Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49412115-49412623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCTA
Entrez Gene ID
6988
Gene Name
T-cell leukemia translocation altered
Gene Aliases
-
UniGene
Hs.517962
Gene Chromosome Location
Chr.3: 49412206-49416476 on Build GRCh38
Transcripts
NM_022171.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3595273 c.117C>T p.F39F Chr.3: 49412543-49412543 on Build GRCh38
3595272 c.81G>A p.R27R Chr.3: 49412507-49412507 on Build GRCh38
4610139 c.130C>G p.L44V Chr.3: 49412556-49412556 on Build GRCh38
5671773 c.170delG p.F59fs*6 Chr.3: 49412596-49412596 on Build GRCh38
4940105 c.30G>T p.L10F Chr.3: 49412456-49412456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772580835 Chr.3: 49412556-49412557 on Build GRCh38
rs773813045 Chr.3: 49412237-49412237 on Build GRCh38
rs759191076 Chr.3: 49412379-49412379 on Build GRCh38
rs764544359 Chr.3: 49412556-49412556 on Build GRCh38
rs756563225 Chr.3: 49412486-49412486 on Build GRCh38
rs780173259 Chr.3: 49412576-49412576 on Build GRCh38
rs868617938 Chr.3: 49412348-49412348 on Build GRCh38
rs776147858 Chr.3: 49412554-49412554 on Build GRCh38
rs779029527 Chr.3: 49412505-49412505 on Build GRCh38
rs756360985 Chr.3: 49412420-49412420 on Build GRCh38
rs746935879 Chr.3: 49412577-49412577 on Build GRCh38
rs764788956 Chr.3: 49412466-49412466 on Build GRCh38
rs772735343 Chr.3: 49412547-49412547 on Build GRCh38
rs976356038 Chr.3: 49412281-49412281 on Build GRCh38
rs576415237 Chr.3: 49412375-49412375 on Build GRCh38
rs765410971 Chr.3: 49412565-49412565 on Build GRCh38
rs746839681 Chr.3: 49412446-49412446 on Build GRCh38
rs530630431 Chr.3: 49412247-49412247 on Build GRCh38
rs775236156 Chr.3: 49412437-49412437 on Build GRCh38
rs903269329 Chr.3: 49412298-49412298 on Build GRCh38
rs757470686 Chr.3: 49412562-49412562 on Build GRCh38
rs918269749 Chr.3: 49412356-49412356 on Build GRCh38
rs779979564 Chr.3: 49412522-49412522 on Build GRCh38
rs530311756 Chr.3: 49412214-49412214 on Build GRCh38
rs750468556 Chr.3: 49412567-49412567 on Build GRCh38
rs117693529 Chr.3: 49412368-49412368 on Build GRCh38
rs940045 Chr.3: 49412205-49412205 on Build GRCh38
rs558648115 Chr.3: 49412408-49412408 on Build GRCh38
rs761376273 Chr.3: 49412422-49412423 on Build GRCh38
rs1012106343 Chr.3: 49412404-49412404 on Build GRCh38
rs762389867 Chr.3: 49412553-49412553 on Build GRCh38
rs763246567 Chr.3: 49412238-49412238 on Build GRCh38
rs994519572 Chr.3: 49412216-49412216 on Build GRCh38
rs745805193 Chr.3: 49412429-49412429 on Build GRCh38
rs768238869 Chr.3: 49412448-49412448 on Build GRCh38
rs146792880 Chr.3: 49412496-49412496 on Build GRCh38
rs770428371 Chr.3: 49412217-49412217 on Build GRCh38
rs768435275 Chr.3: 49412532-49412532 on Build GRCh38
rs764746241 Chr.3: 49412545-49412546 on Build GRCh38
rs140184270 Chr.3: 49412492-49412492 on Build GRCh38
rs768166136 Chr.3: 49412476-49412476 on Build GRCh38
rs1056084334 Chr.3: 49412260-49412260 on Build GRCh38
rs753295631 Chr.3: 49412398-49412398 on Build GRCh38
rs747923015 Chr.3: 49412596-49412596 on Build GRCh38
rs1045634749 Chr.3: 49412367-49412367 on Build GRCh38
rs1029561919 Chr.3: 49412196-49412196 on Build GRCh38
rs776267933 Chr.3: 49412452-49412452 on Build GRCh38
rs769368638 Chr.3: 49412538-49412538 on Build GRCh38
rs765182068 Chr.3: 49412218-49412218 on Build GRCh38
rs941141730 Chr.3: 49412321-49412321 on Build GRCh38
rs775322471 Chr.3: 49412393-49412393 on Build GRCh38
rs774544348 Chr.3: 49412277-49412277 on Build GRCh38
rs747811502 Chr.3: 49412537-49412537 on Build GRCh38
rs534355000 Chr.3: 49412312-49412312 on Build GRCh38
rs745977657 Chr.3: 49412517-49412517 on Build GRCh38
rs892964879 Chr.3: 49412403-49412403 on Build GRCh38
rs569607483 Chr.3: 49412241-49412241 on Build GRCh38
rs563928830 Chr.3: 49412480-49412480 on Build GRCh38
rs753149286 Chr.3: 49412418-49412418 on Build GRCh38
rs771897675 Chr.3: 49412391-49412391 on Build GRCh38
rs772642481 Chr.3: 49412467-49412467 on Build GRCh38
rs758317433 Chr.3: 49412518-49412518 on Build GRCh38
rs140484970 Chr.3: 49412588-49412588 on Build GRCh38
rs996753446 Chr.3: 49412183-49412183 on Build GRCh38
rs548838140 Chr.3: 49412232-49412232 on Build GRCh38
rs752673050 Chr.3: 49412314-49412314 on Build GRCh38
rs1030005958 Chr.3: 49412361-49412361 on Build GRCh38
rs139004176 Chr.3: 49412252-49412252 on Build GRCh38
rs199582584 Chr.3: 49412500-49412500 on Build GRCh38
rs776467205 Chr.3: 49412536-49412536 on Build GRCh38
rs535262167 Chr.3: 49412366-49412366 on Build GRCh38
rs955243735 Chr.3: 49412200-49412200 on Build GRCh38
rs758475959 Chr.3: 49412573-49412573 on Build GRCh38
rs963937809 Chr.3: 49412173-49412173 on Build GRCh38
rs950109717 Chr.3: 49412284-49412284 on Build GRCh38
rs896810896 Chr.3: 49412215-49412215 on Build GRCh38
rs907062207 Chr.3: 49412227-49412227 on Build GRCh38
rs996621113 Chr.3: 49412359-49412359 on Build GRCh38
rs1054462131 Chr.3: 49412331-49412331 on Build GRCh38
rs76667473 Chr.3: 49412209-49412209 on Build GRCh38
rs1020351456 Chr.3: 49412406-49412406 on Build GRCh38
rs12488416 Chr.3: 49412461-49412461 on Build GRCh38
rs1004287291 Chr.3: 49412382-49412382 on Build GRCh38
rs565287211 Chr.3: 49412180-49412180 on Build GRCh38
rs932120229 Chr.3: 49412165-49412165 on Build GRCh38
rs957647537 Chr.3: 49412242-49412242 on Build GRCh38
rs923494319 Chr.3: 49412290-49412290 on Build GRCh38
rs967802036 Chr.3: 49412415-49412415 on Build GRCh38
rs1038109766 Chr.3: 49412344-49412344 on Build GRCh38
rs577154770 Chr.3: 49412409-49412409 on Build GRCh38
rs953539508 Chr.3: 49412291-49412291 on Build GRCh38
rs115911627 Chr.3: 49412426-49412426 on Build GRCh38
rs1038482905 Chr.3: 49412212-49412212 on Build GRCh38
rs371507539 Chr.3: 49412399-49412399 on Build GRCh38
rs764350015 Chr.3: 49412490-49412490 on Build GRCh38
rs754205315 Chr.3: 49412557-49412557 on Build GRCh38
rs754956265 Chr.3: 49412587-49412587 on Build GRCh38
rs749325066 Chr.3: 49412423-49412423 on Build GRCh38
rs761335887 Chr.3: 49412555-49412555 on Build GRCh38
rs778165868 Chr.3: 49412421-49412421 on Build GRCh38
rs867069542 Chr.3: 49412199-49412199 on Build GRCh38
rs375625597 Chr.3: 49412417-49412417 on Build GRCh38
rs560125749 Chr.3: 49412210-49412210 on Build GRCh38
rs760300241 Chr.3: 49412395-49412395 on Build GRCh38
rs552828302 Chr.3: 49412323-49412323 on Build GRCh38
rs575217400 Chr.3: 49412431-49412431 on Build GRCh38
rs760055221 Chr.3: 49412473-49412473 on Build GRCh38
rs568061425 Chr.3: 49412357-49412357 on Build GRCh38
rs564052717 Chr.3: 49412524-49412524 on Build GRCh38
rs570442136 Chr.3: 49412254-49412254 on Build GRCh38
rs1024156581 Chr.3: 49412222-49412222 on Build GRCh38
rs920794940 Chr.3: 49412239-49412239 on Build GRCh38
rs1000203507 Chr.3: 49412305-49412305 on Build GRCh38
rs1049523278 Chr.3: 49412179-49412179 on Build GRCh38

View Full Product Details