Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50296310-50296799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYAL3
Entrez Gene ID
8372
Gene Name
hyaluronoglucosaminidase 3
Gene Aliases
HYAL-3, LUCA-3, LUCA3
UniGene
Hs.129910
Gene Chromosome Location
Chr.3: 50292828-50299468 on Build GRCh38
Transcripts
NM_001200029.1 NM_001200030.1 NM_001200031.1 NM_001200032.1 NM_003549.3 NM_001200016.1 NM_001200018.1 NM_012191.3

Gene Symbol
NAT6
Entrez Gene ID
24142
Gene Name
N-acetyltransferase 6
Gene Aliases
FUS-2, FUS2
UniGene
Hs.129910
Gene Chromosome Location
Chr.3: 50296402-50299421 on Build GRCh38
Transcripts
NM_001200029.1 NM_001200030.1 NM_001200031.1 NM_001200032.1 NM_003549.3 NM_001200016.1 NM_001200018.1 NM_012191.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2851254 c.742A>T p.N248Y Chr.3: 50296788-50296788 on Build GRCh38
4584484 c.924C>G p.I308M Chr.3: 50296606-50296606 on Build GRCh38
1424156 c.782C>A p.P261H Chr.3: 50296748-50296748 on Build GRCh38
6896049 c.892G>T p.G298W Chr.3: 50296638-50296638 on Build GRCh38
1424155 c.851C>A p.P284H Chr.3: 50296679-50296679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200163623 Chr.3: 50296554-50296555 on Build GRCh38
rs782070086 Chr.3: 50296345-50296345 on Build GRCh38
rs370500153 Chr.3: 50296546-50296546 on Build GRCh38
rs990717436 Chr.3: 50296555-50296555 on Build GRCh38
rs1002895615 Chr.3: 50296517-50296517 on Build GRCh38
rs371233677 Chr.3: 50296682-50296682 on Build GRCh38
rs370006421 Chr.3: 50296620-50296620 on Build GRCh38
rs958693797 Chr.3: 50296776-50296776 on Build GRCh38
rs782807592 Chr.3: 50296567-50296567 on Build GRCh38
rs373888358 Chr.3: 50296623-50296623 on Build GRCh38
rs782557024 Chr.3: 50296618-50296618 on Build GRCh38
rs782250530 Chr.3: 50296730-50296730 on Build GRCh38
rs782189821 Chr.3: 50296520-50296520 on Build GRCh38
rs74426485 Chr.3: 50296635-50296635 on Build GRCh38
rs368362937 Chr.3: 50296724-50296724 on Build GRCh38
rs377507869 Chr.3: 50296637-50296637 on Build GRCh38
rs782098242 Chr.3: 50296646-50296646 on Build GRCh38
rs782066929 Chr.3: 50296572-50296572 on Build GRCh38
rs782212075 Chr.3: 50296713-50296713 on Build GRCh38
rs782820007 Chr.3: 50296660-50296660 on Build GRCh38
rs960626077 Chr.3: 50296545-50296545 on Build GRCh38
rs370663191 Chr.3: 50296650-50296650 on Build GRCh38
rs372686842 Chr.3: 50296663-50296663 on Build GRCh38
rs781962043 Chr.3: 50296768-50296768 on Build GRCh38
rs782626712 Chr.3: 50296474-50296474 on Build GRCh38
rs782360508 Chr.3: 50296556-50296556 on Build GRCh38
rs782089165 Chr.3: 50296553-50296553 on Build GRCh38
rs1047241281 Chr.3: 50296669-50296669 on Build GRCh38
rs374067234 Chr.3: 50296734-50296734 on Build GRCh38
rs781888148 Chr.3: 50296606-50296606 on Build GRCh38
rs996074708 Chr.3: 50296475-50296475 on Build GRCh38
rs782650389 Chr.3: 50296699-50296699 on Build GRCh38
rs782053569 Chr.3: 50296744-50296744 on Build GRCh38
rs34185226 Chr.3: 50296734-50296735 on Build GRCh38
rs587703549 Chr.3: 50296680-50296680 on Build GRCh38
rs192428304 Chr.3: 50296668-50296668 on Build GRCh38
rs923716071 Chr.3: 50296609-50296609 on Build GRCh38
rs781870766 Chr.3: 50296654-50296654 on Build GRCh38
rs782571527 Chr.3: 50296619-50296619 on Build GRCh38
rs781836755 Chr.3: 50296589-50296589 on Build GRCh38
rs587768945 Chr.3: 50296344-50296344 on Build GRCh38
rs782740787 Chr.3: 50296576-50296576 on Build GRCh38
rs782427011 Chr.3: 50296701-50296701 on Build GRCh38
rs782513639 Chr.3: 50296457-50296457 on Build GRCh38
rs782664262 Chr.3: 50296634-50296634 on Build GRCh38
rs782511363 Chr.3: 50296594-50296594 on Build GRCh38
rs587637246 Chr.3: 50296384-50296384 on Build GRCh38
rs368118572 Chr.3: 50296735-50296735 on Build GRCh38
rs782709893 Chr.3: 50296746-50296746 on Build GRCh38
rs372053533 Chr.3: 50296773-50296773 on Build GRCh38
rs587724778 Chr.3: 50296386-50296386 on Build GRCh38
rs374208572 Chr.3: 50296582-50296582 on Build GRCh38
rs201771785 Chr.3: 50296685-50296685 on Build GRCh38
rs200295394 Chr.3: 50296731-50296731 on Build GRCh38
rs782643393 Chr.3: 50296604-50296604 on Build GRCh38
rs781785577 Chr.3: 50296569-50296569 on Build GRCh38
rs1035668942 Chr.3: 50296536-50296536 on Build GRCh38
rs781996881 Chr.3: 50296558-50296558 on Build GRCh38
rs782030593 Chr.3: 50296737-50296737 on Build GRCh38
rs771193453 Chr.3: 50296387-50296387 on Build GRCh38
rs201246355 Chr.3: 50296633-50296633 on Build GRCh38
rs782312544 Chr.3: 50296374-50296374 on Build GRCh38
rs782146142 Chr.3: 50296658-50296658 on Build GRCh38
rs782633160 Chr.3: 50296631-50296631 on Build GRCh38
rs1025759737 Chr.3: 50296483-50296483 on Build GRCh38
rs1013329791 Chr.3: 50296779-50296779 on Build GRCh38
rs202131471 Chr.3: 50296565-50296565 on Build GRCh38
rs781974605 Chr.3: 50296485-50296485 on Build GRCh38
rs951428261 Chr.3: 50296503-50296503 on Build GRCh38

View Full Product Details