Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53888288-53888734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SELK
Entrez Gene ID
58515
Gene Name
selenoprotein K
Gene Aliases
HSPC030, HSPC297, SelK
UniGene
Hs.58471
Gene Chromosome Location
Chr.3: 53885199-53891962 on Build GRCh38
Transcripts
NM_021237.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1047347 c.78C>A p.F26L Chr.3: 53888425-53888425 on Build GRCh38
1047346 c.81C>A p.F27L Chr.3: 53888422-53888422 on Build GRCh38
337862 c.22C>T p.Q8* Chr.3: 53888481-53888481 on Build GRCh38
7447415 c.110+2T>C p.? Chr.3: 53888391-53888391 on Build GRCh38
1665982 c.110+1G>T p.? Chr.3: 53888392-53888392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965955368 Chr.3: 53888588-53888588 on Build GRCh38
rs757104638 Chr.3: 53888355-53888355 on Build GRCh38
rs759309127 Chr.3: 53888354-53888354 on Build GRCh38
rs753906390 Chr.3: 53888511-53888511 on Build GRCh38
rs977256755 Chr.3: 53888346-53888346 on Build GRCh38
rs3816144 Chr.3: 53888593-53888593 on Build GRCh38
rs376258471 Chr.3: 53888332-53888332 on Build GRCh38
rs983270388 Chr.3: 53888687-53888687 on Build GRCh38
rs751533368 Chr.3: 53888478-53888478 on Build GRCh38
rs774923314 Chr.3: 53888466-53888466 on Build GRCh38
rs749802486 Chr.3: 53888385-53888385 on Build GRCh38
rs528689867 Chr.3: 53888509-53888509 on Build GRCh38
rs560821192 Chr.3: 53888556-53888556 on Build GRCh38
rs375950292 Chr.3: 53888477-53888477 on Build GRCh38
rs779481266 Chr.3: 53888400-53888400 on Build GRCh38
rs759250099 Chr.3: 53888428-53888428 on Build GRCh38
rs953263155 Chr.3: 53888664-53888664 on Build GRCh38
rs750673209 Chr.3: 53888505-53888505 on Build GRCh38
rs115392613 Chr.3: 53888571-53888571 on Build GRCh38
rs922687230 Chr.3: 53888462-53888462 on Build GRCh38
rs1041038866 Chr.3: 53888430-53888430 on Build GRCh38
rs546499317 Chr.3: 53888427-53888427 on Build GRCh38
rs748424200 Chr.3: 53888402-53888402 on Build GRCh38
rs35258834 Chr.3: 53888567-53888568 on Build GRCh38
rs1017448436 Chr.3: 53888576-53888576 on Build GRCh38
rs780352601 Chr.3: 53888367-53888367 on Build GRCh38
rs772508566 Chr.3: 53888404-53888404 on Build GRCh38
rs930028757 Chr.3: 53888654-53888654 on Build GRCh38
rs569280980 Chr.3: 53888648-53888648 on Build GRCh38
rs756234259 Chr.3: 53888365-53888365 on Build GRCh38
rs771231767 Chr.3: 53888319-53888319 on Build GRCh38
rs1005761424 Chr.3: 53888487-53888487 on Build GRCh38
rs762528129 Chr.3: 53888474-53888474 on Build GRCh38
rs942692368 Chr.3: 53888407-53888407 on Build GRCh38
rs986377187 Chr.3: 53888358-53888358 on Build GRCh38
rs947315586 Chr.3: 53888382-53888382 on Build GRCh38
rs371839775 Chr.3: 53888527-53888527 on Build GRCh38
rs957184778 Chr.3: 53888567-53888567 on Build GRCh38
rs986001406 Chr.3: 53888388-53888388 on Build GRCh38
rs894757912 Chr.3: 53888515-53888515 on Build GRCh38
rs201843824 Chr.3: 53888392-53888392 on Build GRCh38
rs767457982 Chr.3: 53888502-53888502 on Build GRCh38
rs1032795600 Chr.3: 53888564-53888564 on Build GRCh38
rs781218366 Chr.3: 53888362-53888362 on Build GRCh38
rs182816092 Chr.3: 53888340-53888340 on Build GRCh38
rs760524250 Chr.3: 53888660-53888660 on Build GRCh38
rs751243531 Chr.3: 53888347-53888347 on Build GRCh38
rs547667658 Chr.3: 53888649-53888649 on Build GRCh38
rs186628092 Chr.3: 53888465-53888465 on Build GRCh38
rs372300406 Chr.3: 53888364-53888364 on Build GRCh38
rs774745266 Chr.3: 53888383-53888383 on Build GRCh38
rs999016592 Chr.3: 53888558-53888558 on Build GRCh38
rs756465009 Chr.3: 53888508-53888508 on Build GRCh38
rs1013899545 Chr.3: 53888530-53888530 on Build GRCh38
rs535217934 Chr.3: 53888568-53888568 on Build GRCh38
rs150968318 Chr.3: 53888544-53888544 on Build GRCh38
rs761488603 Chr.3: 53888481-53888481 on Build GRCh38
rs563667094 Chr.3: 53888348-53888348 on Build GRCh38

View Full Product Details