Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108136499-108137018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A24
Entrez Gene ID
29957
Gene Name
solute carrier family 25 member 24
Gene Aliases
APC1, SCAMC-1
UniGene
Hs.656870
Gene Chromosome Location
Chr.1: 108134722-108200358 on Build GRCh38
Transcripts
NM_013386.4 NM_213651.2 XM_017001104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7196723 c.1315G>T p.G439* Chr.1: 108136772-108136772 on Build GRCh38
4462928 c.1210C>T p.P404S Chr.1: 108136820-108136820 on Build GRCh38
1736965 c.1265C>T p.S422F Chr.1: 108136822-108136822 on Build GRCh38
5509704 c.1250-10delT p.? Chr.1: 108136847-108136847 on Build GRCh38
5509705 c.1193-10delT p.? Chr.1: 108136847-108136847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756247539 Chr.1: 108136722-108136722 on Build GRCh38
rs760344943 Chr.1: 108136787-108136787 on Build GRCh38
rs765947046 Chr.1: 108136663-108136663 on Build GRCh38
rs149670605 Chr.1: 108136789-108136789 on Build GRCh38
rs144420834 Chr.1: 108136792-108136792 on Build GRCh38
rs754957381 Chr.1: 108136691-108136691 on Build GRCh38
rs560890849 Chr.1: 108136805-108136805 on Build GRCh38
rs772520980 Chr.1: 108136829-108136829 on Build GRCh38
rs910734681 Chr.1: 108136983-108136983 on Build GRCh38
rs919697593 Chr.1: 108136556-108136556 on Build GRCh38
rs952497421 Chr.1: 108136945-108136945 on Build GRCh38
rs773526548 Chr.1: 108136785-108136785 on Build GRCh38
rs746053129 Chr.1: 108136706-108136706 on Build GRCh38
rs904934130 Chr.1: 108136555-108136555 on Build GRCh38
rs765813497 Chr.1: 108136846-108136847 on Build GRCh38
rs572466613 Chr.1: 108136584-108136584 on Build GRCh38
rs765493447 Chr.1: 108136675-108136675 on Build GRCh38
rs985590733 Chr.1: 108136976-108136976 on Build GRCh38
rs763003506 Chr.1: 108136878-108136878 on Build GRCh38
rs1009766940 Chr.1: 108136608-108136608 on Build GRCh38
rs367963175 Chr.1: 108136862-108136862 on Build GRCh38
rs763334679 Chr.1: 108136883-108136885 on Build GRCh38
rs41278470 Chr.1: 108136621-108136621 on Build GRCh38
rs923514128 Chr.1: 108136636-108136636 on Build GRCh38
rs774716010 Chr.1: 108136751-108136751 on Build GRCh38
rs200970220 Chr.1: 108136628-108136628 on Build GRCh38
rs780069314 Chr.1: 108136814-108136814 on Build GRCh38
rs747271413 Chr.1: 108136835-108136835 on Build GRCh38
rs772402918 Chr.1: 108136771-108136771 on Build GRCh38
rs760226357 Chr.1: 108136651-108136651 on Build GRCh38
rs375592750 Chr.1: 108136808-108136808 on Build GRCh38
rs1042983976 Chr.1: 108136938-108136938 on Build GRCh38
rs749830114 Chr.1: 108136609-108136609 on Build GRCh38
rs890637164 Chr.1: 108136946-108136946 on Build GRCh38
rs758863591 Chr.1: 108136926-108136926 on Build GRCh38
rs41278468 Chr.1: 108136612-108136612 on Build GRCh38
rs781617889 Chr.1: 108136701-108136701 on Build GRCh38
rs972377992 Chr.1: 108136546-108136546 on Build GRCh38
rs758258978 Chr.1: 108136700-108136700 on Build GRCh38
rs1054966655 Chr.1: 108136776-108136776 on Build GRCh38
rs770685399 Chr.1: 108136846-108136846 on Build GRCh38
rs775096775 Chr.1: 108136874-108136874 on Build GRCh38
rs775899730 Chr.1: 108136602-108136602 on Build GRCh38
rs973434663 Chr.1: 108136676-108136676 on Build GRCh38
rs544018515 Chr.1: 108136693-108136693 on Build GRCh38
rs575093838 Chr.1: 108136521-108136521 on Build GRCh38
rs982059147 Chr.1: 108136836-108136836 on Build GRCh38
rs377138462 Chr.1: 108136698-108136698 on Build GRCh38
rs749266397 Chr.1: 108136741-108136741 on Build GRCh38
rs17021096 Chr.1: 108136529-108136529 on Build GRCh38
rs200257541 Chr.1: 108136815-108136815 on Build GRCh38
rs375761402 Chr.1: 108136684-108136684 on Build GRCh38
rs376157270 Chr.1: 108136793-108136793 on Build GRCh38
rs1009521837 Chr.1: 108136982-108136982 on Build GRCh38
rs762097430 Chr.1: 108136626-108136626 on Build GRCh38
rs952566550 Chr.1: 108136581-108136581 on Build GRCh38
rs1001320483 Chr.1: 108136753-108136753 on Build GRCh38
rs772835903 Chr.1: 108136639-108136639 on Build GRCh38
rs552936936 Chr.1: 108136536-108136536 on Build GRCh38
rs778125454 Chr.1: 108136833-108136833 on Build GRCh38
rs937710236 Chr.1: 108136571-108136571 on Build GRCh38
rs748568887 Chr.1: 108136755-108136755 on Build GRCh38
rs17513812 Chr.1: 108136910-108136910 on Build GRCh38
rs544745749 Chr.1: 108136585-108136585 on Build GRCh38
rs748622050 Chr.1: 108136825-108136825 on Build GRCh38
rs779443602 Chr.1: 108136817-108136817 on Build GRCh38
rs964682263 Chr.1: 108136654-108136654 on Build GRCh38
rs372726918 Chr.1: 108136790-108136790 on Build GRCh38
rs769210704 Chr.1: 108136749-108136749 on Build GRCh38
rs560573928 Chr.1: 108136996-108136996 on Build GRCh38
rs199598099 Chr.1: 108136860-108136860 on Build GRCh38
rs1053504822 Chr.1: 108136909-108136909 on Build GRCh38

View Full Product Details