Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109625891-109626410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD2
Entrez Gene ID
271
Gene Name
adenosine monophosphate deaminase 2
Gene Aliases
PCH9, SPG63
UniGene
Hs.82927
Gene Chromosome Location
Chr.1: 109619813-109632055 on Build GRCh38
Transcripts
NM_001257360.1 NM_001257361.1 NM_001308170.1 NM_004037.7 NM_139156.3 NM_203404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5177048 c.255delG p.G86fs*158 Chr.1: 109626346-109626346 on Build GRCh38
6057053 c.247G>T p.E83* Chr.1: 109626338-109626338 on Build GRCh38
4661430 c.629G>A p.R210Q Chr.1: 109626363-109626363 on Build GRCh38
4020397 c.538C>T p.R180W Chr.1: 109626182-109626182 on Build GRCh38
1320159 c.601G>T p.G201C Chr.1: 109626335-109626335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752716360 Chr.1: 109626219-109626219 on Build GRCh38
rs1047829882 Chr.1: 109626096-109626096 on Build GRCh38
rs1045549669 Chr.1: 109626153-109626153 on Build GRCh38
rs952574498 Chr.1: 109626016-109626029 on Build GRCh38
rs930354267 Chr.1: 109626091-109626091 on Build GRCh38
rs909031529 Chr.1: 109625972-109625972 on Build GRCh38
rs771249006 Chr.1: 109626145-109626145 on Build GRCh38
rs921369081 Chr.1: 109625967-109625967 on Build GRCh38
rs200906971 Chr.1: 109626123-109626123 on Build GRCh38
rs1038777242 Chr.1: 109626075-109626075 on Build GRCh38
rs138175138 Chr.1: 109626384-109626384 on Build GRCh38
rs868501220 Chr.1: 109625978-109625978 on Build GRCh38
rs763135190 Chr.1: 109626220-109626220 on Build GRCh38
rs758231653 Chr.1: 109626351-109626351 on Build GRCh38
rs776115972 Chr.1: 109626289-109626289 on Build GRCh38
rs571185024 Chr.1: 109625959-109625959 on Build GRCh38
rs182256435 Chr.1: 109625918-109625918 on Build GRCh38
rs1001539693 Chr.1: 109625956-109625956 on Build GRCh38
rs774472908 Chr.1: 109626083-109626083 on Build GRCh38
rs771729287 Chr.1: 109626282-109626282 on Build GRCh38
rs373334637 Chr.1: 109626353-109626353 on Build GRCh38
rs553616355 Chr.1: 109626254-109626254 on Build GRCh38
rs774088979 Chr.1: 109626131-109626131 on Build GRCh38
rs199618590 Chr.1: 109626272-109626272 on Build GRCh38
rs765794204 Chr.1: 109626182-109626182 on Build GRCh38
rs114639310 Chr.1: 109626117-109626117 on Build GRCh38
rs550959557 Chr.1: 109625935-109625935 on Build GRCh38
rs770959153 Chr.1: 109626032-109626032 on Build GRCh38
rs369853440 Chr.1: 109626249-109626249 on Build GRCh38
rs775750465 Chr.1: 109626373-109626373 on Build GRCh38
rs575582124 Chr.1: 109626324-109626324 on Build GRCh38
rs551776531 Chr.1: 109626001-109626001 on Build GRCh38
rs766200601 Chr.1: 109626326-109626326 on Build GRCh38
rs951956085 Chr.1: 109626202-109626202 on Build GRCh38
rs774577364 Chr.1: 109626311-109626311 on Build GRCh38
rs754879932 Chr.1: 109626340-109626340 on Build GRCh38
rs753757082 Chr.1: 109626329-109626329 on Build GRCh38
rs757584392 Chr.1: 109626242-109626242 on Build GRCh38
rs943100302 Chr.1: 109626049-109626049 on Build GRCh38
rs999158342 Chr.1: 109626116-109626116 on Build GRCh38
rs368102578 Chr.1: 109626376-109626376 on Build GRCh38
rs957674325 Chr.1: 109626294-109626294 on Build GRCh38
rs777443721 Chr.1: 109626363-109626363 on Build GRCh38
rs764193360 Chr.1: 109626310-109626310 on Build GRCh38
rs984263188 Chr.1: 109626227-109626227 on Build GRCh38
rs955796743 Chr.1: 109626000-109626000 on Build GRCh38
rs756105491 Chr.1: 109626251-109626251 on Build GRCh38
rs928184387 Chr.1: 109626009-109626009 on Build GRCh38
rs764200213 Chr.1: 109626223-109626223 on Build GRCh38
rs762067648 Chr.1: 109626313-109626313 on Build GRCh38
rs768185545 Chr.1: 109626124-109626124 on Build GRCh38
rs746212106 Chr.1: 109626364-109626364 on Build GRCh38
rs955686512 Chr.1: 109626362-109626362 on Build GRCh38
rs776988264 Chr.1: 109626147-109626147 on Build GRCh38
rs970149816 Chr.1: 109625926-109625926 on Build GRCh38
rs779371189 Chr.1: 109626118-109626118 on Build GRCh38
rs749420176 Chr.1: 109626025-109626025 on Build GRCh38
rs920269042 Chr.1: 109626076-109626076 on Build GRCh38
rs536798841 Chr.1: 109626164-109626164 on Build GRCh38
rs867331549 Chr.1: 109626128-109626128 on Build GRCh38
rs768258207 Chr.1: 109626259-109626259 on Build GRCh38
rs750235935 Chr.1: 109626321-109626321 on Build GRCh38
rs493972 Chr.1: 109626266-109626266 on Build GRCh38
rs150418456 Chr.1: 109626383-109626383 on Build GRCh38
rs957371314 Chr.1: 109625970-109625970 on Build GRCh38
rs112705694 Chr.1: 109626299-109626299 on Build GRCh38
rs200750655 Chr.1: 109626221-109626221 on Build GRCh38
rs1010263262 Chr.1: 109626309-109626309 on Build GRCh38
rs750254503 Chr.1: 109626250-109626250 on Build GRCh38
rs770170283 Chr.1: 109626287-109626287 on Build GRCh38
rs767908314 Chr.1: 109626315-109626315 on Build GRCh38
rs372087784 Chr.1: 109626377-109626377 on Build GRCh38
rs902210887 Chr.1: 109626105-109626105 on Build GRCh38
rs544609345 Chr.1: 109626368-109626368 on Build GRCh38
rs373313669 Chr.1: 109626258-109626258 on Build GRCh38
rs751756777 Chr.1: 109626236-109626236 on Build GRCh38
rs913514687 Chr.1: 109625989-109625989 on Build GRCh38
rs946516316 Chr.1: 109626070-109626070 on Build GRCh38
rs201869959 Chr.1: 109626283-109626283 on Build GRCh38
rs894900638 Chr.1: 109626039-109626039 on Build GRCh38
rs1043847124 Chr.1: 109626090-109626090 on Build GRCh38
rs972990702 Chr.1: 109626008-109626008 on Build GRCh38
rs761001167 Chr.1: 109626142-109626142 on Build GRCh38

View Full Product Details