Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57560183-57560729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE12
Entrez Gene ID
201626
Gene Name
phosphodiesterase 12
Gene Aliases
2'-PDE, 2-PDE
UniGene
Hs.572993
Gene Chromosome Location
Chr.3: 57556247-57656480 on Build GRCh38
Transcripts
NM_001322176.1 NM_001322177.1 NM_177966.6

SNP Detail

SNP ID Chromosome Location
rs758672373 Chr.3: 57560450-57560450 on Build GRCh38
rs963611054 Chr.3: 57560444-57560444 on Build GRCh38
rs529715446 Chr.3: 57560300-57560300 on Build GRCh38
rs59592393 Chr.3: 57560450-57560451 on Build GRCh38
rs778201357 Chr.3: 57560252-57560255 on Build GRCh38
rs1038714008 Chr.3: 57560449-57560449 on Build GRCh38
rs980123176 Chr.3: 57560303-57560306 on Build GRCh38
rs902706535 Chr.3: 57560548-57560548 on Build GRCh38
rs141300932 Chr.3: 57560306-57560309 on Build GRCh38
rs552031796 Chr.3: 57560400-57560400 on Build GRCh38
rs150853169 Chr.3: 57560338-57560338 on Build GRCh38
rs748894438 Chr.3: 57560660-57560660 on Build GRCh38
rs939823627 Chr.3: 57560692-57560692 on Build GRCh38
rs776853522 Chr.3: 57560298-57560298 on Build GRCh38
rs532901949 Chr.3: 57560410-57560410 on Build GRCh38
rs1046076960 Chr.3: 57560227-57560227 on Build GRCh38
rs534551274 Chr.3: 57560451-57560451 on Build GRCh38
rs141489470 Chr.3: 57560222-57560222 on Build GRCh38
rs549859645 Chr.3: 57560317-57560317 on Build GRCh38
rs765179643 Chr.3: 57560314-57560325 on Build GRCh38
rs1039159022 Chr.3: 57560307-57560307 on Build GRCh38
rs192798402 Chr.3: 57560522-57560522 on Build GRCh38
rs1004831400 Chr.3: 57560609-57560609 on Build GRCh38
rs1005052406 Chr.3: 57560418-57560418 on Build GRCh38
rs538414065 Chr.3: 57560381-57560381 on Build GRCh38
rs566577608 Chr.3: 57560577-57560577 on Build GRCh38
rs1038032845 Chr.3: 57560265-57560265 on Build GRCh38
rs955120377 Chr.3: 57560292-57560292 on Build GRCh38
rs925290057 Chr.3: 57560305-57560306 on Build GRCh38
rs1004576456 Chr.3: 57560258-57560258 on Build GRCh38
rs1016851909 Chr.3: 57560435-57560435 on Build GRCh38
rs915141107 Chr.3: 57560523-57560523 on Build GRCh38
rs990849996 Chr.3: 57560314-57560319 on Build GRCh38
rs959612611 Chr.3: 57560313-57560314 on Build GRCh38
rs144212569 Chr.3: 57560482-57560482 on Build GRCh38
rs996588367 Chr.3: 57560268-57560268 on Build GRCh38
rs7618684 Chr.3: 57560266-57560266 on Build GRCh38
rs886407666 Chr.3: 57560605-57560605 on Build GRCh38
rs981237651 Chr.3: 57560375-57560375 on Build GRCh38
rs935213619 Chr.3: 57560274-57560274 on Build GRCh38
rs1001059860 Chr.3: 57560551-57560551 on Build GRCh38
rs923564584 Chr.3: 57560485-57560485 on Build GRCh38
rs576041338 Chr.3: 57560557-57560557 on Build GRCh38
rs755741714 Chr.3: 57560574-57560574 on Build GRCh38
rs565125728 Chr.3: 57560647-57560647 on Build GRCh38
rs1032978272 Chr.3: 57560556-57560556 on Build GRCh38
rs899392625 Chr.3: 57560632-57560632 on Build GRCh38
rs970451374 Chr.3: 57560474-57560474 on Build GRCh38
rs753060450 Chr.3: 57560411-57560411 on Build GRCh38
rs950986193 Chr.3: 57560414-57560414 on Build GRCh38
rs146506134 Chr.3: 57560668-57560668 on Build GRCh38
rs544787037 Chr.3: 57560616-57560616 on Build GRCh38
rs752375915 Chr.3: 57560320-57560325 on Build GRCh38
rs991190956 Chr.3: 57560596-57560596 on Build GRCh38
rs1030258974 Chr.3: 57560386-57560386 on Build GRCh38
rs556207572 Chr.3: 57560528-57560528 on Build GRCh38
rs900608816 Chr.3: 57560570-57560570 on Build GRCh38
rs571834364 Chr.3: 57560412-57560412 on Build GRCh38
rs766435276 Chr.3: 57560297-57560297 on Build GRCh38
rs1028465030 Chr.3: 57560273-57560274 on Build GRCh38
rs1016553448 Chr.3: 57560617-57560617 on Build GRCh38
rs917967113 Chr.3: 57560326-57560329 on Build GRCh38
rs1979074 Chr.3: 57560471-57560471 on Build GRCh38
rs942573209 Chr.3: 57560527-57560527 on Build GRCh38
rs935246351 Chr.3: 57560487-57560487 on Build GRCh38

View Full Product Details