Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 64561436-64561954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS9
Entrez Gene ID
56999
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 9
Gene Aliases
-
UniGene
Hs.656071
Gene Chromosome Location
Chr.3: 64515654-64688000 on Build GRCh38
Transcripts
NM_001318781.1 NM_182920.1 NR_110150.1 NR_110151.1

Gene Symbol
ADAMTS9-AS1
Entrez Gene ID
101929335
Gene Name
ADAMTS9 antisense RNA 1
Gene Aliases
-
UniGene
Hs.476604
Gene Chromosome Location
Chr.3: 64561346-64590084 on Build GRCh38
Transcripts
NM_001318781.1 NM_182920.1 NR_110150.1 NR_110151.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5810518 c.4634C>A p.S1545* Chr.3: 64561642-64561642 on Build GRCh38
5905527 c.4579G>A p.G1527R Chr.3: 64561697-64561697 on Build GRCh38
22819 c.4605C>T p.T1535T Chr.3: 64561671-64561671 on Build GRCh38
731711 c.4696G>A p.E1566K Chr.3: 64561580-64561580 on Build GRCh38
6011071 c.4680G>A p.R1560R Chr.3: 64561596-64561596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770813155 Chr.3: 64561546-64561546 on Build GRCh38
rs892722430 Chr.3: 64561669-64561669 on Build GRCh38
rs17071048 Chr.3: 64561502-64561502 on Build GRCh38
rs910645601 Chr.3: 64561907-64561907 on Build GRCh38
rs769261684 Chr.3: 64561645-64561645 on Build GRCh38
rs576887535 Chr.3: 64561515-64561515 on Build GRCh38
rs747389868 Chr.3: 64561751-64561754 on Build GRCh38
rs775114165 Chr.3: 64561648-64561648 on Build GRCh38
rs780324244 Chr.3: 64561612-64561612 on Build GRCh38
rs773889358 Chr.3: 64561662-64561662 on Build GRCh38
rs755013991 Chr.3: 64561697-64561697 on Build GRCh38
rs747686401 Chr.3: 64561802-64561802 on Build GRCh38
rs917431401 Chr.3: 64561830-64561830 on Build GRCh38
rs772636677 Chr.3: 64561541-64561541 on Build GRCh38
rs745869372 Chr.3: 64561641-64561641 on Build GRCh38
rs544278182 Chr.3: 64561573-64561573 on Build GRCh38
rs561544201 Chr.3: 64561728-64561728 on Build GRCh38
rs764323037 Chr.3: 64561786-64561786 on Build GRCh38
rs374726166 Chr.3: 64561535-64561535 on Build GRCh38
rs948868718 Chr.3: 64561840-64561840 on Build GRCh38
rs150552517 Chr.3: 64561714-64561714 on Build GRCh38
rs780489993 Chr.3: 64561571-64561574 on Build GRCh38
rs780411856 Chr.3: 64561532-64561532 on Build GRCh38
rs551592736 Chr.3: 64561511-64561512 on Build GRCh38
rs776318441 Chr.3: 64561769-64561770 on Build GRCh38
rs749201678 Chr.3: 64561589-64561589 on Build GRCh38
rs201971866 Chr.3: 64561712-64561712 on Build GRCh38
rs752881125 Chr.3: 64561784-64561784 on Build GRCh38
rs543387674 Chr.3: 64561715-64561715 on Build GRCh38
rs568951629 Chr.3: 64561910-64561910 on Build GRCh38
rs756277187 Chr.3: 64561531-64561531 on Build GRCh38
rs753989691 Chr.3: 64561617-64561617 on Build GRCh38
rs920626098 Chr.3: 64561838-64561838 on Build GRCh38
rs371145408 Chr.3: 64561915-64561915 on Build GRCh38
rs368382773 Chr.3: 64561769-64561769 on Build GRCh38
rs971970632 Chr.3: 64561831-64561831 on Build GRCh38
rs748769698 Chr.3: 64561656-64561656 on Build GRCh38
rs940305058 Chr.3: 64561824-64561825 on Build GRCh38
rs1009760206 Chr.3: 64561713-64561713 on Build GRCh38
rs952031469 Chr.3: 64561846-64561846 on Build GRCh38
rs746749259 Chr.3: 64561621-64561621 on Build GRCh38
rs370056695 Chr.3: 64561572-64561572 on Build GRCh38
rs999835269 Chr.3: 64561618-64561618 on Build GRCh38
rs904292542 Chr.3: 64561853-64561853 on Build GRCh38
rs768061654 Chr.3: 64561674-64561674 on Build GRCh38
rs1034226608 Chr.3: 64561487-64561487 on Build GRCh38
rs759645218 Chr.3: 64561770-64561770 on Build GRCh38
rs748775134 Chr.3: 64561726-64561726 on Build GRCh38
rs762257619 Chr.3: 64561670-64561670 on Build GRCh38
rs80265358 Chr.3: 64561826-64561826 on Build GRCh38
rs752916517 Chr.3: 64561707-64561707 on Build GRCh38
rs573042306 Chr.3: 64561471-64561471 on Build GRCh38
rs761317544 Chr.3: 64561689-64561689 on Build GRCh38
rs143448269 Chr.3: 64561904-64561904 on Build GRCh38
rs377076231 Chr.3: 64561688-64561688 on Build GRCh38
rs761417785 Chr.3: 64561784-64561784 on Build GRCh38
rs565451972 Chr.3: 64561813-64561813 on Build GRCh38
rs780029734 Chr.3: 64561800-64561800 on Build GRCh38
rs755864267 Chr.3: 64561795-64561795 on Build GRCh38
rs372148637 Chr.3: 64561632-64561632 on Build GRCh38
rs201456312 Chr.3: 64561756-64561756 on Build GRCh38
rs758668736 Chr.3: 64561785-64561785 on Build GRCh38
rs751901830 Chr.3: 64561583-64561583 on Build GRCh38
rs750177958 Chr.3: 64561789-64561789 on Build GRCh38
rs868532784 Chr.3: 64561727-64561727 on Build GRCh38
rs768955715 Chr.3: 64561769-64561769 on Build GRCh38
rs768313543 Chr.3: 64561659-64561659 on Build GRCh38
rs762455066 Chr.3: 64561800-64561800 on Build GRCh38
rs755182181 Chr.3: 64561619-64561619 on Build GRCh38
rs374414296 Chr.3: 64561605-64561605 on Build GRCh38
rs765438916 Chr.3: 64561702-64561702 on Build GRCh38
rs964564018 Chr.3: 64561924-64561924 on Build GRCh38
rs1053202803 Chr.3: 64561884-64561884 on Build GRCh38
rs768081992 Chr.3: 64561602-64561602 on Build GRCh38
rs769349136 Chr.3: 64561552-64561552 on Build GRCh38
rs765295809 Chr.3: 64561776-64561776 on Build GRCh38
rs1031267747 Chr.3: 64561642-64561642 on Build GRCh38
rs772653864 Chr.3: 64561799-64561800 on Build GRCh38
rs757038316 Chr.3: 64561708-64561708 on Build GRCh38
rs367778526 Chr.3: 64561477-64561477 on Build GRCh38
rs1040971936 Chr.3: 64561921-64561921 on Build GRCh38
rs143017614 Chr.3: 64561476-64561476 on Build GRCh38
rs931290441 Chr.3: 64561523-64561523 on Build GRCh38
rs779037653 Chr.3: 64561620-64561620 on Build GRCh38
rs377542091 Chr.3: 64561777-64561777 on Build GRCh38
rs979280190 Chr.3: 64561698-64561698 on Build GRCh38
rs769459921 Chr.3: 64561794-64561794 on Build GRCh38
rs756258679 Chr.3: 64561611-64561611 on Build GRCh38
rs766845570 Chr.3: 64561767-64561767 on Build GRCh38
rs750593389 Chr.3: 64561550-64561550 on Build GRCh38
rs764341290 Chr.3: 64561577-64561577 on Build GRCh38
rs774048233 Chr.3: 64561732-64561732 on Build GRCh38
rs1024114836 Chr.3: 64561817-64561817 on Build GRCh38
rs549081938 Chr.3: 64561505-64561505 on Build GRCh38
rs878986582 Chr.3: 64561799-64561799 on Build GRCh38
rs756262621 Chr.3: 64561562-64561562 on Build GRCh38
rs999869631 Chr.3: 64561876-64561876 on Build GRCh38
rs908498409 Chr.3: 64561808-64561808 on Build GRCh38
rs762089819 Chr.3: 64561595-64561595 on Build GRCh38
rs562473718 Chr.3: 64561634-64561634 on Build GRCh38
rs779885369 Chr.3: 64561724-64561724 on Build GRCh38
rs750462868 Chr.3: 64561528-64561528 on Build GRCh38
rs747843929 Chr.3: 64561752-64561752 on Build GRCh38
rs202061376 Chr.3: 64561763-64561763 on Build GRCh38
rs35667547 Chr.3: 64561801-64561801 on Build GRCh38
rs531153050 Chr.3: 64561590-64561590 on Build GRCh38
rs1012654622 Chr.3: 64561610-64561610 on Build GRCh38
rs767826229 Chr.3: 64561537-64561537 on Build GRCh38
rs766819593 Chr.3: 64561696-64561696 on Build GRCh38
rs920575073 Chr.3: 64561540-64561540 on Build GRCh38

View Full Product Details