Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101327671-101328221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SENP7
Entrez Gene ID
57337
Gene Name
SUMO1/sentrin specific peptidase 7
Gene Aliases
-
UniGene
Hs.529551
Gene Chromosome Location
Chr.3: 101324189-101513241 on Build GRCh38
Transcripts
NM_001077203.2 NM_001282801.1 NM_001282802.1 NM_001282803.1 NM_020654.4 XM_005247662.4 XM_011513038.2 XM_011513039.2 XM_011513040.2 XM_011513041.2 XM_011513042.2 XM_017006924.1 XM_017006925.1 XM_017006926.1 XM_017006927.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6326363 c.2906G>A p.R969H Chr.3: 101327775-101327775 on Build GRCh38
5750552 c.570C>A p.S190R Chr.3: 101327675-101327675 on Build GRCh38
5629423 c.496G>T p.D166Y Chr.3: 101327749-101327749 on Build GRCh38
3408070 c.2890A>G p.K964E Chr.3: 101327791-101327791 on Build GRCh38
4759442 c.2809T>G p.F937V Chr.3: 101327674-101327674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754029999 Chr.3: 101327859-101327859 on Build GRCh38
rs757009066 Chr.3: 101327770-101327770 on Build GRCh38
rs748921668 Chr.3: 101328035-101328042 on Build GRCh38
rs935988979 Chr.3: 101328189-101328189 on Build GRCh38
rs990201368 Chr.3: 101327852-101327852 on Build GRCh38
rs769464565 Chr.3: 101327805-101327805 on Build GRCh38
rs902078550 Chr.3: 101327979-101327979 on Build GRCh38
rs899689378 Chr.3: 101327932-101327932 on Build GRCh38
rs1006613482 Chr.3: 101328025-101328025 on Build GRCh38
rs760850518 Chr.3: 101327846-101327846 on Build GRCh38
rs955265011 Chr.3: 101327999-101327999 on Build GRCh38
rs376088352 Chr.3: 101327837-101327837 on Build GRCh38
rs141682754 Chr.3: 101327888-101327888 on Build GRCh38
rs745526738 Chr.3: 101327795-101327795 on Build GRCh38
rs895867475 Chr.3: 101327778-101327778 on Build GRCh38
rs775267345 Chr.3: 101327940-101327940 on Build GRCh38
rs991624544 Chr.3: 101328128-101328128 on Build GRCh38
rs889496395 Chr.3: 101328005-101328005 on Build GRCh38
rs762311324 Chr.3: 101327830-101327830 on Build GRCh38
rs937497679 Chr.3: 101328118-101328118 on Build GRCh38
rs147999457 Chr.3: 101327838-101327838 on Build GRCh38
rs769204505 Chr.3: 101327809-101327809 on Build GRCh38
rs950177433 Chr.3: 101328146-101328146 on Build GRCh38
rs774204894 Chr.3: 101327845-101327845 on Build GRCh38
rs182946375 Chr.3: 101327982-101327982 on Build GRCh38
rs1002825488 Chr.3: 101328151-101328151 on Build GRCh38
rs1038130853 Chr.3: 101328061-101328061 on Build GRCh38
rs768248822 Chr.3: 101328046-101328046 on Build GRCh38
rs770484888 Chr.3: 101327765-101327765 on Build GRCh38
rs1015672448 Chr.3: 101328010-101328010 on Build GRCh38
rs994015646 Chr.3: 101328126-101328126 on Build GRCh38
rs1032837356 Chr.3: 101327900-101327900 on Build GRCh38
rs562063622 Chr.3: 101327945-101327945 on Build GRCh38
rs903170947 Chr.3: 101328181-101328182 on Build GRCh38
rs748787962 Chr.3: 101327807-101327807 on Build GRCh38
rs117388797 Chr.3: 101328140-101328140 on Build GRCh38
rs9849770 Chr.3: 101327827-101327827 on Build GRCh38
rs147181609 Chr.3: 101327980-101327980 on Build GRCh38
rs779578577 Chr.3: 101327806-101327806 on Build GRCh38
rs520558 Chr.3: 101328089-101328089 on Build GRCh38
rs971716627 Chr.3: 101328143-101328143 on Build GRCh38
rs548135689 Chr.3: 101327891-101327891 on Build GRCh38
rs778761426 Chr.3: 101327771-101327771 on Build GRCh38
rs545234979 Chr.3: 101328014-101328014 on Build GRCh38
rs1055899376 Chr.3: 101327775-101327775 on Build GRCh38
rs974049419 Chr.3: 101328016-101328016 on Build GRCh38
rs190636203 Chr.3: 101327923-101327923 on Build GRCh38
rs375796723 Chr.3: 101327763-101327763 on Build GRCh38
rs369034392 Chr.3: 101327858-101327858 on Build GRCh38
rs112537671 Chr.3: 101327733-101327733 on Build GRCh38
rs542576091 Chr.3: 101328175-101328175 on Build GRCh38

View Full Product Details