Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101756966-101757511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP97
Entrez Gene ID
79598
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, LRRIQ2
UniGene
Hs.444135
Gene Chromosome Location
Chr.3: 101724098-101770562 on Build GRCh38
Transcripts
NM_001303401.1 NM_024548.3 XM_006713743.3 XM_011513127.1 XM_017007178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7352780 c.955G>C p.V319L Chr.3: 101757124-101757124 on Build GRCh38
6298973 c.997G>C p.V333L Chr.3: 101757166-101757166 on Build GRCh38
5568050 c.941C>T p.S314F Chr.3: 101757110-101757110 on Build GRCh38
727527 c.1021G>A p.E341K Chr.3: 101757190-101757190 on Build GRCh38
4842357 c.934G>C p.E312Q Chr.3: 101757103-101757103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757566858 Chr.3: 101757131-101757131 on Build GRCh38
rs373790394 Chr.3: 101757183-101757183 on Build GRCh38
rs1041235896 Chr.3: 101757346-101757346 on Build GRCh38
rs766127476 Chr.3: 101757078-101757078 on Build GRCh38
rs775502112 Chr.3: 101757040-101757040 on Build GRCh38
rs532126800 Chr.3: 101757052-101757052 on Build GRCh38
rs928240470 Chr.3: 101757184-101757184 on Build GRCh38
rs997167398 Chr.3: 101757396-101757396 on Build GRCh38
rs375068070 Chr.3: 101757230-101757230 on Build GRCh38
rs368306158 Chr.3: 101757253-101757253 on Build GRCh38
rs1048118360 Chr.3: 101757278-101757278 on Build GRCh38
rs576216332 Chr.3: 101757426-101757426 on Build GRCh38
rs1028350101 Chr.3: 101757406-101757406 on Build GRCh38
rs756290846 Chr.3: 101757136-101757136 on Build GRCh38
rs781563241 Chr.3: 101757133-101757133 on Build GRCh38
rs377071978 Chr.3: 101757171-101757171 on Build GRCh38
rs111350199 Chr.3: 101757447-101757447 on Build GRCh38
rs754893022 Chr.3: 101757007-101757007 on Build GRCh38
rs183185194 Chr.3: 101757153-101757153 on Build GRCh38
rs769877351 Chr.3: 101757024-101757024 on Build GRCh38
rs769321916 Chr.3: 101757222-101757223 on Build GRCh38
rs746112255 Chr.3: 101757135-101757135 on Build GRCh38
rs759026157 Chr.3: 101757115-101757115 on Build GRCh38
rs769549839 Chr.3: 101757237-101757237 on Build GRCh38
rs759213129 Chr.3: 101757098-101757098 on Build GRCh38
rs909995611 Chr.3: 101757286-101757286 on Build GRCh38
rs548717536 Chr.3: 101757359-101757359 on Build GRCh38
rs752227089 Chr.3: 101757114-101757114 on Build GRCh38
rs749354683 Chr.3: 101757170-101757170 on Build GRCh38
rs752032259 Chr.3: 101757128-101757128 on Build GRCh38
rs985544245 Chr.3: 101757251-101757251 on Build GRCh38
rs776826802 Chr.3: 101757016-101757017 on Build GRCh38
rs775200869 Chr.3: 101757248-101757248 on Build GRCh38
rs764869260 Chr.3: 101757100-101757100 on Build GRCh38
rs769196163 Chr.3: 101757017-101757017 on Build GRCh38
rs190066405 Chr.3: 101757427-101757427 on Build GRCh38
rs746902638 Chr.3: 101757212-101757212 on Build GRCh38
rs59720918 Chr.3: 101757124-101757124 on Build GRCh38
rs770650498 Chr.3: 101757223-101757223 on Build GRCh38
rs201473764 Chr.3: 101757446-101757448 on Build GRCh38
rs558812520 Chr.3: 101757379-101757379 on Build GRCh38
rs1045846518 Chr.3: 101757437-101757437 on Build GRCh38
rs186139092 Chr.3: 101757364-101757364 on Build GRCh38
rs914102451 Chr.3: 101757122-101757122 on Build GRCh38
rs958013485 Chr.3: 101757407-101757407 on Build GRCh38
rs564831319 Chr.3: 101757009-101757009 on Build GRCh38
rs759299247 Chr.3: 101757234-101757234 on Build GRCh38
rs976558420 Chr.3: 101757197-101757197 on Build GRCh38
rs1021070407 Chr.3: 101757431-101757431 on Build GRCh38
rs557327055 Chr.3: 101757296-101757296 on Build GRCh38
rs565761991 Chr.3: 101757079-101757079 on Build GRCh38
rs748121588 Chr.3: 101757043-101757043 on Build GRCh38
rs966746257 Chr.3: 101757469-101757469 on Build GRCh38
rs1014341390 Chr.3: 101757001-101757001 on Build GRCh38
rs776444063 Chr.3: 101757228-101757228 on Build GRCh38
rs1024428185 Chr.3: 101757008-101757008 on Build GRCh38
rs547201342 Chr.3: 101757063-101757063 on Build GRCh38
rs537314684 Chr.3: 101757372-101757372 on Build GRCh38
rs112580461 Chr.3: 101757350-101757350 on Build GRCh38
rs770915426 Chr.3: 101757261-101757261 on Build GRCh38
rs372730278 Chr.3: 101757023-101757023 on Build GRCh38
rs775429615 Chr.3: 101757313-101757313 on Build GRCh38
rs762309270 Chr.3: 101757196-101757196 on Build GRCh38
rs932507016 Chr.3: 101757250-101757250 on Build GRCh38

View Full Product Details