Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101827526-101828031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFKBIZ
Entrez Gene ID
64332
Gene Name
NFKB inhibitor zeta
Gene Aliases
IKBZ, INAP, MAIL
UniGene
Hs.319171
Gene Chromosome Location
Chr.3: 101827990-101861025 on Build GRCh38
Transcripts
NM_001005474.2 NM_001134456.1 NM_145037.2

Gene Symbol
NXPE3
Entrez Gene ID
91775
Gene Name
neurexophilin and PC-esterase domain family member 3
Gene Aliases
FAM55C, MST115, MSTP115
UniGene
Hs.595933
Gene Chromosome Location
Chr.3: 101779185-101828231 on Build GRCh38
Transcripts
NM_001005474.2 NM_001134456.1 NM_145037.2

SNP Detail

SNP ID Chromosome Location
rs932229617 Chr.3: 101827768-101827768 on Build GRCh38
rs896849400 Chr.3: 101827564-101827564 on Build GRCh38
rs966168057 Chr.3: 101827998-101827998 on Build GRCh38
rs13317050 Chr.3: 101827940-101827940 on Build GRCh38
rs538163565 Chr.3: 101827723-101827723 on Build GRCh38
rs905746552 Chr.3: 101827926-101827926 on Build GRCh38
rs1000537403 Chr.3: 101827838-101827838 on Build GRCh38
rs941083524 Chr.3: 101827813-101827813 on Build GRCh38
rs922150361 Chr.3: 101827756-101827756 on Build GRCh38
rs573745500 Chr.3: 101827684-101827687 on Build GRCh38
rs969959524 Chr.3: 101827661-101827661 on Build GRCh38
rs980763209 Chr.3: 101827868-101827868 on Build GRCh38
rs538699138 Chr.3: 101827751-101827751 on Build GRCh38
rs1010304069 Chr.3: 101827976-101827976 on Build GRCh38
rs909422003 Chr.3: 101827794-101827794 on Build GRCh38
rs752835328 Chr.3: 101827705-101827705 on Build GRCh38
rs745634227 Chr.3: 101827774-101827774 on Build GRCh38
rs1024618876 Chr.3: 101827658-101827658 on Build GRCh38
rs966465785 Chr.3: 101827746-101827746 on Build GRCh38
rs573855380 Chr.3: 101827865-101827865 on Build GRCh38
rs867894558 Chr.3: 101827588-101827588 on Build GRCh38
rs544400368 Chr.3: 101827889-101827889 on Build GRCh38
rs549166827 Chr.3: 101827556-101827556 on Build GRCh38
rs1032798423 Chr.3: 101827954-101827954 on Build GRCh38
rs749618752 Chr.3: 101827779-101827779 on Build GRCh38
rs956657252 Chr.3: 101827659-101827659 on Build GRCh38
rs1033642576 Chr.3: 101827924-101827924 on Build GRCh38
rs571886654 Chr.3: 101827731-101827731 on Build GRCh38
rs912138708 Chr.3: 101827983-101827983 on Build GRCh38
rs572450947 Chr.3: 101827819-101827819 on Build GRCh38
rs957448955 Chr.3: 101827707-101827707 on Build GRCh38
rs942212910 Chr.3: 101827732-101827732 on Build GRCh38
rs973868082 Chr.3: 101827936-101827936 on Build GRCh38
rs1014153426 Chr.3: 101827644-101827644 on Build GRCh38
rs779456694 Chr.3: 101827985-101827985 on Build GRCh38
rs917942744 Chr.3: 101827687-101827687 on Build GRCh38
rs756222880 Chr.3: 101827740-101827740 on Build GRCh38
rs17347617 Chr.3: 101828003-101828003 on Build GRCh38
rs558481494 Chr.3: 101827777-101827777 on Build GRCh38
rs567477718 Chr.3: 101827695-101827695 on Build GRCh38
rs1032550449 Chr.3: 101827631-101827631 on Build GRCh38
rs1025297094 Chr.3: 101827859-101827859 on Build GRCh38
rs369798395 Chr.3: 101827784-101827784 on Build GRCh38
rs773622431 Chr.3: 101827882-101827882 on Build GRCh38
rs192578865 Chr.3: 101827820-101827820 on Build GRCh38
rs879459386 Chr.3: 101827682-101827682 on Build GRCh38
rs537201491 Chr.3: 101827555-101827555 on Build GRCh38
rs778171574 Chr.3: 101827772-101827772 on Build GRCh38
rs987771248 Chr.3: 101827958-101827958 on Build GRCh38
rs72937687 Chr.3: 101827841-101827841 on Build GRCh38
rs1001287949 Chr.3: 101827680-101827680 on Build GRCh38
rs957229438 Chr.3: 101827955-101827955 on Build GRCh38
rs556549614 Chr.3: 101827729-101827729 on Build GRCh38
rs1014307182 Chr.3: 101827854-101827854 on Build GRCh38
rs980879677 Chr.3: 101827699-101827699 on Build GRCh38
rs189274492 Chr.3: 101827764-101827764 on Build GRCh38
rs943746978 Chr.3: 101827993-101827993 on Build GRCh38

View Full Product Details