Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101849714-101850234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFKBIZ
Entrez Gene ID
64332
Gene Name
NFKB inhibitor zeta
Gene Aliases
IKBZ, INAP, MAIL
UniGene
Hs.319171
Gene Chromosome Location
Chr.3: 101827990-101861025 on Build GRCh38
Transcripts
NM_001005474.2 NM_031419.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5798297 c.283C>A p.Q95K Chr.3: 101849911-101849911 on Build GRCh38
6313214 c.246G>T p.V82V Chr.3: 101849874-101849874 on Build GRCh38
6398281 c.127G>A p.A43T Chr.3: 101849755-101849755 on Build GRCh38
6210045 c.217G>A p.A73T Chr.3: 101849845-101849845 on Build GRCh38
5568749 c.199T>G p.S67A Chr.3: 101849827-101849827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756937236 Chr.3: 101849742-101849742 on Build GRCh38
rs779117101 Chr.3: 101849911-101849911 on Build GRCh38
rs958107572 Chr.3: 101849752-101849752 on Build GRCh38
rs974169056 Chr.3: 101849920-101849920 on Build GRCh38
rs1038282991 Chr.3: 101850006-101850006 on Build GRCh38
rs1010705124 Chr.3: 101850126-101850126 on Build GRCh38
rs1049652510 Chr.3: 101850084-101850084 on Build GRCh38
rs910997135 Chr.3: 101850177-101850177 on Build GRCh38
rs1025453412 Chr.3: 101850027-101850027 on Build GRCh38
rs62280809 Chr.3: 101850077-101850077 on Build GRCh38
rs1034853583 Chr.3: 101850042-101850042 on Build GRCh38
rs539840555 Chr.3: 101849992-101849992 on Build GRCh38
rs753478094 Chr.3: 101849859-101849859 on Build GRCh38
rs946224747 Chr.3: 101849741-101849741 on Build GRCh38
rs1002918758 Chr.3: 101850036-101850036 on Build GRCh38
rs1042111040 Chr.3: 101849760-101849760 on Build GRCh38
rs557155082 Chr.3: 101849904-101849904 on Build GRCh38
rs561824277 Chr.3: 101850020-101850020 on Build GRCh38
rs755421482 Chr.3: 101849873-101849873 on Build GRCh38
rs575628701 Chr.3: 101849905-101849905 on Build GRCh38
rs1017266669 Chr.3: 101849847-101849847 on Build GRCh38
rs867815814 Chr.3: 101849799-101849799 on Build GRCh38
rs889420850 Chr.3: 101850003-101850003 on Build GRCh38
rs914810080 Chr.3: 101850101-101850101 on Build GRCh38
rs1041893316 Chr.3: 101849943-101849943 on Build GRCh38
rs959640744 Chr.3: 101849991-101849991 on Build GRCh38
rs900865058 Chr.3: 101850068-101850068 on Build GRCh38
rs1020953966 Chr.3: 101850130-101850130 on Build GRCh38
rs779785022 Chr.3: 101849841-101849841 on Build GRCh38
rs962752222 Chr.3: 101849748-101849748 on Build GRCh38
rs923419351 Chr.3: 101850118-101850118 on Build GRCh38
rs750193192 Chr.3: 101849796-101849796 on Build GRCh38
rs966650949 Chr.3: 101849973-101849973 on Build GRCh38
rs971237051 Chr.3: 101850034-101850034 on Build GRCh38
rs893263734 Chr.3: 101849740-101849740 on Build GRCh38
rs1027922012 Chr.3: 101850203-101850206 on Build GRCh38
rs755947769 Chr.3: 101849838-101849838 on Build GRCh38
rs1029360603 Chr.3: 101849828-101849828 on Build GRCh38
rs946218337 Chr.3: 101849938-101849938 on Build GRCh38
rs932567218 Chr.3: 101850069-101850069 on Build GRCh38
rs939805544 Chr.3: 101850028-101850028 on Build GRCh38
rs898439217 Chr.3: 101850007-101850007 on Build GRCh38
rs997201642 Chr.3: 101849926-101849926 on Build GRCh38
rs902313383 Chr.3: 101849786-101849786 on Build GRCh38
rs572831094 Chr.3: 101849946-101849946 on Build GRCh38
rs375895348 Chr.3: 101849927-101849927 on Build GRCh38
rs977658559 Chr.3: 101850114-101850114 on Build GRCh38
rs952157235 Chr.3: 101850203-101850203 on Build GRCh38
rs1050832371 Chr.3: 101850143-101850143 on Build GRCh38
rs967634503 Chr.3: 101850104-101850104 on Build GRCh38
rs766089296 Chr.3: 101849791-101849791 on Build GRCh38
rs1038070709 Chr.3: 101850189-101850189 on Build GRCh38
rs1021009709 Chr.3: 101849940-101849940 on Build GRCh38
rs933728001 Chr.3: 101849984-101849984 on Build GRCh38
rs982135685 Chr.3: 101849988-101849988 on Build GRCh38
rs577809892 Chr.3: 101850094-101850094 on Build GRCh38
rs181272860 Chr.3: 101850109-101850109 on Build GRCh38
rs898175861 Chr.3: 101850197-101850197 on Build GRCh38
rs767309092 Chr.3: 101849743-101849743 on Build GRCh38
rs938137495 Chr.3: 101849855-101849855 on Build GRCh38
rs1010924529 Chr.3: 101849937-101849937 on Build GRCh38
rs1006786471 Chr.3: 101849836-101849836 on Build GRCh38
rs749241699 Chr.3: 101849870-101849870 on Build GRCh38
rs545226790 Chr.3: 101850125-101850125 on Build GRCh38
rs557746328 Chr.3: 101849932-101849932 on Build GRCh38
rs1035467931 Chr.3: 101849990-101849990 on Build GRCh38
rs535550319 Chr.3: 101850076-101850076 on Build GRCh38
rs889464150 Chr.3: 101849831-101849831 on Build GRCh38
rs546092647 Chr.3: 101849908-101849908 on Build GRCh38
rs972601070 Chr.3: 101849866-101849866 on Build GRCh38
rs1035013293 Chr.3: 101850164-101850164 on Build GRCh38
rs1004068524 Chr.3: 101850159-101850159 on Build GRCh38
rs1006976687 Chr.3: 101850005-101850005 on Build GRCh38
rs748505249 Chr.3: 101850078-101850078 on Build GRCh38
rs189392097 Chr.3: 101849801-101849801 on Build GRCh38
rs942362557 Chr.3: 101850184-101850184 on Build GRCh38
rs997790777 Chr.3: 101849809-101849809 on Build GRCh38
rs893666589 Chr.3: 101849935-101849935 on Build GRCh38
rs933488506 Chr.3: 101850127-101850127 on Build GRCh38
rs990592443 Chr.3: 101850098-101850098 on Build GRCh38
rs9867267 Chr.3: 101850181-101850181 on Build GRCh38
rs535651244 Chr.3: 101849901-101849901 on Build GRCh38
rs971499460 Chr.3: 101849882-101849882 on Build GRCh38

View Full Product Details