Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119203827-119204324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UPK1B
Entrez Gene ID
7348
Gene Name
uroplakin 1B
Gene Aliases
TSPAN20, UPIB, UPK1
UniGene
Hs.271580
Gene Chromosome Location
Chr.3: 119173578-119205153 on Build GRCh38
Transcripts
NM_006952.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6817654 c.782A>G p.*261* Chr.3: 119203966-119203966 on Build GRCh38
286360 c.742C>T p.L248F Chr.3: 119203926-119203926 on Build GRCh38
1666085 c.770_771insT p.R257fs*3 Chr.3: 119203954-119203955 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747835180 Chr.3: 119203896-119203896 on Build GRCh38
rs202057233 Chr.3: 119203906-119203906 on Build GRCh38
rs895736566 Chr.3: 119203861-119203861 on Build GRCh38
rs142595322 Chr.3: 119204100-119204100 on Build GRCh38
rs200923434 Chr.3: 119203902-119203902 on Build GRCh38
rs769037799 Chr.3: 119203939-119203939 on Build GRCh38
rs76337943 Chr.3: 119204275-119204275 on Build GRCh38
rs998363059 Chr.3: 119204293-119204293 on Build GRCh38
rs1044729127 Chr.3: 119204130-119204130 on Build GRCh38
rs893258624 Chr.3: 119204263-119204263 on Build GRCh38
rs758283438 Chr.3: 119203900-119203900 on Build GRCh38
rs925700905 Chr.3: 119204096-119204096 on Build GRCh38
rs561840255 Chr.3: 119203862-119203862 on Build GRCh38
rs780115448 Chr.3: 119203933-119203933 on Build GRCh38
rs200460242 Chr.3: 119203905-119203905 on Build GRCh38
rs965954527 Chr.3: 119204279-119204279 on Build GRCh38
rs1046322117 Chr.3: 119204101-119204101 on Build GRCh38
rs760301948 Chr.3: 119203970-119203970 on Build GRCh38
rs746125035 Chr.3: 119203908-119203908 on Build GRCh38
rs760146855 Chr.3: 119204238-119204238 on Build GRCh38
rs747992285 Chr.3: 119204113-119204113 on Build GRCh38
rs749442831 Chr.3: 119203936-119203936 on Build GRCh38
rs565402265 Chr.3: 119204075-119204075 on Build GRCh38
rs765834851 Chr.3: 119203866-119203866 on Build GRCh38
rs867946258 Chr.3: 119203984-119203984 on Build GRCh38
rs778980644 Chr.3: 119203890-119203890 on Build GRCh38
rs768673308 Chr.3: 119203980-119203980 on Build GRCh38
rs766117150 Chr.3: 119203975-119203975 on Build GRCh38
rs754861348 Chr.3: 119203887-119203887 on Build GRCh38
rs749185748 Chr.3: 119203959-119203959 on Build GRCh38
rs770179195 Chr.3: 119203910-119203910 on Build GRCh38
rs775407506 Chr.3: 119203922-119203922 on Build GRCh38
rs1020526629 Chr.3: 119203968-119203968 on Build GRCh38
rs921508699 Chr.3: 119204079-119204079 on Build GRCh38
rs1048345755 Chr.3: 119204212-119204212 on Build GRCh38
rs888531197 Chr.3: 119204267-119204267 on Build GRCh38
rs893268146 Chr.3: 119203947-119203947 on Build GRCh38
rs867885959 Chr.3: 119203993-119203993 on Build GRCh38
rs188911128 Chr.3: 119204023-119204023 on Build GRCh38
rs372939005 Chr.3: 119203989-119203989 on Build GRCh38
rs965672137 Chr.3: 119204032-119204032 on Build GRCh38
rs1029027719 Chr.3: 119204302-119204302 on Build GRCh38
rs369195775 Chr.3: 119203863-119203863 on Build GRCh38
rs997172025 Chr.3: 119203990-119203990 on Build GRCh38
rs752166080 Chr.3: 119203890-119203891 on Build GRCh38
rs917041393 Chr.3: 119204114-119204114 on Build GRCh38
rs772809374 Chr.3: 119203962-119203962 on Build GRCh38
rs115262398 Chr.3: 119204048-119204048 on Build GRCh38
rs766839177 Chr.3: 119203915-119203915 on Build GRCh38
rs752106466 Chr.3: 119204022-119204022 on Build GRCh38
rs753440425 Chr.3: 119203868-119203868 on Build GRCh38
rs963181755 Chr.3: 119204081-119204081 on Build GRCh38
rs978416578 Chr.3: 119204089-119204089 on Build GRCh38
rs777682196 Chr.3: 119204000-119204000 on Build GRCh38
rs905219856 Chr.3: 119203943-119203943 on Build GRCh38
rs953884893 Chr.3: 119204087-119204087 on Build GRCh38
rs772916634 Chr.3: 119204140-119204140 on Build GRCh38
rs1002193840 Chr.3: 119204179-119204179 on Build GRCh38
rs1001589434 Chr.3: 119203877-119203877 on Build GRCh38
rs114878363 Chr.3: 119203996-119203996 on Build GRCh38
rs186059070 Chr.3: 119203957-119203957 on Build GRCh38
rs758228581 Chr.3: 119203999-119203999 on Build GRCh38
rs755740080 Chr.3: 119204268-119204268 on Build GRCh38
rs539091282 Chr.3: 119204123-119204123 on Build GRCh38
rs760094694 Chr.3: 119203914-119203915 on Build GRCh38
rs754525263 Chr.3: 119203987-119203987 on Build GRCh38
rs545390054 Chr.3: 119203856-119203856 on Build GRCh38
rs1055884543 Chr.3: 119204214-119204216 on Build GRCh38

View Full Product Details