Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119544471-119544976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD80
Entrez Gene ID
941
Gene Name
CD80 molecule
Gene Aliases
B7, B7-1, B7.1, BB1, CD28LG, CD28LG1, LAB7
UniGene
Hs.838
Gene Chromosome Location
Chr.3: 119523909-119559709 on Build GRCh38
Transcripts
NM_005191.3 XM_011513327.2 XM_017007520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1036758 c.289A>C p.N97H Chr.3: 119544679-119544679 on Build GRCh38
4112545 c.396T>C p.A132A Chr.3: 119544572-119544572 on Build GRCh38
4112546 c.188G>A p.R63H Chr.3: 119544780-119544780 on Build GRCh38
6623901 c.372C>T p.D124D Chr.3: 119544596-119544596 on Build GRCh38
4112544 c.404C>T p.T135M Chr.3: 119544564-119544564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023309227 Chr.3: 119544498-119544498 on Build GRCh38
rs750432224 Chr.3: 119544913-119544913 on Build GRCh38
rs771359795 Chr.3: 119544706-119544706 on Build GRCh38
rs767373131 Chr.3: 119544780-119544780 on Build GRCh38
rs762668335 Chr.3: 119544611-119544611 on Build GRCh38
rs758270977 Chr.3: 119544567-119544567 on Build GRCh38
rs749569459 Chr.3: 119544593-119544593 on Build GRCh38
rs755938741 Chr.3: 119544916-119544916 on Build GRCh38
rs528831990 Chr.3: 119544950-119544950 on Build GRCh38
rs777767800 Chr.3: 119544579-119544579 on Build GRCh38
rs183378805 Chr.3: 119544829-119544829 on Build GRCh38
rs149271868 Chr.3: 119544582-119544582 on Build GRCh38
rs554004644 Chr.3: 119544752-119544752 on Build GRCh38
rs370971089 Chr.3: 119544564-119544564 on Build GRCh38
rs368745104 Chr.3: 119544712-119544712 on Build GRCh38
rs185402094 Chr.3: 119544942-119544942 on Build GRCh38
rs369146723 Chr.3: 119544874-119544874 on Build GRCh38
rs772241262 Chr.3: 119544691-119544691 on Build GRCh38
rs746220473 Chr.3: 119544835-119544835 on Build GRCh38
rs9282638 Chr.3: 119544923-119544923 on Build GRCh38
rs750697479 Chr.3: 119544540-119544540 on Build GRCh38
rs147732996 Chr.3: 119544518-119544518 on Build GRCh38
rs762000038 Chr.3: 119544606-119544606 on Build GRCh38
rs752132958 Chr.3: 119544647-119544647 on Build GRCh38
rs372271427 Chr.3: 119544596-119544596 on Build GRCh38
rs775739462 Chr.3: 119544759-119544760 on Build GRCh38
rs761167098 Chr.3: 119544556-119544556 on Build GRCh38
rs760922746 Chr.3: 119544911-119544911 on Build GRCh38
rs140149683 Chr.3: 119544723-119544723 on Build GRCh38
rs376720196 Chr.3: 119544898-119544898 on Build GRCh38
rs9282639 Chr.3: 119544897-119544897 on Build GRCh38
rs977184373 Chr.3: 119544920-119544920 on Build GRCh38
rs762428605 Chr.3: 119544742-119544742 on Build GRCh38
rs944044905 Chr.3: 119544642-119544642 on Build GRCh38
rs199861577 Chr.3: 119544626-119544626 on Build GRCh38
rs765932547 Chr.3: 119544563-119544563 on Build GRCh38
rs778350268 Chr.3: 119544566-119544566 on Build GRCh38
rs781001516 Chr.3: 119544667-119544667 on Build GRCh38
rs1014349892 Chr.3: 119544810-119544810 on Build GRCh38
rs116944420 Chr.3: 119544671-119544671 on Build GRCh38
rs780121133 Chr.3: 119544586-119544586 on Build GRCh38
rs570995916 Chr.3: 119544517-119544517 on Build GRCh38
rs895447298 Chr.3: 119544776-119544776 on Build GRCh38
rs750178788 Chr.3: 119544786-119544786 on Build GRCh38
rs759136851 Chr.3: 119544638-119544638 on Build GRCh38
rs760924166 Chr.3: 119544630-119544630 on Build GRCh38
rs775093170 Chr.3: 119544857-119544857 on Build GRCh38
rs62264489 Chr.3: 119544858-119544858 on Build GRCh38
rs748417002 Chr.3: 119544821-119544821 on Build GRCh38
rs766806612 Chr.3: 119544632-119544632 on Build GRCh38
rs756974714 Chr.3: 119544662-119544662 on Build GRCh38
rs770707039 Chr.3: 119544673-119544673 on Build GRCh38
rs745680949 Chr.3: 119544668-119544668 on Build GRCh38
rs921535147 Chr.3: 119544659-119544659 on Build GRCh38
rs757278848 Chr.3: 119544699-119544699 on Build GRCh38
rs779283878 Chr.3: 119544674-119544674 on Build GRCh38
rs761204102 Chr.3: 119544700-119544700 on Build GRCh38
rs759577101 Chr.3: 119544508-119544508 on Build GRCh38
rs776993724 Chr.3: 119544707-119544707 on Build GRCh38
rs142547094 Chr.3: 119544825-119544825 on Build GRCh38
rs376672655 Chr.3: 119544637-119544637 on Build GRCh38
rs777859790 Chr.3: 119544827-119544827 on Build GRCh38
rs867968226 Chr.3: 119544847-119544847 on Build GRCh38
rs1022857991 Chr.3: 119544819-119544819 on Build GRCh38
rs756028722 Chr.3: 119544794-119544794 on Build GRCh38
rs764879350 Chr.3: 119544641-119544641 on Build GRCh38
rs764862930 Chr.3: 119544713-119544713 on Build GRCh38
rs756594891 Chr.3: 119544871-119544871 on Build GRCh38
rs766233590 Chr.3: 119544918-119544918 on Build GRCh38
rs772700006 Chr.3: 119544625-119544625 on Build GRCh38
rs770820543 Chr.3: 119544585-119544585 on Build GRCh38
rs370099351 Chr.3: 119544764-119544764 on Build GRCh38
rs373759978 Chr.3: 119544896-119544896 on Build GRCh38
rs751567741 Chr.3: 119544657-119544657 on Build GRCh38
rs375549265 Chr.3: 119544763-119544763 on Build GRCh38
rs748224407 Chr.3: 119544681-119544681 on Build GRCh38
rs770194191 Chr.3: 119544848-119544848 on Build GRCh38
rs2228017 Chr.3: 119544833-119544833 on Build GRCh38
rs752552367 Chr.3: 119544648-119544648 on Build GRCh38
rs142958721 Chr.3: 119544864-119544864 on Build GRCh38
rs777560868 Chr.3: 119544649-119544649 on Build GRCh38
rs146886200 Chr.3: 119544856-119544856 on Build GRCh38
rs201443429 Chr.3: 119544828-119544828 on Build GRCh38
rs931956626 Chr.3: 119544795-119544795 on Build GRCh38
rs763700778 Chr.3: 119544747-119544747 on Build GRCh38

View Full Product Details