Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120324647-120325160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC58
Entrez Gene ID
116064
Gene Name
leucine rich repeat containing 58
Gene Aliases
-
UniGene
Hs.518084
Gene Chromosome Location
Chr.3: 120324729-120349339 on Build GRCh38
Transcripts
NM_001099678.1

SNP Detail

SNP ID Chromosome Location
rs955746719 Chr.3: 120324897-120324897 on Build GRCh38
rs374147203 Chr.3: 120324852-120324852 on Build GRCh38
rs556809467 Chr.3: 120325032-120325032 on Build GRCh38
rs1052582782 Chr.3: 120324678-120324678 on Build GRCh38
rs1014153701 Chr.3: 120324849-120324849 on Build GRCh38
rs1022665789 Chr.3: 120324851-120324851 on Build GRCh38
rs372526673 Chr.3: 120324912-120324912 on Build GRCh38
rs560214169 Chr.3: 120324997-120324997 on Build GRCh38
rs988736112 Chr.3: 120324905-120324905 on Build GRCh38
rs1041468826 Chr.3: 120324731-120324731 on Build GRCh38
rs201759328 Chr.3: 120324955-120324955 on Build GRCh38
rs944281256 Chr.3: 120324690-120324690 on Build GRCh38
rs576712562 Chr.3: 120325117-120325117 on Build GRCh38
rs944319793 Chr.3: 120324941-120324941 on Build GRCh38
rs758013127 Chr.3: 120324734-120324734 on Build GRCh38
rs1015397505 Chr.3: 120324822-120324822 on Build GRCh38
rs568124392 Chr.3: 120324791-120324791 on Build GRCh38
rs531727221 Chr.3: 120324740-120324740 on Build GRCh38
rs548130258 Chr.3: 120324758-120324758 on Build GRCh38
rs1048347094 Chr.3: 120325112-120325112 on Build GRCh38
rs193052429 Chr.3: 120324735-120324735 on Build GRCh38
rs895738755 Chr.3: 120324823-120324823 on Build GRCh38
rs10511400 Chr.3: 120324989-120324989 on Build GRCh38
rs190686962 Chr.3: 120324732-120324732 on Build GRCh38
rs142768626 Chr.3: 120325124-120325124 on Build GRCh38
rs936825748 Chr.3: 120324674-120324674 on Build GRCh38
rs914057284 Chr.3: 120324918-120324918 on Build GRCh38
rs185680218 Chr.3: 120324689-120324689 on Build GRCh38
rs1048457809 Chr.3: 120324774-120324774 on Build GRCh38
rs571003802 Chr.3: 120324960-120324960 on Build GRCh38
rs369286488 Chr.3: 120325125-120325125 on Build GRCh38
rs184498984 Chr.3: 120324801-120324801 on Build GRCh38
rs116389131 Chr.3: 120324883-120324883 on Build GRCh38
rs978385226 Chr.3: 120324872-120324872 on Build GRCh38
rs932655263 Chr.3: 120325022-120325022 on Build GRCh38
rs925481302 Chr.3: 120324673-120324673 on Build GRCh38

View Full Product Details