Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120329016-120329559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC58
Entrez Gene ID
116064
Gene Name
leucine rich repeat containing 58
Gene Aliases
-
UniGene
Hs.518084
Gene Chromosome Location
Chr.3: 120324729-120349339 on Build GRCh38
Transcripts
NM_001099678.1

SNP Detail

SNP ID Chromosome Location
rs573242486 Chr.3: 120329124-120329124 on Build GRCh38
rs145749987 Chr.3: 120329121-120329121 on Build GRCh38
rs966746171 Chr.3: 120329235-120329235 on Build GRCh38
rs141281821 Chr.3: 120329283-120329283 on Build GRCh38
rs11716277 Chr.3: 120329246-120329246 on Build GRCh38
rs1035116268 Chr.3: 120329377-120329377 on Build GRCh38
rs942324607 Chr.3: 120329499-120329499 on Build GRCh38
rs753836777 Chr.3: 120329247-120329247 on Build GRCh38
rs890817673 Chr.3: 120329053-120329053 on Build GRCh38
rs935136485 Chr.3: 120329174-120329174 on Build GRCh38
rs528153479 Chr.3: 120329524-120329524 on Build GRCh38
rs148307306 Chr.3: 120329355-120329355 on Build GRCh38
rs918449140 Chr.3: 120329387-120329387 on Build GRCh38
rs757318149 Chr.3: 120329272-120329272 on Build GRCh38
rs562864640 Chr.3: 120329226-120329226 on Build GRCh38
rs1011020995 Chr.3: 120329220-120329220 on Build GRCh38
rs573787091 Chr.3: 120329102-120329103 on Build GRCh38
rs755537897 Chr.3: 120329072-120329072 on Build GRCh38
rs946605603 Chr.3: 120329075-120329075 on Build GRCh38
rs1018232530 Chr.3: 120329132-120329132 on Build GRCh38
rs76566364 Chr.3: 120329437-120329437 on Build GRCh38
rs188739327 Chr.3: 120329418-120329418 on Build GRCh38
rs890843735 Chr.3: 120329217-120329217 on Build GRCh38
rs1051261182 Chr.3: 120329440-120329440 on Build GRCh38
rs960755890 Chr.3: 120329390-120329390 on Build GRCh38
rs559093760 Chr.3: 120329502-120329502 on Build GRCh38
rs9859833 Chr.3: 120329204-120329204 on Build GRCh38
rs1050807180 Chr.3: 120329184-120329184 on Build GRCh38
rs536686232 Chr.3: 120329076-120329076 on Build GRCh38
rs573025454 Chr.3: 120329345-120329348 on Build GRCh38
rs781548092 Chr.3: 120329141-120329141 on Build GRCh38
rs929700501 Chr.3: 120329420-120329420 on Build GRCh38
rs34715000 Chr.3: 120329342-120329343 on Build GRCh38
rs1043424612 Chr.3: 120329111-120329115 on Build GRCh38
rs890880466 Chr.3: 120329485-120329485 on Build GRCh38
rs145013589 Chr.3: 120329348-120329348 on Build GRCh38
rs916384548 Chr.3: 120329481-120329481 on Build GRCh38
rs960968289 Chr.3: 120329251-120329251 on Build GRCh38

View Full Product Details