Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120331011-120331506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC58
Entrez Gene ID
116064
Gene Name
leucine rich repeat containing 58
Gene Aliases
-
UniGene
Hs.518084
Gene Chromosome Location
Chr.3: 120324729-120349339 on Build GRCh38
Transcripts
NM_001099678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1418081 c.1076G>C p.S359T Chr.3: 120331240-120331240 on Build GRCh38
4456527 c.1012C>T p.P338S Chr.3: 120331304-120331304 on Build GRCh38
1318742 c.1078G>A p.V360I Chr.3: 120331238-120331238 on Build GRCh38
6703993 c.1073C>T p.A358V Chr.3: 120331243-120331243 on Build GRCh38
6096080 c.1063G>C p.E355Q Chr.3: 120331253-120331253 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs904330012 Chr.3: 120331110-120331110 on Build GRCh38
rs746947407 Chr.3: 120331288-120331288 on Build GRCh38
rs546885697 Chr.3: 120331292-120331292 on Build GRCh38
rs746570514 Chr.3: 120331451-120331451 on Build GRCh38
rs755236722 Chr.3: 120331422-120331422 on Build GRCh38
rs767674607 Chr.3: 120331380-120331380 on Build GRCh38
rs750579351 Chr.3: 120331408-120331408 on Build GRCh38
rs776343293 Chr.3: 120331154-120331154 on Build GRCh38
rs761230451 Chr.3: 120331413-120331413 on Build GRCh38
rs77005092 Chr.3: 120331187-120331187 on Build GRCh38
rs1031393478 Chr.3: 120331136-120331136 on Build GRCh38
rs757355348 Chr.3: 120331173-120331173 on Build GRCh38
rs772753939 Chr.3: 120331282-120331282 on Build GRCh38
rs780443520 Chr.3: 120331218-120331218 on Build GRCh38
rs771518348 Chr.3: 120331267-120331267 on Build GRCh38
rs561092787 Chr.3: 120331122-120331122 on Build GRCh38
rs542629162 Chr.3: 120331072-120331072 on Build GRCh38
rs114533645 Chr.3: 120331083-120331083 on Build GRCh38
rs201989674 Chr.3: 120331228-120331228 on Build GRCh38
rs1021405409 Chr.3: 120331098-120331098 on Build GRCh38
rs375549625 Chr.3: 120331457-120331457 on Build GRCh38
rs866771170 Chr.3: 120331423-120331423 on Build GRCh38
rs111898403 Chr.3: 120331048-120331048 on Build GRCh38
rs1004121377 Chr.3: 120331364-120331364 on Build GRCh38
rs188827595 Chr.3: 120331465-120331465 on Build GRCh38
rs908612213 Chr.3: 120331389-120331389 on Build GRCh38
rs764292107 Chr.3: 120331164-120331164 on Build GRCh38
rs1022722176 Chr.3: 120331478-120331478 on Build GRCh38
rs770834372 Chr.3: 120331290-120331290 on Build GRCh38
rs774667295 Chr.3: 120331338-120331338 on Build GRCh38
rs564893232 Chr.3: 120331059-120331059 on Build GRCh38
rs200466593 Chr.3: 120331229-120331229 on Build GRCh38
rs931446134 Chr.3: 120331344-120331344 on Build GRCh38
rs748564857 Chr.3: 120331263-120331263 on Build GRCh38
rs754372386 Chr.3: 120331421-120331421 on Build GRCh38
rs887209503 Chr.3: 120331053-120331053 on Build GRCh38
rs959807826 Chr.3: 120331119-120331119 on Build GRCh38
rs766169633 Chr.3: 120331153-120331153 on Build GRCh38
rs1040888688 Chr.3: 120331046-120331046 on Build GRCh38
rs779268480 Chr.3: 120331428-120331428 on Build GRCh38
rs760624843 Chr.3: 120331152-120331152 on Build GRCh38
rs372468489 Chr.3: 120331189-120331189 on Build GRCh38
rs1048682846 Chr.3: 120331362-120331362 on Build GRCh38
rs756619195 Chr.3: 120331199-120331199 on Build GRCh38
rs766860444 Chr.3: 120331417-120331417 on Build GRCh38
rs532436092 Chr.3: 120331442-120331442 on Build GRCh38
rs562802423 Chr.3: 120331088-120331088 on Build GRCh38
rs983368643 Chr.3: 120331375-120331375 on Build GRCh38
rs376883169 Chr.3: 120331167-120331167 on Build GRCh38
rs778468490 Chr.3: 120331233-120331233 on Build GRCh38
rs932661818 Chr.3: 120331450-120331450 on Build GRCh38
rs888394750 Chr.3: 120331363-120331363 on Build GRCh38
rs9850986 Chr.3: 120331437-120331437 on Build GRCh38
rs747808266 Chr.3: 120331239-120331239 on Build GRCh38
rs368394965 Chr.3: 120331374-120331374 on Build GRCh38
rs763227362 Chr.3: 120331162-120331162 on Build GRCh38
rs201166538 Chr.3: 120331309-120331309 on Build GRCh38
rs76906770 Chr.3: 120331316-120331316 on Build GRCh38
rs960070905 Chr.3: 120331142-120331142 on Build GRCh38
rs752302442 Chr.3: 120331429-120331429 on Build GRCh38
rs899729076 Chr.3: 120331084-120331084 on Build GRCh38
rs969587391 Chr.3: 120331140-120331140 on Build GRCh38
rs769726430 Chr.3: 120331329-120331329 on Build GRCh38

View Full Product Details