Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 123629388-123629896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYLK
Entrez Gene ID
4638
Gene Name
myosin light chain kinase
Gene Aliases
AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK
UniGene
Hs.477375
Gene Chromosome Location
Chr.3: 123612296-123884302 on Build GRCh38
Transcripts
NM_001321309.1 NM_053025.3 NM_053026.3 NM_053027.3 NM_053028.3 XM_011512860.2 XM_011512861.2 XM_017006469.1 XM_017006470.1 XM_017006471.1 NR_038266.2 NR_121654.1

Gene Symbol
MYLK-AS1
Entrez Gene ID
100506826
Gene Name
MYLK antisense RNA 1
Gene Aliases
-
UniGene
Hs.556600
Gene Chromosome Location
Chr.3: 123585513-123630821 on Build GRCh38
Transcripts
NM_001321309.1 NM_053025.3 NM_053026.3 NM_053027.3 NM_053028.3 XM_011512860.2 XM_011512861.2 XM_017006469.1 XM_017006470.1 XM_017006471.1 NR_038266.2 NR_121654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1418425 c.5002G>A p.E1668K Chr.3: 123629586-123629586 on Build GRCh38
6569864 c.5047G>A p.E1683K Chr.3: 123629541-123629541 on Build GRCh38
3057139 c.5107G>A p.D1703N Chr.3: 123629481-123629481 on Build GRCh38
124619 c.5071G>A p.D1691N Chr.3: 123629517-123629517 on Build GRCh38
3057140 c.5056G>A p.D1686N Chr.3: 123629532-123629532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41426350 Chr.3: 123629636-123629636 on Build GRCh38
rs1009862305 Chr.3: 123629850-123629850 on Build GRCh38
rs746510648 Chr.3: 123629672-123629672 on Build GRCh38
rs777818668 Chr.3: 123629671-123629671 on Build GRCh38
rs912331049 Chr.3: 123629726-123629726 on Build GRCh38
rs752645090 Chr.3: 123629665-123629665 on Build GRCh38
rs367656778 Chr.3: 123629605-123629605 on Build GRCh38
rs143398165 Chr.3: 123629513-123629513 on Build GRCh38
rs781190664 Chr.3: 123629535-123629535 on Build GRCh38
rs146600235 Chr.3: 123629477-123629477 on Build GRCh38
rs984532764 Chr.3: 123629818-123629818 on Build GRCh38
rs908450520 Chr.3: 123629631-123629631 on Build GRCh38
rs577501563 Chr.3: 123629573-123629573 on Build GRCh38
rs1054002712 Chr.3: 123629680-123629680 on Build GRCh38
rs201934398 Chr.3: 123629469-123629469 on Build GRCh38
rs1049913386 Chr.3: 123629710-123629710 on Build GRCh38
rs890051502 Chr.3: 123629706-123629706 on Build GRCh38
rs148364244 Chr.3: 123629533-123629533 on Build GRCh38
rs943666454 Chr.3: 123629730-123629730 on Build GRCh38
rs749571448 Chr.3: 123629870-123629870 on Build GRCh38
rs150392984 Chr.3: 123629547-123629547 on Build GRCh38
rs185783762 Chr.3: 123629670-123629670 on Build GRCh38
rs766692784 Chr.3: 123629595-123629595 on Build GRCh38
rs368571172 Chr.3: 123629521-123629521 on Build GRCh38
rs763359005 Chr.3: 123629480-123629480 on Build GRCh38
rs780756829 Chr.3: 123629675-123629675 on Build GRCh38
rs770549888 Chr.3: 123629558-123629558 on Build GRCh38
rs1052799439 Chr.3: 123629751-123629751 on Build GRCh38
rs774862641 Chr.3: 123629572-123629572 on Build GRCh38
rs374944537 Chr.3: 123629673-123629673 on Build GRCh38
rs561674421 Chr.3: 123629782-123629782 on Build GRCh38
rs750437332 Chr.3: 123629586-123629586 on Build GRCh38
rs767591736 Chr.3: 123629576-123629576 on Build GRCh38
rs151291675 Chr.3: 123629813-123629813 on Build GRCh38
rs764447883 Chr.3: 123629571-123629571 on Build GRCh38
rs541486076 Chr.3: 123629679-123629679 on Build GRCh38
rs764572695 Chr.3: 123629484-123629484 on Build GRCh38
rs111321419 Chr.3: 123629862-123629862 on Build GRCh38
rs527480711 Chr.3: 123629807-123629807 on Build GRCh38
rs754874304 Chr.3: 123629526-123629526 on Build GRCh38
rs372695434 Chr.3: 123629542-123629542 on Build GRCh38
rs769962460 Chr.3: 123629794-123629794 on Build GRCh38
rs747677351 Chr.3: 123629428-123629428 on Build GRCh38
rs763303731 Chr.3: 123629567-123629567 on Build GRCh38
rs965542616 Chr.3: 123629733-123629733 on Build GRCh38
rs555101909 Chr.3: 123629475-123629475 on Build GRCh38
rs927213113 Chr.3: 123629548-123629548 on Build GRCh38
rs879151680 Chr.3: 123629426-123629426 on Build GRCh38
rs202229368 Chr.3: 123629466-123629466 on Build GRCh38
rs776324960 Chr.3: 123629563-123629563 on Build GRCh38
rs757296690 Chr.3: 123629503-123629503 on Build GRCh38
rs141467675 Chr.3: 123629509-123629509 on Build GRCh38
rs554444072 Chr.3: 123629520-123629520 on Build GRCh38
rs535983286 Chr.3: 123629812-123629812 on Build GRCh38
rs1033041988 Chr.3: 123629437-123629437 on Build GRCh38
rs1025721182 Chr.3: 123629776-123629776 on Build GRCh38
rs904208692 Chr.3: 123629411-123629411 on Build GRCh38
rs370587479 Chr.3: 123629422-123629422 on Build GRCh38
rs898325708 Chr.3: 123629712-123629712 on Build GRCh38
rs375904060 Chr.3: 123629653-123629653 on Build GRCh38
rs56068068 Chr.3: 123629676-123629676 on Build GRCh38
rs375470297 Chr.3: 123629488-123629488 on Build GRCh38
rs756434518 Chr.3: 123629518-123629518 on Build GRCh38
rs201104217 Chr.3: 123629512-123629512 on Build GRCh38
rs771545471 Chr.3: 123629454-123629454 on Build GRCh38
rs140456660 Chr.3: 123629544-123629544 on Build GRCh38
rs1006985772 Chr.3: 123629707-123629707 on Build GRCh38
rs375038682 Chr.3: 123629587-123629587 on Build GRCh38
rs994209567 Chr.3: 123629756-123629756 on Build GRCh38
rs968157617 Chr.3: 123629541-123629541 on Build GRCh38

View Full Product Details