Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 125230435-125230934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF148
Entrez Gene ID
7707
Gene Name
zinc finger protein 148
Gene Aliases
BERF-1, BFCOL1, HT-BETA, ZBP-89, ZFP148, pHZ-52
UniGene
Hs.592591
Gene Chromosome Location
Chr.3: 125225669-125375354 on Build GRCh38
Transcripts
NM_021964.2

SNP Detail

SNP ID Chromosome Location
rs941190937 Chr.3: 125230632-125230632 on Build GRCh38
rs937572793 Chr.3: 125230657-125230657 on Build GRCh38
rs957937082 Chr.3: 125230457-125230457 on Build GRCh38
rs939258172 Chr.3: 125230574-125230574 on Build GRCh38
rs999090747 Chr.3: 125230752-125230752 on Build GRCh38
rs928609391 Chr.3: 125230859-125230859 on Build GRCh38
rs985127280 Chr.3: 125230507-125230507 on Build GRCh38
rs1056016774 Chr.3: 125230688-125230688 on Build GRCh38
rs749823067 Chr.3: 125230512-125230512 on Build GRCh38
rs890826129 Chr.3: 125230707-125230707 on Build GRCh38
rs555738769 Chr.3: 125230468-125230468 on Build GRCh38
rs1041826549 Chr.3: 125230772-125230772 on Build GRCh38
rs1032360320 Chr.3: 125230770-125230770 on Build GRCh38
rs775303161 Chr.3: 125230904-125230906 on Build GRCh38
rs879683758 Chr.3: 125230845-125230845 on Build GRCh38
rs981519441 Chr.3: 125230846-125230846 on Build GRCh38
rs941320344 Chr.3: 125230865-125230865 on Build GRCh38
rs999345117 Chr.3: 125230790-125230790 on Build GRCh38
rs150767504 Chr.3: 125230621-125230621 on Build GRCh38
rs543639649 Chr.3: 125230849-125230849 on Build GRCh38
rs1006765143 Chr.3: 125230803-125230803 on Build GRCh38
rs541436855 Chr.3: 125230800-125230800 on Build GRCh38
rs372968519 Chr.3: 125230892-125230892 on Build GRCh38
rs770692942 Chr.3: 125230629-125230632 on Build GRCh38
rs929326262 Chr.3: 125230860-125230860 on Build GRCh38
rs774830678 Chr.3: 125230871-125230871 on Build GRCh38
rs562376339 Chr.3: 125230851-125230851 on Build GRCh38
rs982680379 Chr.3: 125230862-125230862 on Build GRCh38
rs778271652 Chr.3: 125230476-125230476 on Build GRCh38
rs996636821 Chr.3: 125230693-125230693 on Build GRCh38
rs976805911 Chr.3: 125230906-125230906 on Build GRCh38
rs1038086746 Chr.3: 125230635-125230635 on Build GRCh38
rs554661240 Chr.3: 125230585-125230585 on Build GRCh38
rs774263998 Chr.3: 125230736-125230736 on Build GRCh38
rs895822744 Chr.3: 125230702-125230702 on Build GRCh38
rs748397575 Chr.3: 125230611-125230611 on Build GRCh38
rs930151836 Chr.3: 125230618-125230618 on Build GRCh38
rs970113879 Chr.3: 125230819-125230819 on Build GRCh38
rs114482952 Chr.3: 125230629-125230629 on Build GRCh38
rs375425289 Chr.3: 125230838-125230838 on Build GRCh38
rs928679688 Chr.3: 125230546-125230546 on Build GRCh38
rs908460587 Chr.3: 125230864-125230864 on Build GRCh38
rs1029315944 Chr.3: 125230699-125230699 on Build GRCh38
rs1014639208 Chr.3: 125230763-125230763 on Build GRCh38
rs899573886 Chr.3: 125230641-125230641 on Build GRCh38
rs753022505 Chr.3: 125230860-125230860 on Build GRCh38
rs965377122 Chr.3: 125230578-125230578 on Build GRCh38
rs948689447 Chr.3: 125230494-125230494 on Build GRCh38
rs576600464 Chr.3: 125230806-125230806 on Build GRCh38
rs771983454 Chr.3: 125230816-125230816 on Build GRCh38
rs560651236 Chr.3: 125230705-125230705 on Build GRCh38
rs185363972 Chr.3: 125230650-125230650 on Build GRCh38
rs538577643 Chr.3: 125230517-125230517 on Build GRCh38
rs903249686 Chr.3: 125230773-125230773 on Build GRCh38
rs957522882 Chr.3: 125230794-125230794 on Build GRCh38

View Full Product Details