Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 125529834-125530378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSBPL11
Entrez Gene ID
114885
Gene Name
oxysterol binding protein like 11
Gene Aliases
ORP-11, ORP11, OSBP12, TCCCIA00292
UniGene
Hs.477440
Gene Chromosome Location
Chr.3: 125528858-125595537 on Build GRCh38
Transcripts
NM_022776.4

SNP Detail

SNP ID Chromosome Location
rs868225052 Chr.3: 125530068-125530068 on Build GRCh38
rs527992245 Chr.3: 125529924-125529924 on Build GRCh38
rs780768599 Chr.3: 125530225-125530225 on Build GRCh38
rs986258582 Chr.3: 125530328-125530328 on Build GRCh38
rs534812148 Chr.3: 125530267-125530267 on Build GRCh38
rs573937815 Chr.3: 125530265-125530265 on Build GRCh38
rs11458216 Chr.3: 125529994-125529995 on Build GRCh38
rs911944168 Chr.3: 125530332-125530332 on Build GRCh38
rs533403015 Chr.3: 125530015-125530015 on Build GRCh38
rs79254144 Chr.3: 125530006-125530006 on Build GRCh38
rs978899244 Chr.3: 125529968-125529968 on Build GRCh38
rs746905848 Chr.3: 125529916-125529916 on Build GRCh38
rs895512886 Chr.3: 125530233-125530233 on Build GRCh38
rs953431087 Chr.3: 125529991-125529991 on Build GRCh38
rs1057027074 Chr.3: 125530331-125530331 on Build GRCh38
rs1040987850 Chr.3: 125530079-125530079 on Build GRCh38
rs564978172 Chr.3: 125530054-125530054 on Build GRCh38
rs1033103165 Chr.3: 125530319-125530319 on Build GRCh38
rs185536792 Chr.3: 125530204-125530204 on Build GRCh38
rs905428626 Chr.3: 125530277-125530277 on Build GRCh38
rs944444599 Chr.3: 125530152-125530152 on Build GRCh38
rs1001268824 Chr.3: 125530200-125530200 on Build GRCh38
rs187508619 Chr.3: 125530263-125530263 on Build GRCh38
rs953211435 Chr.3: 125530057-125530057 on Build GRCh38
rs768428765 Chr.3: 125530018-125530018 on Build GRCh38
rs747678767 Chr.3: 125530242-125530243 on Build GRCh38
rs1020481386 Chr.3: 125529946-125529946 on Build GRCh38
rs973303869 Chr.3: 125530042-125530042 on Build GRCh38
rs547823253 Chr.3: 125529935-125529935 on Build GRCh38
rs397705160 Chr.3: 125530005-125530006 on Build GRCh38
rs571117383 Chr.3: 125529986-125529986 on Build GRCh38
rs11544320 Chr.3: 125530155-125530155 on Build GRCh38
rs1035127675 Chr.3: 125529865-125529865 on Build GRCh38
rs1000330558 Chr.3: 125530317-125530317 on Build GRCh38
rs911653080 Chr.3: 125530098-125530098 on Build GRCh38
rs35676757 Chr.3: 125530287-125530288 on Build GRCh38
rs192024253 Chr.3: 125530313-125530313 on Build GRCh38
rs967543959 Chr.3: 125529950-125529950 on Build GRCh38
rs961042642 Chr.3: 125529867-125529867 on Build GRCh38
rs888198958 Chr.3: 125530185-125530185 on Build GRCh38
rs550179468 Chr.3: 125530130-125530130 on Build GRCh38
rs111748383 Chr.3: 125530335-125530335 on Build GRCh38
rs375040206 Chr.3: 125530178-125530180 on Build GRCh38
rs1014479751 Chr.3: 125529923-125529923 on Build GRCh38
rs953598042 Chr.3: 125530322-125530322 on Build GRCh38
rs986328144 Chr.3: 125530090-125530090 on Build GRCh38
rs193258072 Chr.3: 125530191-125530191 on Build GRCh38
rs145177475 Chr.3: 125530005-125530005 on Build GRCh38
rs555544463 Chr.3: 125530247-125530247 on Build GRCh38
rs938190806 Chr.3: 125530316-125530316 on Build GRCh38
rs553871331 Chr.3: 125530260-125530260 on Build GRCh38
rs1027545323 Chr.3: 125530053-125530053 on Build GRCh38
rs527642863 Chr.3: 125530100-125530100 on Build GRCh38

View Full Product Details