Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 125551896-125552395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSBPL11
Entrez Gene ID
114885
Gene Name
oxysterol binding protein like 11
Gene Aliases
ORP-11, ORP11, OSBP12, TCCCIA00292
UniGene
Hs.477440
Gene Chromosome Location
Chr.3: 125528858-125595537 on Build GRCh38
Transcripts
NM_022776.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7192779 c.1594C>T p.H532Y Chr.3: 125552241-125552241 on Build GRCh38
3058287 c.1635C>T p.G545G Chr.3: 125552200-125552200 on Build GRCh38
1037670 c.1617C>A p.F539L Chr.3: 125552218-125552218 on Build GRCh38
1670531 c.1570G>T p.E524* Chr.3: 125552265-125552265 on Build GRCh38
1218916 c.1616T>G p.F539C Chr.3: 125552219-125552219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746998693 Chr.3: 125552142-125552149 on Build GRCh38
rs1055490628 Chr.3: 125552133-125552133 on Build GRCh38
rs200224916 Chr.3: 125552110-125552111 on Build GRCh38
rs753174656 Chr.3: 125552327-125552327 on Build GRCh38
rs578002517 Chr.3: 125552243-125552243 on Build GRCh38
rs199706756 Chr.3: 125552145-125552145 on Build GRCh38
rs756663222 Chr.3: 125552131-125552132 on Build GRCh38
rs2979371 Chr.3: 125551984-125551984 on Build GRCh38
rs780875300 Chr.3: 125552152-125552152 on Build GRCh38
rs1039446563 Chr.3: 125552111-125552111 on Build GRCh38
rs1019037013 Chr.3: 125552068-125552068 on Build GRCh38
rs761338505 Chr.3: 125552257-125552257 on Build GRCh38
rs755334451 Chr.3: 125552160-125552160 on Build GRCh38
rs1049305879 Chr.3: 125552158-125552158 on Build GRCh38
rs924622916 Chr.3: 125552064-125552064 on Build GRCh38
rs904231643 Chr.3: 125552029-125552029 on Build GRCh38
rs60664244 Chr.3: 125552158-125552161 on Build GRCh38
rs966058659 Chr.3: 125552070-125552070 on Build GRCh38
rs10541213 Chr.3: 125552142-125552145 on Build GRCh38
rs778140217 Chr.3: 125552214-125552214 on Build GRCh38
rs541038110 Chr.3: 125552203-125552203 on Build GRCh38
rs1015156409 Chr.3: 125551983-125551983 on Build GRCh38
rs1009615592 Chr.3: 125552319-125552319 on Build GRCh38
rs915083230 Chr.3: 125552097-125552097 on Build GRCh38
rs78040650 Chr.3: 125552166-125552166 on Build GRCh38
rs945956332 Chr.3: 125552089-125552089 on Build GRCh38
rs989369230 Chr.3: 125552236-125552236 on Build GRCh38
rs765936259 Chr.3: 125552287-125552287 on Build GRCh38
rs762878129 Chr.3: 125552136-125552136 on Build GRCh38
rs779394620 Chr.3: 125552080-125552080 on Build GRCh38
rs768536012 Chr.3: 125552160-125552161 on Build GRCh38
rs201489753 Chr.3: 125552143-125552143 on Build GRCh38
rs72038182 Chr.3: 125552147-125552150 on Build GRCh38
rs1013875449 Chr.3: 125551939-125551939 on Build GRCh38
rs779271741 Chr.3: 125552199-125552199 on Build GRCh38
rs556756770 Chr.3: 125552052-125552052 on Build GRCh38
rs762447928 Chr.3: 125552141-125552144 on Build GRCh38
rs867804529 Chr.3: 125552301-125552301 on Build GRCh38
rs750032510 Chr.3: 125552159-125552159 on Build GRCh38
rs377242399 Chr.3: 125552349-125552349 on Build GRCh38
rs750875557 Chr.3: 125552144-125552145 on Build GRCh38
rs757443297 Chr.3: 125552361-125552361 on Build GRCh38
rs757994615 Chr.3: 125552142-125552143 on Build GRCh38
rs924646572 Chr.3: 125552190-125552190 on Build GRCh38
rs757694954 Chr.3: 125551994-125551994 on Build GRCh38
rs766977557 Chr.3: 125552262-125552262 on Build GRCh38
rs77761851 Chr.3: 125552120-125552120 on Build GRCh38
rs1028239137 Chr.3: 125552015-125552015 on Build GRCh38
rs751479684 Chr.3: 125552256-125552256 on Build GRCh38
rs755734850 Chr.3: 125552274-125552274 on Build GRCh38
rs966125146 Chr.3: 125552293-125552293 on Build GRCh38
rs895522310 Chr.3: 125551930-125551930 on Build GRCh38
rs777015706 Chr.3: 125552240-125552240 on Build GRCh38
rs895564743 Chr.3: 125552135-125552135 on Build GRCh38
rs769477065 Chr.3: 125552242-125552242 on Build GRCh38
rs754214160 Chr.3: 125552140-125552147 on Build GRCh38
rs988127216 Chr.3: 125552071-125552071 on Build GRCh38
rs745496035 Chr.3: 125552142-125552147 on Build GRCh38
rs977527056 Chr.3: 125552172-125552172 on Build GRCh38
rs771300850 Chr.3: 125552224-125552224 on Build GRCh38
rs1047201429 Chr.3: 125552126-125552126 on Build GRCh38
rs942602423 Chr.3: 125552366-125552366 on Build GRCh38
rs572429737 Chr.3: 125552200-125552200 on Build GRCh38
rs200206436 Chr.3: 125552141-125552141 on Build GRCh38
rs756731716 Chr.3: 125552147-125552147 on Build GRCh38
rs758658010 Chr.3: 125552335-125552335 on Build GRCh38
rs778472130 Chr.3: 125552348-125552348 on Build GRCh38
rs540041855 Chr.3: 125552026-125552026 on Build GRCh38
rs190608562 Chr.3: 125552131-125552131 on Build GRCh38
rs978676251 Chr.3: 125552106-125552106 on Build GRCh38
rs867251984 Chr.3: 125552176-125552176 on Build GRCh38
rs750080262 Chr.3: 125552265-125552265 on Build GRCh38
rs971443031 Chr.3: 125552055-125552055 on Build GRCh38
rs900952124 Chr.3: 125552122-125552123 on Build GRCh38
rs576814146 Chr.3: 125552085-125552085 on Build GRCh38
rs756973883 Chr.3: 125552140-125552143 on Build GRCh38
rs150910514 Chr.3: 125552140-125552145 on Build GRCh38
rs753284839 Chr.3: 125552132-125552133 on Build GRCh38
rs564619745 Chr.3: 125552221-125552221 on Build GRCh38
rs779401692 Chr.3: 125552141-125552142 on Build GRCh38
rs759788380 Chr.3: 125552241-125552241 on Build GRCh38
rs551482946 Chr.3: 125551981-125551981 on Build GRCh38
rs186979174 Chr.3: 125552108-125552108 on Build GRCh38
rs958876722 Chr.3: 125552146-125552146 on Build GRCh38
rs79433472 Chr.3: 125552119-125552119 on Build GRCh38
rs997930927 Chr.3: 125552123-125552124 on Build GRCh38

View Full Product Details