Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128080989-128081493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RUVBL1
Entrez Gene ID
8607
Gene Name
RuvB like AAA ATPase 1
Gene Aliases
ECP-54, ECP54, INO80H, NMP 238, NMP238, PONTIN, Pontin52, RVB1, TIH1, TIP49, TIP49A
UniGene
Hs.272822
Gene Chromosome Location
Chr.3: 128064612-128153914 on Build GRCh38
Transcripts
NM_001319084.1 NM_001319086.1 NM_003707.2 XM_005247841.3 XM_011513248.1 XM_011513249.2 XM_017007356.1 XM_017007357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5749229 c.1259A>G p.N420S Chr.3: 128081362-128081362 on Build GRCh38
230532 c.1361A>T p.Y454F Chr.3: 128081260-128081260 on Build GRCh38
6511508 c.1311C>T p.F437F Chr.3: 128081310-128081310 on Build GRCh38
445525 c.1272C>T p.S424S Chr.3: 128081349-128081349 on Build GRCh38
6380435 c.1268A>G p.D423G Chr.3: 128081353-128081353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370080824 Chr.3: 128081243-128081243 on Build GRCh38
rs1057156 Chr.3: 128081228-128081228 on Build GRCh38
rs372381267 Chr.3: 128081331-128081331 on Build GRCh38
rs764386458 Chr.3: 128081091-128081091 on Build GRCh38
rs750497962 Chr.3: 128081270-128081270 on Build GRCh38
rs983025702 Chr.3: 128081159-128081159 on Build GRCh38
rs749767641 Chr.3: 128081428-128081428 on Build GRCh38
rs779351228 Chr.3: 128081440-128081440 on Build GRCh38
rs763641171 Chr.3: 128081417-128081417 on Build GRCh38
rs72974082 Chr.3: 128081077-128081077 on Build GRCh38
rs780805649 Chr.3: 128081425-128081425 on Build GRCh38
rs982691759 Chr.3: 128081448-128081448 on Build GRCh38
rs751296279 Chr.3: 128081419-128081419 on Build GRCh38
rs535682052 Chr.3: 128081453-128081453 on Build GRCh38
rs748533150 Chr.3: 128081443-128081443 on Build GRCh38
rs749854545 Chr.3: 128081347-128081347 on Build GRCh38
rs754761052 Chr.3: 128081302-128081302 on Build GRCh38
rs375523902 Chr.3: 128081461-128081461 on Build GRCh38
rs368834831 Chr.3: 128081454-128081454 on Build GRCh38
rs1017966304 Chr.3: 128081081-128081081 on Build GRCh38
rs569769466 Chr.3: 128081184-128081184 on Build GRCh38
rs778523801 Chr.3: 128081214-128081214 on Build GRCh38
rs1802411 Chr.3: 128081249-128081249 on Build GRCh38
rs949485193 Chr.3: 128081183-128081183 on Build GRCh38
rs754566615 Chr.3: 128081208-128081208 on Build GRCh38
rs773281133 Chr.3: 128081366-128081366 on Build GRCh38
rs760657332 Chr.3: 128081385-128081385 on Build GRCh38
rs775866819 Chr.3: 128081229-128081229 on Build GRCh38
rs887477871 Chr.3: 128081147-128081147 on Build GRCh38
rs774685925 Chr.3: 128081235-128081235 on Build GRCh38
rs766547190 Chr.3: 128081386-128081386 on Build GRCh38
rs956905462 Chr.3: 128081088-128081088 on Build GRCh38
rs142162397 Chr.3: 128081132-128081132 on Build GRCh38
rs890597585 Chr.3: 128081313-128081313 on Build GRCh38
rs201991974 Chr.3: 128081215-128081215 on Build GRCh38
rs15373 Chr.3: 128081120-128081120 on Build GRCh38
rs770891695 Chr.3: 128081460-128081460 on Build GRCh38
rs777170930 Chr.3: 128081218-128081218 on Build GRCh38
rs892748496 Chr.3: 128081288-128081288 on Build GRCh38
rs759517387 Chr.3: 128081394-128081394 on Build GRCh38
rs765055859 Chr.3: 128081308-128081308 on Build GRCh38
rs780424922 Chr.3: 128081338-128081338 on Build GRCh38
rs767652542 Chr.3: 128081258-128081258 on Build GRCh38
rs376409650 Chr.3: 128081336-128081336 on Build GRCh38
rs6779679 Chr.3: 128081092-128081092 on Build GRCh38
rs937144063 Chr.3: 128081468-128081468 on Build GRCh38
rs552198810 Chr.3: 128081226-128081226 on Build GRCh38
rs769214063 Chr.3: 128081234-128081234 on Build GRCh38
rs757988824 Chr.3: 128081416-128081416 on Build GRCh38
rs917725149 Chr.3: 128081161-128081161 on Build GRCh38
rs949915631 Chr.3: 128081121-128081121 on Build GRCh38
rs115163125 Chr.3: 128081334-128081334 on Build GRCh38
rs879231486 Chr.3: 128081324-128081324 on Build GRCh38
rs566226179 Chr.3: 128081093-128081093 on Build GRCh38
rs988505713 Chr.3: 128081437-128081437 on Build GRCh38
rs927102668 Chr.3: 128081133-128081133 on Build GRCh38
rs951384599 Chr.3: 128081447-128081447 on Build GRCh38
rs769123849 Chr.3: 128081358-128081358 on Build GRCh38
rs776517604 Chr.3: 128081391-128081391 on Build GRCh38
rs927133878 Chr.3: 128081462-128081462 on Build GRCh38
rs917968115 Chr.3: 128081108-128081108 on Build GRCh38
rs756694637 Chr.3: 128081424-128081424 on Build GRCh38
rs562008395 Chr.3: 128081105-128081105 on Build GRCh38
rs752298560 Chr.3: 128081318-128081318 on Build GRCh38
rs763399487 Chr.3: 128081230-128081230 on Build GRCh38
rs752496410 Chr.3: 128081402-128081402 on Build GRCh38
rs371363267 Chr.3: 128081154-128081154 on Build GRCh38
rs940321350 Chr.3: 128081156-128081156 on Build GRCh38
rs764840101 Chr.3: 128081144-128081144 on Build GRCh38
rs955300556 Chr.3: 128081053-128081053 on Build GRCh38
rs772183542 Chr.3: 128081362-128081362 on Build GRCh38
rs369637214 Chr.3: 128081360-128081360 on Build GRCh38
rs1057220 Chr.3: 128081140-128081140 on Build GRCh38
rs375970015 Chr.3: 128081452-128081452 on Build GRCh38
rs372747593 Chr.3: 128081401-128081401 on Build GRCh38
rs766239636 Chr.3: 128081291-128081291 on Build GRCh38
rs748338865 Chr.3: 128081361-128081361 on Build GRCh38
rs760590643 Chr.3: 128081287-128081287 on Build GRCh38
rs753870834 Chr.3: 128081292-128081292 on Build GRCh38
rs755571111 Chr.3: 128081429-128081429 on Build GRCh38

View Full Product Details