Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128575723-128576218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01565
Entrez Gene ID
23434
Gene Name
long intergenic non-protein coding RNA 1565
Gene Aliases
C3orf27, GR6
UniGene
Hs.194283
Gene Chromosome Location
Chr.3: 128572000-128576086 on Build GRCh38
Transcripts
NR_125802.1

SNP Detail

SNP ID Chromosome Location
rs575439095 Chr.3: 128576110-128576110 on Build GRCh38
rs139226944 Chr.3: 128575787-128575787 on Build GRCh38
rs1032354488 Chr.3: 128576081-128576081 on Build GRCh38
rs535191137 Chr.3: 128576065-128576065 on Build GRCh38
rs988652983 Chr.3: 128576128-128576128 on Build GRCh38
rs935408020 Chr.3: 128575907-128575907 on Build GRCh38
rs960882868 Chr.3: 128575793-128575793 on Build GRCh38
rs537195877 Chr.3: 128575911-128575911 on Build GRCh38
rs935992764 Chr.3: 128575962-128575962 on Build GRCh38
rs767759010 Chr.3: 128576167-128576167 on Build GRCh38
rs2712415 Chr.3: 128576022-128576022 on Build GRCh38
rs2465354 Chr.3: 128576054-128576054 on Build GRCh38
rs984400698 Chr.3: 128575804-128575804 on Build GRCh38
rs993897309 Chr.3: 128576003-128576003 on Build GRCh38
rs1026246435 Chr.3: 128575792-128575792 on Build GRCh38
rs779938072 Chr.3: 128575758-128575758 on Build GRCh38
rs569872802 Chr.3: 128575892-128575892 on Build GRCh38
rs556599419 Chr.3: 128576082-128576082 on Build GRCh38
rs564517465 Chr.3: 128576197-128576197 on Build GRCh38
rs923366217 Chr.3: 128576133-128576133 on Build GRCh38
rs913072686 Chr.3: 128575946-128575946 on Build GRCh38
rs1015229545 Chr.3: 128576186-128576186 on Build GRCh38
rs969830529 Chr.3: 128575857-128575857 on Build GRCh38
rs2734019 Chr.3: 128576163-128576163 on Build GRCh38
rs7626811 Chr.3: 128575956-128575956 on Build GRCh38
rs961657177 Chr.3: 128576188-128576188 on Build GRCh38
rs1016672576 Chr.3: 128575786-128575786 on Build GRCh38
rs534598209 Chr.3: 128576011-128576011 on Build GRCh38
rs932783503 Chr.3: 128575921-128575921 on Build GRCh38
rs993838588 Chr.3: 128575790-128575790 on Build GRCh38
rs1051480734 Chr.3: 128575955-128575955 on Build GRCh38
rs969633475 Chr.3: 128576061-128576061 on Build GRCh38
rs976948999 Chr.3: 128575868-128575868 on Build GRCh38
rs940242878 Chr.3: 128575862-128575862 on Build GRCh38
rs896676295 Chr.3: 128576029-128576029 on Build GRCh38
rs755434291 Chr.3: 128575869-128575869 on Build GRCh38
rs76259946 Chr.3: 128576127-128576127 on Build GRCh38
rs1006550534 Chr.3: 128575963-128575963 on Build GRCh38
rs190354509 Chr.3: 128576104-128576104 on Build GRCh38
rs747113930 Chr.3: 128575805-128575805 on Build GRCh38
rs559030862 Chr.3: 128576121-128576121 on Build GRCh38
rs777475457 Chr.3: 128576123-128576123 on Build GRCh38
rs889472205 Chr.3: 128575751-128575751 on Build GRCh38
rs1009417335 Chr.3: 128575759-128575759 on Build GRCh38
rs552083402 Chr.3: 128575937-128575937 on Build GRCh38
rs770183928 Chr.3: 128576073-128576073 on Build GRCh38
rs2713596 Chr.3: 128576198-128576198 on Build GRCh38
rs529426679 Chr.3: 128575778-128575778 on Build GRCh38
rs1038002498 Chr.3: 128575975-128575975 on Build GRCh38
rs898137903 Chr.3: 128575999-128575999 on Build GRCh38
rs933851302 Chr.3: 128576148-128576148 on Build GRCh38
rs933819602 Chr.3: 128575951-128575951 on Build GRCh38
rs185164887 Chr.3: 128576018-128576018 on Build GRCh38
rs1006497296 Chr.3: 128575752-128575752 on Build GRCh38
rs561911293 Chr.3: 128575757-128575757 on Build GRCh38
rs1052488039 Chr.3: 128575919-128575919 on Build GRCh38
rs2713595 Chr.3: 128576146-128576146 on Build GRCh38
rs908806927 Chr.3: 128575855-128575855 on Build GRCh38
rs1024349029 Chr.3: 128576055-128576055 on Build GRCh38
rs1025207939 Chr.3: 128575777-128575777 on Build GRCh38
rs916827098 Chr.3: 128575817-128575817 on Build GRCh38
rs547015519 Chr.3: 128575756-128575756 on Build GRCh38

View Full Product Details