Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 131018366-131018877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTE1
Entrez Gene ID
28990
Gene Name
asteroid homolog 1 (Drosophila)
Gene Aliases
HT001
UniGene
Hs.100878
Gene Chromosome Location
Chr.3: 131013873-131026854 on Build GRCh38
Transcripts
NM_001288950.1 NM_014065.3 NR_110229.1 XM_017006266.1 XM_017006267.1 XM_017006268.1 XM_017006269.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4619803 c.1500A>C p.I500I Chr.3: 131018519-131018519 on Build GRCh38
3944857 c.1321C>T p.Q441* Chr.3: 131018698-131018698 on Build GRCh38
5398174 c.1510C>T p.P504S Chr.3: 131018509-131018509 on Build GRCh38
4583945 c.1315A>C p.R439R Chr.3: 131018704-131018704 on Build GRCh38
5350897 c.1483G>T p.G495W Chr.3: 131018536-131018536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901391433 Chr.3: 131018846-131018847 on Build GRCh38
rs776729377 Chr.3: 131018609-131018609 on Build GRCh38
rs745757966 Chr.3: 131018622-131018622 on Build GRCh38
rs368067508 Chr.3: 131018712-131018712 on Build GRCh38
rs218503 Chr.3: 131018788-131018788 on Build GRCh38
rs754307526 Chr.3: 131018683-131018683 on Build GRCh38
rs745936746 Chr.3: 131018482-131018484 on Build GRCh38
rs367878698 Chr.3: 131018744-131018744 on Build GRCh38
rs773420294 Chr.3: 131018494-131018494 on Build GRCh38
rs560359533 Chr.3: 131018590-131018590 on Build GRCh38
rs749622274 Chr.3: 131018462-131018462 on Build GRCh38
rs763709345 Chr.3: 131018649-131018649 on Build GRCh38
rs950175716 Chr.3: 131018831-131018831 on Build GRCh38
rs766958752 Chr.3: 131018666-131018666 on Build GRCh38
rs938073561 Chr.3: 131018765-131018765 on Build GRCh38
rs763382554 Chr.3: 131018422-131018422 on Build GRCh38
rs759277815 Chr.3: 131018516-131018516 on Build GRCh38
rs758633153 Chr.3: 131018573-131018573 on Build GRCh38
rs929264657 Chr.3: 131018406-131018406 on Build GRCh38
rs148013706 Chr.3: 131018557-131018557 on Build GRCh38
rs759652851 Chr.3: 131018584-131018584 on Build GRCh38
rs762566062 Chr.3: 131018647-131018647 on Build GRCh38
rs374756661 Chr.3: 131018461-131018461 on Build GRCh38
rs973248092 Chr.3: 131018715-131018715 on Build GRCh38
rs534138604 Chr.3: 131018814-131018814 on Build GRCh38
rs372033215 Chr.3: 131018601-131018601 on Build GRCh38
rs775345305 Chr.3: 131018635-131018635 on Build GRCh38
rs927451792 Chr.3: 131018848-131018848 on Build GRCh38
rs17282320 Chr.3: 131018835-131018835 on Build GRCh38
rs531359525 Chr.3: 131018752-131018752 on Build GRCh38
rs778047224 Chr.3: 131018588-131018588 on Build GRCh38
rs775911203 Chr.3: 131018566-131018568 on Build GRCh38
rs748383591 Chr.3: 131018560-131018560 on Build GRCh38
rs779027365 Chr.3: 131018470-131018470 on Build GRCh38
rs148800707 Chr.3: 131018489-131018489 on Build GRCh38
rs201151851 Chr.3: 131018505-131018505 on Build GRCh38
rs1055263908 Chr.3: 131018710-131018710 on Build GRCh38
rs747434653 Chr.3: 131018725-131018728 on Build GRCh38
rs1046493778 Chr.3: 131018415-131018415 on Build GRCh38
rs193049914 Chr.3: 131018748-131018748 on Build GRCh38
rs765217896 Chr.3: 131018490-131018491 on Build GRCh38
rs758588526 Chr.3: 131018702-131018702 on Build GRCh38
rs765030322 Chr.3: 131018519-131018519 on Build GRCh38
rs888738412 Chr.3: 131018487-131018487 on Build GRCh38
rs757424449 Chr.3: 131018706-131018706 on Build GRCh38
rs768857716 Chr.3: 131018746-131018746 on Build GRCh38
rs770966234 Chr.3: 131018501-131018501 on Build GRCh38
rs143427732 Chr.3: 131018548-131018548 on Build GRCh38
rs755504198 Chr.3: 131018687-131018687 on Build GRCh38
rs544261075 Chr.3: 131018429-131018429 on Build GRCh38
rs779758381 Chr.3: 131018739-131018739 on Build GRCh38
rs772582652 Chr.3: 131018518-131018518 on Build GRCh38
rs748788320 Chr.3: 131018576-131018576 on Build GRCh38
rs774357333 Chr.3: 131018570-131018570 on Build GRCh38
rs761336147 Chr.3: 131018665-131018665 on Build GRCh38
rs372161995 Chr.3: 131018729-131018729 on Build GRCh38
rs762665001 Chr.3: 131018530-131018530 on Build GRCh38
rs747000434 Chr.3: 131018498-131018498 on Build GRCh38
rs375067832 Chr.3: 131018704-131018704 on Build GRCh38
rs769751757 Chr.3: 131018737-131018737 on Build GRCh38
rs773953587 Chr.3: 131018653-131018653 on Build GRCh38
rs769546265 Chr.3: 131018623-131018623 on Build GRCh38
rs983824014 Chr.3: 131018787-131018787 on Build GRCh38
rs775014075 Chr.3: 131018529-131018529 on Build GRCh38
rs769025040 Chr.3: 131018463-131018463 on Build GRCh38
rs527868033 Chr.3: 131018700-131018700 on Build GRCh38
rs1037302856 Chr.3: 131018836-131018836 on Build GRCh38
rs755312386 Chr.3: 131018553-131018553 on Build GRCh38
rs763574548 Chr.3: 131018538-131018538 on Build GRCh38
rs950615193 Chr.3: 131018766-131018766 on Build GRCh38
rs556717620 Chr.3: 131018669-131018669 on Build GRCh38
rs964570443 Chr.3: 131018714-131018714 on Build GRCh38
rs545207928 Chr.3: 131018549-131018549 on Build GRCh38
rs954607750 Chr.3: 131018659-131018659 on Build GRCh38
rs776603269 Chr.3: 131018507-131018507 on Build GRCh38
rs920581936 Chr.3: 131018390-131018390 on Build GRCh38
rs778169586 Chr.3: 131018703-131018703 on Build GRCh38
rs748345801 Chr.3: 131018474-131018474 on Build GRCh38
rs548700211 Chr.3: 131018701-131018701 on Build GRCh38
rs988079159 Chr.3: 131018602-131018602 on Build GRCh38
rs78238509 Chr.3: 131018539-131018539 on Build GRCh38
rs771090825 Chr.3: 131018591-131018591 on Build GRCh38
rs375832359 Chr.3: 131018619-131018619 on Build GRCh38

View Full Product Details