Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133573382-133573869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDV3
Entrez Gene ID
55573
Gene Name
CDV3 homolog
Gene Aliases
H41
UniGene
Hs.518265
Gene Chromosome Location
Chr.3: 133573563-133590274 on Build GRCh38
Transcripts
NM_001134422.1 NM_001282762.1 NM_017548.4 XM_005247588.4 XM_005247589.4 XM_005247590.4 XM_005247591.4 XM_011512961.2

SNP Detail

SNP ID Chromosome Location
rs183107326 Chr.3: 133573432-133573432 on Build GRCh38
rs991896259 Chr.3: 133573800-133573801 on Build GRCh38
rs993861299 Chr.3: 133573841-133573841 on Build GRCh38
rs952601384 Chr.3: 133573702-133573702 on Build GRCh38
rs930980616 Chr.3: 133573641-133573641 on Build GRCh38
rs982322294 Chr.3: 133573613-133573613 on Build GRCh38
rs750941401 Chr.3: 133573637-133573637 on Build GRCh38
rs975680206 Chr.3: 133573741-133573741 on Build GRCh38
rs984085310 Chr.3: 133573558-133573558 on Build GRCh38
rs986849303 Chr.3: 133573732-133573732 on Build GRCh38
rs1025470989 Chr.3: 133573545-133573545 on Build GRCh38
rs542459749 Chr.3: 133573532-133573532 on Build GRCh38
rs370300187 Chr.3: 133573458-133573458 on Build GRCh38
rs1040030276 Chr.3: 133573431-133573431 on Build GRCh38
rs545704350 Chr.3: 133573638-133573638 on Build GRCh38
rs1013247869 Chr.3: 133573662-133573662 on Build GRCh38
rs576980932 Chr.3: 133573627-133573627 on Build GRCh38
rs563408959 Chr.3: 133573618-133573618 on Build GRCh38
rs758810319 Chr.3: 133573688-133573688 on Build GRCh38
rs904716618 Chr.3: 133573664-133573664 on Build GRCh38
rs906238366 Chr.3: 133573805-133573805 on Build GRCh38
rs886887190 Chr.3: 133573462-133573462 on Build GRCh38
rs1056415642 Chr.3: 133573793-133573794 on Build GRCh38
rs751991651 Chr.3: 133573710-133573710 on Build GRCh38
rs976753108 Chr.3: 133573745-133573745 on Build GRCh38
rs1046684291 Chr.3: 133573654-133573654 on Build GRCh38
rs548505487 Chr.3: 133573744-133573744 on Build GRCh38
rs766681404 Chr.3: 133573700-133573700 on Build GRCh38
rs990580974 Chr.3: 133573519-133573519 on Build GRCh38
rs71317414 Chr.3: 133573588-133573588 on Build GRCh38
rs553496974 Chr.3: 133573477-133573477 on Build GRCh38
rs539677486 Chr.3: 133573464-133573464 on Build GRCh38
rs963892841 Chr.3: 133573736-133573736 on Build GRCh38
rs886793807 Chr.3: 133573658-133573658 on Build GRCh38
rs912673307 Chr.3: 133573734-133573734 on Build GRCh38
rs868799512 Chr.3: 133573411-133573411 on Build GRCh38
rs936194735 Chr.3: 133573772-133573772 on Build GRCh38
rs1024401983 Chr.3: 133573467-133573467 on Build GRCh38
rs986901475 Chr.3: 133573554-133573554 on Build GRCh38
rs964358523 Chr.3: 133573717-133573717 on Build GRCh38
rs1002147117 Chr.3: 133573670-133573670 on Build GRCh38
rs1056066678 Chr.3: 133573676-133573676 on Build GRCh38
rs1001758358 Chr.3: 133573479-133573479 on Build GRCh38
rs950342112 Chr.3: 133573801-133573809 on Build GRCh38
rs928086506 Chr.3: 133573617-133573617 on Build GRCh38
rs1011851207 Chr.3: 133573706-133573706 on Build GRCh38
rs188856134 Chr.3: 133573530-133573530 on Build GRCh38
rs559452916 Chr.3: 133573691-133573691 on Build GRCh38
rs1046829435 Chr.3: 133573842-133573843 on Build GRCh38
rs967374329 Chr.3: 133573720-133573720 on Build GRCh38
rs549361313 Chr.3: 133573801-133573801 on Build GRCh38
rs942609433 Chr.3: 133573412-133573412 on Build GRCh38
rs200294568 Chr.3: 133573621-133573622 on Build GRCh38
rs894772937 Chr.3: 133573677-133573677 on Build GRCh38
rs952732220 Chr.3: 133573546-133573546 on Build GRCh38
rs993910729 Chr.3: 133573694-133573694 on Build GRCh38
rs71317412 Chr.3: 133573436-133573436 on Build GRCh38
rs912360577 Chr.3: 133573557-133573557 on Build GRCh38
rs766419041 Chr.3: 133573489-133573489 on Build GRCh38
rs1046137019 Chr.3: 133573667-133573667 on Build GRCh38
rs778176078 Chr.3: 133573844-133573844 on Build GRCh38
rs894832611 Chr.3: 133573461-133573461 on Build GRCh38
rs967441135 Chr.3: 133573762-133573762 on Build GRCh38
rs1046092678 Chr.3: 133573683-133573683 on Build GRCh38
rs111722096 Chr.3: 133573695-133573695 on Build GRCh38
rs977525176 Chr.3: 133573764-133573764 on Build GRCh38
rs71317413 Chr.3: 133573541-133573541 on Build GRCh38
rs373449225 Chr.3: 133573599-133573599 on Build GRCh38
rs574523950 Chr.3: 133573556-133573556 on Build GRCh38
rs907359163 Chr.3: 133573668-133573668 on Build GRCh38
rs1011874824 Chr.3: 133573679-133573679 on Build GRCh38
rs1000485103 Chr.3: 133573450-133573450 on Build GRCh38
rs922638689 Chr.3: 133573759-133573759 on Build GRCh38
rs370687204 Chr.3: 133573766-133573766 on Build GRCh38
rs1005858596 Chr.3: 133573704-133573704 on Build GRCh38
rs957392261 Chr.3: 133573500-133573500 on Build GRCh38
rs529595638 Chr.3: 133573697-133573697 on Build GRCh38
rs765373028 Chr.3: 133573586-133573586 on Build GRCh38
rs971973373 Chr.3: 133573472-133573472 on Build GRCh38
rs549735511 Chr.3: 133573419-133573419 on Build GRCh38

View Full Product Details