Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110947929-110948445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRIF1
Entrez Gene ID
55791
Gene Name
ligand dependent nuclear receptor interacting factor 1
Gene Aliases
C1orf103, RIF1
UniGene
Hs.25245
Gene Chromosome Location
Chr.1: 110877252-110964203 on Build GRCh38
Transcripts
NM_001006945.1 NM_018372.3 XM_005271029.3 XM_011541768.1 XM_017001768.1 XM_017001769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7110401 c.2012C>A p.P671Q Chr.1: 110948257-110948257 on Build GRCh38
7111899 c.2043C>T p.L681L Chr.1: 110948226-110948226 on Build GRCh38
6577220 c.2037delC p.N679fs*30 Chr.1: 110948232-110948232 on Build GRCh38
1283763 c.2080G>T p.E694* Chr.1: 110948189-110948189 on Build GRCh38
1332639 c.2275C>G p.L759V Chr.1: 110947994-110947994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368369866 Chr.1: 110948365-110948365 on Build GRCh38
rs900074377 Chr.1: 110948120-110948120 on Build GRCh38
rs757829085 Chr.1: 110948358-110948358 on Build GRCh38
rs376501948 Chr.1: 110947982-110947982 on Build GRCh38
rs764241552 Chr.1: 110948341-110948344 on Build GRCh38
rs772757214 Chr.1: 110948078-110948078 on Build GRCh38
rs955412724 Chr.1: 110948236-110948236 on Build GRCh38
rs759763152 Chr.1: 110948094-110948094 on Build GRCh38
rs761774344 Chr.1: 110948285-110948285 on Build GRCh38
rs747482932 Chr.1: 110948172-110948172 on Build GRCh38
rs1045525081 Chr.1: 110948107-110948107 on Build GRCh38
rs778203277 Chr.1: 110948170-110948170 on Build GRCh38
rs766550641 Chr.1: 110947998-110947998 on Build GRCh38
rs568800560 Chr.1: 110948315-110948315 on Build GRCh38
rs199995205 Chr.1: 110948134-110948134 on Build GRCh38
rs371082233 Chr.1: 110948317-110948317 on Build GRCh38
rs768378091 Chr.1: 110948263-110948263 on Build GRCh38
rs990580681 Chr.1: 110948240-110948240 on Build GRCh38
rs146605389 Chr.1: 110948048-110948048 on Build GRCh38
rs780862697 Chr.1: 110947960-110947960 on Build GRCh38
rs143295716 Chr.1: 110948068-110948068 on Build GRCh38
rs750001410 Chr.1: 110948299-110948299 on Build GRCh38
rs750077474 Chr.1: 110948145-110948145 on Build GRCh38
rs779832911 Chr.1: 110948152-110948152 on Build GRCh38
rs949169665 Chr.1: 110947978-110947978 on Build GRCh38
rs754090645 Chr.1: 110948357-110948359 on Build GRCh38
rs200670185 Chr.1: 110948117-110948117 on Build GRCh38
rs370405768 Chr.1: 110948067-110948067 on Build GRCh38
rs775682328 Chr.1: 110948255-110948255 on Build GRCh38
rs764081477 Chr.1: 110948096-110948096 on Build GRCh38
rs770946803 Chr.1: 110948176-110948176 on Build GRCh38
rs757482653 Chr.1: 110948375-110948379 on Build GRCh38
rs761494634 Chr.1: 110948418-110948418 on Build GRCh38
rs748690553 Chr.1: 110948073-110948073 on Build GRCh38
rs745508016 Chr.1: 110947988-110947988 on Build GRCh38
rs771030930 Chr.1: 110948084-110948084 on Build GRCh38
rs200763276 Chr.1: 110948363-110948363 on Build GRCh38
rs374381226 Chr.1: 110948293-110948293 on Build GRCh38
rs769113814 Chr.1: 110948403-110948403 on Build GRCh38
rs946116993 Chr.1: 110948064-110948064 on Build GRCh38
rs752661913 Chr.1: 110948023-110948023 on Build GRCh38
rs980598758 Chr.1: 110948221-110948221 on Build GRCh38
rs78742177 Chr.1: 110948297-110948297 on Build GRCh38
rs375688507 Chr.1: 110948286-110948286 on Build GRCh38
rs758847416 Chr.1: 110948327-110948327 on Build GRCh38
rs745981621 Chr.1: 110948383-110948385 on Build GRCh38
rs757755996 Chr.1: 110948291-110948291 on Build GRCh38
rs749678246 Chr.1: 110947967-110947967 on Build GRCh38
rs762790969 Chr.1: 110948258-110948258 on Build GRCh38
rs148620213 Chr.1: 110948148-110948148 on Build GRCh38
rs140706787 Chr.1: 110947970-110947970 on Build GRCh38
rs374452637 Chr.1: 110948348-110948348 on Build GRCh38
rs145932328 Chr.1: 110948361-110948361 on Build GRCh38
rs764944479 Chr.1: 110948011-110948011 on Build GRCh38
rs763995417 Chr.1: 110948047-110948047 on Build GRCh38
rs747126394 Chr.1: 110948082-110948082 on Build GRCh38
rs767481981 Chr.1: 110948000-110948002 on Build GRCh38
rs1045389510 Chr.1: 110948414-110948414 on Build GRCh38
rs774150050 Chr.1: 110948416-110948416 on Build GRCh38
rs780264484 Chr.1: 110948367-110948367 on Build GRCh38
rs779033999 Chr.1: 110948378-110948380 on Build GRCh38
rs148419989 Chr.1: 110948320-110948320 on Build GRCh38
rs995618979 Chr.1: 110948135-110948135 on Build GRCh38
rs147565982 Chr.1: 110948050-110948050 on Build GRCh38
rs371134004 Chr.1: 110948312-110948312 on Build GRCh38
rs748565882 Chr.1: 110948157-110948157 on Build GRCh38
rs199904906 Chr.1: 110948124-110948124 on Build GRCh38
rs752447521 Chr.1: 110948021-110948021 on Build GRCh38
rs758831233 Chr.1: 110948161-110948161 on Build GRCh38
rs775014991 Chr.1: 110947980-110947980 on Build GRCh38
rs974248922 Chr.1: 110948316-110948316 on Build GRCh38
rs758281136 Chr.1: 110948029-110948029 on Build GRCh38
rs776908021 Chr.1: 110948005-110948005 on Build GRCh38
rs773825411 Chr.1: 110948080-110948080 on Build GRCh38
rs936599617 Chr.1: 110948231-110948231 on Build GRCh38
rs756735360 Chr.1: 110948121-110948121 on Build GRCh38
rs142104111 Chr.1: 110948215-110948215 on Build GRCh38
rs542067453 Chr.1: 110948093-110948093 on Build GRCh38
rs768371364 Chr.1: 110948395-110948395 on Build GRCh38
rs773379392 Chr.1: 110947987-110947987 on Build GRCh38
rs190274947 Chr.1: 110947981-110947981 on Build GRCh38
rs753528581 Chr.1: 110948318-110948318 on Build GRCh38
rs915562796 Chr.1: 110948070-110948070 on Build GRCh38
rs996755422 Chr.1: 110948223-110948223 on Build GRCh38
rs769846738 Chr.1: 110948364-110948364 on Build GRCh38
rs755924956 Chr.1: 110948083-110948083 on Build GRCh38
rs775310244 Chr.1: 110948095-110948095 on Build GRCh38
rs940212158 Chr.1: 110948016-110948016 on Build GRCh38
rs2232047 Chr.1: 110948346-110948346 on Build GRCh38
rs752029389 Chr.1: 110948357-110948357 on Build GRCh38
rs749448676 Chr.1: 110948389-110948389 on Build GRCh38
rs372607968 Chr.1: 110948224-110948224 on Build GRCh38
rs147205230 Chr.1: 110948228-110948228 on Build GRCh38
rs151182141 Chr.1: 110948362-110948362 on Build GRCh38
rs149285020 Chr.1: 110948268-110948268 on Build GRCh38
rs760788005 Chr.1: 110947995-110947995 on Build GRCh38
rs774163210 Chr.1: 110948274-110948274 on Build GRCh38

View Full Product Details