Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111722643-111723189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM212B
Entrez Gene ID
55924
Gene Name
family with sequence similarity 212 member B
Gene Aliases
C1orf183
UniGene
Hs.193406
Gene Chromosome Location
Chr.1: 111722064-111755797 on Build GRCh38
Transcripts
NM_019099.4 NM_198926.2 XM_011541783.2

SNP Detail

SNP ID Chromosome Location
rs138098294 Chr.1: 111722914-111722914 on Build GRCh38
rs944767634 Chr.1: 111722775-111722775 on Build GRCh38
rs189992724 Chr.1: 111723113-111723113 on Build GRCh38
rs935954769 Chr.1: 111723053-111723053 on Build GRCh38
rs770602469 Chr.1: 111723135-111723135 on Build GRCh38
rs76860170 Chr.1: 111722710-111722710 on Build GRCh38
rs974046054 Chr.1: 111722843-111722843 on Build GRCh38
rs548667235 Chr.1: 111723074-111723074 on Build GRCh38
rs925764485 Chr.1: 111723040-111723040 on Build GRCh38
rs1005224874 Chr.1: 111722937-111722937 on Build GRCh38
rs148046710 Chr.1: 111722674-111722676 on Build GRCh38
rs197424 Chr.1: 111723120-111723120 on Build GRCh38
rs992757401 Chr.1: 111722946-111722946 on Build GRCh38
rs986300386 Chr.1: 111722782-111722782 on Build GRCh38
rs117364116 Chr.1: 111722688-111722688 on Build GRCh38
rs8179264 Chr.1: 111722910-111722910 on Build GRCh38
rs1044268858 Chr.1: 111723031-111723031 on Build GRCh38
rs752540986 Chr.1: 111723083-111723083 on Build GRCh38
rs780733841 Chr.1: 111723164-111723164 on Build GRCh38
rs749426814 Chr.1: 111723133-111723133 on Build GRCh38
rs867327834 Chr.1: 111722925-111722925 on Build GRCh38
rs953741392 Chr.1: 111722760-111722760 on Build GRCh38
rs946753889 Chr.1: 111722720-111722720 on Build GRCh38
rs147385845 Chr.1: 111722801-111722801 on Build GRCh38
rs997858733 Chr.1: 111723119-111723119 on Build GRCh38
rs917335712 Chr.1: 111722961-111722961 on Build GRCh38
rs563246287 Chr.1: 111722924-111722924 on Build GRCh38
rs1035175745 Chr.1: 111723112-111723112 on Build GRCh38
rs528998223 Chr.1: 111723070-111723070 on Build GRCh38
rs994107671 Chr.1: 111722829-111722829 on Build GRCh38
rs910644394 Chr.1: 111722751-111722751 on Build GRCh38
rs186155763 Chr.1: 111722884-111722884 on Build GRCh38
rs948805319 Chr.1: 111722999-111722999 on Build GRCh38
rs1050825661 Chr.1: 111722746-111722746 on Build GRCh38
rs181996028 Chr.1: 111722828-111722828 on Build GRCh38
rs761998009 Chr.1: 111723151-111723160 on Build GRCh38

View Full Product Details