Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 143001451-143001946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100289361
Entrez Gene ID
100289361
Gene Name
uncharacterized LOC100289361
Gene Aliases
-
UniGene
Hs.599649
Gene Chromosome Location
Chr.3: 143000845-143001467 on Build GRCh38
Transcripts
NR_034032.1 NM_001080415.1 NM_001320219.1 NM_001320220.1 NM_001320222.1 XM_005247248.4 XM_017006038.1

Gene Symbol
U2SURP
Entrez Gene ID
23350
Gene Name
U2 snRNP associated SURP domain containing
Gene Aliases
SR140, fSAPa
UniGene
Hs.596572
Gene Chromosome Location
Chr.3: 143001524-143060725 on Build GRCh38
Transcripts
NR_034032.1 NM_001080415.1 NM_001320219.1 NM_001320220.1 NM_001320222.1 XM_005247248.4 XM_017006038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6054845 c.22G>T p.G8* Chr.3: 143001650-143001650 on Build GRCh38
5984626 c.7G>A p.D3N Chr.3: 143001635-143001635 on Build GRCh38
5687933 c.9C>T p.D3D Chr.3: 143001637-143001637 on Build GRCh38
3588328 c.27T>C p.S9S Chr.3: 143001655-143001655 on Build GRCh38
6814758 c.13A>T p.T5S Chr.3: 143001641-143001641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755269749 Chr.3: 143001651-143001651 on Build GRCh38
rs762834528 Chr.3: 143001645-143001645 on Build GRCh38
rs778388132 Chr.3: 143001620-143001620 on Build GRCh38
rs568106049 Chr.3: 143001866-143001866 on Build GRCh38
rs755038890 Chr.3: 143001616-143001616 on Build GRCh38
rs548359963 Chr.3: 143001827-143001827 on Build GRCh38
rs115761525 Chr.3: 143001884-143001884 on Build GRCh38
rs1000420268 Chr.3: 143001880-143001880 on Build GRCh38
rs890086633 Chr.3: 143001855-143001855 on Build GRCh38
rs749860940 Chr.3: 143001656-143001656 on Build GRCh38
rs767616520 Chr.3: 143001712-143001712 on Build GRCh38
rs182428347 Chr.3: 143001916-143001916 on Build GRCh38
rs970692316 Chr.3: 143001912-143001912 on Build GRCh38
rs113402301 Chr.3: 143001847-143001847 on Build GRCh38
rs776394862 Chr.3: 143001694-143001694 on Build GRCh38
rs770237747 Chr.3: 143001619-143001619 on Build GRCh38
rs552224030 Chr.3: 143001874-143001874 on Build GRCh38
rs758588508 Chr.3: 143001644-143001644 on Build GRCh38
rs768162618 Chr.3: 143001693-143001693 on Build GRCh38
rs897555607 Chr.3: 143001839-143001839 on Build GRCh38
rs777950336 Chr.3: 143001647-143001647 on Build GRCh38
rs752941456 Chr.3: 143001608-143001608 on Build GRCh38
rs926927674 Chr.3: 143001799-143001799 on Build GRCh38
rs749632638 Chr.3: 143001623-143001623 on Build GRCh38
rs760165421 Chr.3: 143001592-143001592 on Build GRCh38
rs890096431 Chr.3: 143001846-143001846 on Build GRCh38
rs375855390 Chr.3: 143001586-143001586 on Build GRCh38
rs905114055 Chr.3: 143001727-143001730 on Build GRCh38
rs538513118 Chr.3: 143001554-143001554 on Build GRCh38
rs775174964 Chr.3: 143001589-143001589 on Build GRCh38
rs1018903545 Chr.3: 143001857-143001857 on Build GRCh38
rs768156136 Chr.3: 143001634-143001634 on Build GRCh38
rs1000726245 Chr.3: 143001514-143001514 on Build GRCh38
rs955589560 Chr.3: 143001614-143001614 on Build GRCh38
rs904479878 Chr.3: 143001870-143001870 on Build GRCh38
rs763579775 Chr.3: 143001597-143001597 on Build GRCh38
rs761426654 Chr.3: 143001638-143001638 on Build GRCh38
rs758314591 Chr.3: 143001602-143001602 on Build GRCh38
rs190954305 Chr.3: 143001791-143001791 on Build GRCh38
rs191623793 Chr.3: 143001563-143001563 on Build GRCh38
rs527443908 Chr.3: 143001496-143001496 on Build GRCh38
rs372971680 Chr.3: 143001618-143001618 on Build GRCh38
rs375686570 Chr.3: 143001710-143001710 on Build GRCh38
rs780408117 Chr.3: 143001603-143001603 on Build GRCh38
rs759153323 Chr.3: 143001576-143001576 on Build GRCh38
rs766223687 Chr.3: 143001705-143001705 on Build GRCh38
rs558572041 Chr.3: 143001490-143001490 on Build GRCh38
rs3902877 Chr.3: 143001888-143001888 on Build GRCh38
rs942189239 Chr.3: 143001812-143001812 on Build GRCh38
rs761670533 Chr.3: 143001600-143001600 on Build GRCh38
rs201100859 Chr.3: 143001704-143001704 on Build GRCh38
rs756338171 Chr.3: 143001614-143001614 on Build GRCh38
rs140095013 Chr.3: 143001716-143001716 on Build GRCh38
rs746616952 Chr.3: 143001670-143001670 on Build GRCh38
rs375415414 Chr.3: 143001697-143001697 on Build GRCh38
rs1032469775 Chr.3: 143001537-143001537 on Build GRCh38
rs751894109 Chr.3: 143001612-143001612 on Build GRCh38
rs112914172 Chr.3: 143001700-143001700 on Build GRCh38
rs776034960 Chr.3: 143001637-143001637 on Build GRCh38
rs866891728 Chr.3: 143001913-143001913 on Build GRCh38
rs370308369 Chr.3: 143001646-143001646 on Build GRCh38
rs766360 Chr.3: 143001506-143001506 on Build GRCh38
rs533864079 Chr.3: 143001856-143001856 on Build GRCh38
rs771346677 Chr.3: 143001625-143001625 on Build GRCh38
rs183628705 Chr.3: 143001666-143001666 on Build GRCh38
rs560427922 Chr.3: 143001654-143001654 on Build GRCh38
rs188965230 Chr.3: 143001789-143001789 on Build GRCh38
rs767597694 Chr.3: 143001652-143001652 on Build GRCh38
rs533594901 Chr.3: 143001473-143001473 on Build GRCh38
rs376927803 Chr.3: 143001628-143001628 on Build GRCh38
rs573249900 Chr.3: 143001598-143001598 on Build GRCh38
rs111340020 Chr.3: 143001601-143001601 on Build GRCh38
rs532293687 Chr.3: 143001745-143001745 on Build GRCh38
rs765060815 Chr.3: 143001642-143001642 on Build GRCh38
rs779264097 Chr.3: 143001669-143001669 on Build GRCh38
rs964333520 Chr.3: 143001865-143001865 on Build GRCh38
rs1051527764 Chr.3: 143001853-143001853 on Build GRCh38
rs550425430 Chr.3: 143001785-143001785 on Build GRCh38
rs556847016 Chr.3: 143001894-143001894 on Build GRCh38
rs1025020314 Chr.3: 143001901-143001901 on Build GRCh38
rs774315767 Chr.3: 143001708-143001708 on Build GRCh38

View Full Product Details