Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150558258-150558758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2A
Entrez Gene ID
83939
Gene Name
eukaryotic translation initiation factor 2A
Gene Aliases
CDA02, EIF-2A, MST089, MSTP004, MSTP089
UniGene
Hs.655782
Gene Chromosome Location
Chr.3: 150546678-150586016 on Build GRCh38
Transcripts
NM_001319043.1 NM_001319044.1 NM_001319045.1 NM_001319046.1 NM_032025.4 XM_011513224.1 XM_011512698.2

Gene Symbol
SERP1
Entrez Gene ID
27230
Gene Name
stress-associated endoplasmic reticulum protein 1
Gene Aliases
RAMP4
UniGene
Hs.518326
Gene Chromosome Location
Chr.3: 150541993-150603177 on Build GRCh38
Transcripts
NM_001319043.1 NM_001319044.1 NM_001319045.1 NM_001319046.1 NM_032025.4 XM_011513224.1 XM_011512698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7366020 c.99G>T p.R33S Chr.3: 150558388-150558388 on Build GRCh38
6397626 c.148T>A p.L50M Chr.3: 150558437-150558437 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747233725 Chr.3: 150558463-150558463 on Build GRCh38
rs757290547 Chr.3: 150558439-150558439 on Build GRCh38
rs778949826 Chr.3: 150558445-150558445 on Build GRCh38
rs1023162478 Chr.3: 150558312-150558312 on Build GRCh38
rs540589744 Chr.3: 150558560-150558560 on Build GRCh38
rs200598431 Chr.3: 150558368-150558368 on Build GRCh38
rs6767495 Chr.3: 150558679-150558679 on Build GRCh38
rs575441945 Chr.3: 150558366-150558366 on Build GRCh38
rs953598722 Chr.3: 150558388-150558388 on Build GRCh38
rs566767572 Chr.3: 150558360-150558360 on Build GRCh38
rs35520275 Chr.3: 150558366-150558367 on Build GRCh38
rs771958898 Chr.3: 150558457-150558457 on Build GRCh38
rs376472201 Chr.3: 150558465-150558465 on Build GRCh38
rs755391863 Chr.3: 150558478-150558478 on Build GRCh38
rs777439208 Chr.3: 150558430-150558430 on Build GRCh38
rs180766420 Chr.3: 150558308-150558308 on Build GRCh38
rs776709910 Chr.3: 150558479-150558479 on Build GRCh38
rs372635613 Chr.3: 150558461-150558461 on Build GRCh38
rs769743883 Chr.3: 150558484-150558484 on Build GRCh38
rs897885474 Chr.3: 150558603-150558603 on Build GRCh38
rs200333608 Chr.3: 150558364-150558365 on Build GRCh38
rs763367995 Chr.3: 150558492-150558492 on Build GRCh38
rs754050662 Chr.3: 150558380-150558381 on Build GRCh38
rs947953321 Chr.3: 150558547-150558547 on Build GRCh38
rs910888211 Chr.3: 150558473-150558473 on Build GRCh38
rs956071321 Chr.3: 150558407-150558408 on Build GRCh38
rs1012133219 Chr.3: 150558302-150558302 on Build GRCh38
rs1023595507 Chr.3: 150558598-150558598 on Build GRCh38
rs201778476 Chr.3: 150558358-150558358 on Build GRCh38
rs79583943 Chr.3: 150558487-150558487 on Build GRCh38
rs1040031175 Chr.3: 150558484-150558485 on Build GRCh38
rs887557566 Chr.3: 150558543-150558543 on Build GRCh38
rs968812975 Chr.3: 150558698-150558698 on Build GRCh38
rs76475914 Chr.3: 150558376-150558376 on Build GRCh38
rs760608703 Chr.3: 150558681-150558681 on Build GRCh38
rs191019624 Chr.3: 150558500-150558500 on Build GRCh38
rs753997558 Chr.3: 150558607-150558607 on Build GRCh38
rs77047707 Chr.3: 150558377-150558377 on Build GRCh38
rs569423093 Chr.3: 150558476-150558476 on Build GRCh38
rs1004031009 Chr.3: 150558347-150558347 on Build GRCh38
rs946502160 Chr.3: 150558415-150558415 on Build GRCh38
rs1003511909 Chr.3: 150558590-150558590 on Build GRCh38
rs554597609 Chr.3: 150558643-150558643 on Build GRCh38
rs906517686 Chr.3: 150558567-150558567 on Build GRCh38
rs979287168 Chr.3: 150558429-150558429 on Build GRCh38
rs6776252 Chr.3: 150558604-150558604 on Build GRCh38
rs11924919 Chr.3: 150558365-150558365 on Build GRCh38
rs1047442155 Chr.3: 150558535-150558535 on Build GRCh38
rs764446204 Chr.3: 150558367-150558368 on Build GRCh38
rs988822918 Chr.3: 150558626-150558626 on Build GRCh38
rs756826748 Chr.3: 150558287-150558287 on Build GRCh38
rs753350813 Chr.3: 150558431-150558431 on Build GRCh38
rs11379788 Chr.3: 150558376-150558377 on Build GRCh38
rs755762502 Chr.3: 150558414-150558414 on Build GRCh38
rs906556567 Chr.3: 150558338-150558338 on Build GRCh38
rs201097440 Chr.3: 150558345-150558346 on Build GRCh38
rs928993520 Chr.3: 150558519-150558519 on Build GRCh38
rs752085271 Chr.3: 150558373-150558373 on Build GRCh38
rs979256141 Chr.3: 150558700-150558700 on Build GRCh38
rs200092124 Chr.3: 150558483-150558483 on Build GRCh38
rs953567017 Chr.3: 150558609-150558609 on Build GRCh38
rs554411471 Chr.3: 150558599-150558599 on Build GRCh38
rs970742130 Chr.3: 150558339-150558339 on Build GRCh38
rs776207394 Chr.3: 150558499-150558499 on Build GRCh38
rs796139294 Chr.3: 150558365-150558366 on Build GRCh38
rs966700310 Chr.3: 150558727-150558727 on Build GRCh38
rs1019175083 Chr.3: 150558722-150558722 on Build GRCh38
rs762945396 Chr.3: 150558367-150558367 on Build GRCh38
rs745769713 Chr.3: 150558453-150558453 on Build GRCh38

View Full Product Details