Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150568067-150568604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2A
Entrez Gene ID
83939
Gene Name
eukaryotic translation initiation factor 2A
Gene Aliases
CDA02, EIF-2A, MST089, MSTP004, MSTP089
UniGene
Hs.655782
Gene Chromosome Location
Chr.3: 150546678-150586016 on Build GRCh38
Transcripts
NM_001319043.1 NM_001319044.1 NM_001319045.1 NM_001319046.1 NM_032025.4 XM_011513224.1 XM_011512698.2

Gene Symbol
SERP1
Entrez Gene ID
27230
Gene Name
stress-associated endoplasmic reticulum protein 1
Gene Aliases
RAMP4
UniGene
Hs.518326
Gene Chromosome Location
Chr.3: 150541993-150603177 on Build GRCh38
Transcripts
NM_001319043.1 NM_001319044.1 NM_001319045.1 NM_001319046.1 NM_032025.4 XM_011513224.1 XM_011512698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5474561 c.811+1G>A p.? Chr.3: 150568293-150568293 on Build GRCh38
3767095 c.811C>T p.P271S Chr.3: 150568292-150568292 on Build GRCh38
5474562 c.736+1G>A p.? Chr.3: 150568293-150568293 on Build GRCh38
3767096 c.736C>T p.P246S Chr.3: 150568292-150568292 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1001711131 Chr.3: 150568363-150568363 on Build GRCh38
rs368575162 Chr.3: 150568157-150568157 on Build GRCh38
rs531008925 Chr.3: 150568155-150568155 on Build GRCh38
rs891862280 Chr.3: 150568186-150568186 on Build GRCh38
rs751289885 Chr.3: 150568224-150568224 on Build GRCh38
rs940564964 Chr.3: 150568151-150568151 on Build GRCh38
rs1001231828 Chr.3: 150568188-150568188 on Build GRCh38
rs963995060 Chr.3: 150568430-150568433 on Build GRCh38
rs139445557 Chr.3: 150568166-150568166 on Build GRCh38
rs775490153 Chr.3: 150568173-150568173 on Build GRCh38
rs752217036 Chr.3: 150568257-150568257 on Build GRCh38
rs983642405 Chr.3: 150568527-150568527 on Build GRCh38
rs775575993 Chr.3: 150568292-150568292 on Build GRCh38
rs576406136 Chr.3: 150568520-150568520 on Build GRCh38
rs771937315 Chr.3: 150568328-150568330 on Build GRCh38
rs780875918 Chr.3: 150568234-150568234 on Build GRCh38
rs765501284 Chr.3: 150568215-150568215 on Build GRCh38
rs374369665 Chr.3: 150568130-150568130 on Build GRCh38
rs967588734 Chr.3: 150568417-150568417 on Build GRCh38
rs115348077 Chr.3: 150568100-150568100 on Build GRCh38
rs1014011174 Chr.3: 150568346-150568346 on Build GRCh38
rs540353682 Chr.3: 150568251-150568251 on Build GRCh38
rs757143516 Chr.3: 150568284-150568284 on Build GRCh38
rs62283993 Chr.3: 150568301-150568301 on Build GRCh38
rs940549347 Chr.3: 150568556-150568556 on Build GRCh38
rs754532571 Chr.3: 150568230-150568230 on Build GRCh38
rs779449615 Chr.3: 150568503-150568503 on Build GRCh38
rs761191099 Chr.3: 150568193-150568193 on Build GRCh38
rs991809388 Chr.3: 150568502-150568502 on Build GRCh38
rs562920233 Chr.3: 150568563-150568563 on Build GRCh38
rs757590156 Chr.3: 150568449-150568449 on Build GRCh38
rs768697359 Chr.3: 150568307-150568307 on Build GRCh38
rs1915945 Chr.3: 150568423-150568423 on Build GRCh38
rs201002165 Chr.3: 150568197-150568197 on Build GRCh38
rs975424011 Chr.3: 150568446-150568446 on Build GRCh38
rs372131915 Chr.3: 150568577-150568577 on Build GRCh38
rs1020830577 Chr.3: 150568348-150568348 on Build GRCh38
rs929073991 Chr.3: 150568101-150568101 on Build GRCh38
rs780077662 Chr.3: 150568348-150568348 on Build GRCh38
rs746115513 Chr.3: 150568291-150568291 on Build GRCh38
rs1047540989 Chr.3: 150568115-150568115 on Build GRCh38
rs997449177 Chr.3: 150568167-150568167 on Build GRCh38
rs755657985 Chr.3: 150568258-150568258 on Build GRCh38
rs922680304 Chr.3: 150568500-150568501 on Build GRCh38
rs960289468 Chr.3: 150568481-150568481 on Build GRCh38
rs898806861 Chr.3: 150568178-150568178 on Build GRCh38
rs754973227 Chr.3: 150568564-150568567 on Build GRCh38
rs754308696 Chr.3: 150568198-150568198 on Build GRCh38
rs1033253283 Chr.3: 150568429-150568432 on Build GRCh38
rs543329156 Chr.3: 150568496-150568496 on Build GRCh38
rs778506109 Chr.3: 150568288-150568288 on Build GRCh38
rs900430084 Chr.3: 150568165-150568165 on Build GRCh38
rs762284788 Chr.3: 150568205-150568205 on Build GRCh38

View Full Product Details