Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 158644409-158644906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFM1
Entrez Gene ID
85476
Gene Name
G elongation factor mitochondrial 1
Gene Aliases
COXPD1, EFG, EFG1, EFGM, EGF1, GFM, hEFG1
UniGene
Hs.518355
Gene Chromosome Location
Chr.3: 158644497-158692572 on Build GRCh38
Transcripts
NM_001308164.1 NM_001308166.1 NM_024996.5 XM_006713795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1040507 c.20C>T p.A7V Chr.3: 158644654-158644654 on Build GRCh38
3721924 c.18T>C p.A6A Chr.3: 158644652-158644652 on Build GRCh38
3589375 c.37G>A p.G13R Chr.3: 158644671-158644671 on Build GRCh38
5594626 c.35delT p.L12fs*10 Chr.3: 158644669-158644669 on Build GRCh38
5794792 c.32C>A p.A11D Chr.3: 158644666-158644666 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780010700 Chr.3: 158644744-158644744 on Build GRCh38
rs893145449 Chr.3: 158644550-158644550 on Build GRCh38
rs780207360 Chr.3: 158644661-158644661 on Build GRCh38
rs564879457 Chr.3: 158644569-158644569 on Build GRCh38
rs777558130 Chr.3: 158644696-158644696 on Build GRCh38
rs930167520 Chr.3: 158644469-158644469 on Build GRCh38
rs928858622 Chr.3: 158644868-158644868 on Build GRCh38
rs909108179 Chr.3: 158644432-158644432 on Build GRCh38
rs1018008994 Chr.3: 158644797-158644797 on Build GRCh38
rs531369602 Chr.3: 158644608-158644608 on Build GRCh38
rs776007974 Chr.3: 158644688-158644688 on Build GRCh38
rs963320998 Chr.3: 158644586-158644586 on Build GRCh38
rs560227828 Chr.3: 158644575-158644575 on Build GRCh38
rs767597426 Chr.3: 158644721-158644721 on Build GRCh38
rs928828435 Chr.3: 158644683-158644683 on Build GRCh38
rs756324887 Chr.3: 158644647-158644647 on Build GRCh38
rs749029419 Chr.3: 158644591-158644591 on Build GRCh38
rs553797646 Chr.3: 158644476-158644477 on Build GRCh38
rs574292828 Chr.3: 158644736-158644736 on Build GRCh38
rs1026668201 Chr.3: 158644827-158644827 on Build GRCh38
rs1055907573 Chr.3: 158644701-158644701 on Build GRCh38
rs1029494701 Chr.3: 158644584-158644584 on Build GRCh38
rs753846091 Chr.3: 158644837-158644837 on Build GRCh38
rs377352238 Chr.3: 158644597-158644597 on Build GRCh38
rs1864507 Chr.3: 158644652-158644652 on Build GRCh38
rs933045446 Chr.3: 158644450-158644450 on Build GRCh38
rs755113549 Chr.3: 158644643-158644643 on Build GRCh38
rs28372853 Chr.3: 158644604-158644604 on Build GRCh38
rs1034401781 Chr.3: 158644535-158644535 on Build GRCh38
rs981720475 Chr.3: 158644853-158644853 on Build GRCh38
rs754995745 Chr.3: 158644763-158644763 on Build GRCh38
rs863224031 Chr.3: 158644650-158644650 on Build GRCh38
rs28372852 Chr.3: 158644602-158644602 on Build GRCh38
rs535060182 Chr.3: 158644458-158644458 on Build GRCh38
rs937454854 Chr.3: 158644685-158644685 on Build GRCh38
rs373016137 Chr.3: 158644596-158644596 on Build GRCh38
rs951085474 Chr.3: 158644847-158644847 on Build GRCh38
rs780703183 Chr.3: 158644765-158644765 on Build GRCh38
rs555757931 Chr.3: 158644714-158644714 on Build GRCh38
rs111428707 Chr.3: 158644446-158644446 on Build GRCh38
rs978352242 Chr.3: 158644572-158644572 on Build GRCh38
rs760933181 Chr.3: 158644729-158644729 on Build GRCh38
rs751477274 Chr.3: 158644746-158644746 on Build GRCh38
rs555913327 Chr.3: 158644476-158644476 on Build GRCh38
rs973699666 Chr.3: 158644607-158644607 on Build GRCh38
rs150750805 Chr.3: 158644874-158644874 on Build GRCh38
rs745780647 Chr.3: 158644598-158644598 on Build GRCh38
rs758326841 Chr.3: 158644742-158644742 on Build GRCh38
rs766920149 Chr.3: 158644635-158644635 on Build GRCh38
rs757131337 Chr.3: 158644595-158644595 on Build GRCh38
rs772161827 Chr.3: 158644601-158644601 on Build GRCh38
rs191210604 Chr.3: 158644444-158644444 on Build GRCh38
rs567474426 Chr.3: 158644605-158644605 on Build GRCh38
rs868390553 Chr.3: 158644686-158644686 on Build GRCh38
rs377591693 Chr.3: 158644618-158644618 on Build GRCh38
rs758378925 Chr.3: 158644659-158644659 on Build GRCh38
rs765266988 Chr.3: 158644684-158644684 on Build GRCh38
rs752744978 Chr.3: 158644727-158644727 on Build GRCh38
rs540510822 Chr.3: 158644457-158644457 on Build GRCh38
rs751603045 Chr.3: 158644636-158644636 on Build GRCh38
rs567086019 Chr.3: 158644690-158644690 on Build GRCh38
rs767710902 Chr.3: 158644645-158644645 on Build GRCh38
rs1041700008 Chr.3: 158644720-158644720 on Build GRCh38
rs141179055 Chr.3: 158644779-158644779 on Build GRCh38
rs1016137610 Chr.3: 158644581-158644581 on Build GRCh38
rs574200635 Chr.3: 158644711-158644711 on Build GRCh38
rs773603488 Chr.3: 158644700-158644700 on Build GRCh38
rs539498045 Chr.3: 158644662-158644662 on Build GRCh38
rs543432802 Chr.3: 158644619-158644619 on Build GRCh38
rs558123018 Chr.3: 158644677-158644677 on Build GRCh38
rs1864505 Chr.3: 158644799-158644799 on Build GRCh38
rs562527408 Chr.3: 158644528-158644528 on Build GRCh38
rs766862933 Chr.3: 158644706-158644706 on Build GRCh38
rs1001559608 Chr.3: 158644546-158644546 on Build GRCh38
rs955235348 Chr.3: 158644772-158644772 on Build GRCh38
rs918252573 Chr.3: 158644813-158644813 on Build GRCh38
rs752904724 Chr.3: 158644646-158644646 on Build GRCh38
rs1025079892 Chr.3: 158644563-158644563 on Build GRCh38
rs112860155 Chr.3: 158644624-158644624 on Build GRCh38
rs1051957121 Chr.3: 158644463-158644463 on Build GRCh38
rs1011561551 Chr.3: 158644560-158644560 on Build GRCh38
rs868019124 Chr.3: 158644641-158644641 on Build GRCh38
rs763344414 Chr.3: 158644703-158644703 on Build GRCh38
rs745704169 Chr.3: 158644656-158644656 on Build GRCh38
rs763401118 Chr.3: 158644627-158644627 on Build GRCh38
rs185840111 Chr.3: 158644798-158644798 on Build GRCh38
rs557827192 Chr.3: 158644494-158644494 on Build GRCh38
rs116614958 Chr.3: 158644562-158644562 on Build GRCh38
rs1007030187 Chr.3: 158644519-158644519 on Build GRCh38
rs754681402 Chr.3: 158644841-158644841 on Build GRCh38
rs544347675 Chr.3: 158644511-158644511 on Build GRCh38
rs764267543 Chr.3: 158644734-158644734 on Build GRCh38
rs370405683 Chr.3: 158644547-158644547 on Build GRCh38
rs1001619445 Chr.3: 158644520-158644520 on Build GRCh38
rs545545302 Chr.3: 158644436-158644436 on Build GRCh38
rs907321342 Chr.3: 158644496-158644496 on Build GRCh38
rs560069814 Chr.3: 158644832-158644832 on Build GRCh38
rs1864506 Chr.3: 158644737-158644737 on Build GRCh38
rs573810305 Chr.3: 158644502-158644502 on Build GRCh38
rs571915979 Chr.3: 158644866-158644866 on Build GRCh38
rs863224030 Chr.3: 158644637-158644637 on Build GRCh38
rs886058119 Chr.3: 158644720-158644720 on Build GRCh38
rs778513623 Chr.3: 158644788-158644788 on Build GRCh38
rs557817234 Chr.3: 158644536-158644536 on Build GRCh38
rs746558595 Chr.3: 158644603-158644603 on Build GRCh38
rs904100673 Chr.3: 158644544-158644544 on Build GRCh38

View Full Product Details