Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 158653200-158653746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFM1
Entrez Gene ID
85476
Gene Name
G elongation factor mitochondrial 1
Gene Aliases
COXPD1, EFG, EFG1, EFGM, EGF1, GFM, hEFG1
UniGene
Hs.518355
Gene Chromosome Location
Chr.3: 158644497-158692572 on Build GRCh38
Transcripts
NM_001308164.1 NM_001308166.1 NM_024996.5 XM_006713795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5457035 c.980C>T p.A327V Chr.3: 158653449-158653449 on Build GRCh38
6670511 c.886delT p.L297fs*5 Chr.3: 158653355-158653355 on Build GRCh38
1208123 c.53G>A p.R18Q Chr.3: 158653320-158653320 on Build GRCh38
1040518 c.930A>C p.L310F Chr.3: 158653399-158653399 on Build GRCh38
5457036 c.182C>T p.A61V Chr.3: 158653449-158653449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376636675 Chr.3: 158653393-158653393 on Build GRCh38
rs763084523 Chr.3: 158653466-158653469 on Build GRCh38
rs200902485 Chr.3: 158653502-158653502 on Build GRCh38
rs202026103 Chr.3: 158653565-158653566 on Build GRCh38
rs973393706 Chr.3: 158653523-158653523 on Build GRCh38
rs775228051 Chr.3: 158653355-158653355 on Build GRCh38
rs769903119 Chr.3: 158653436-158653436 on Build GRCh38
rs563970256 Chr.3: 158653691-158653691 on Build GRCh38
rs9858849 Chr.3: 158653617-158653617 on Build GRCh38
rs781397040 Chr.3: 158653427-158653427 on Build GRCh38
rs753396715 Chr.3: 158653367-158653367 on Build GRCh38
rs926189715 Chr.3: 158653558-158653558 on Build GRCh38
rs890590376 Chr.3: 158653575-158653575 on Build GRCh38
rs771890880 Chr.3: 158653319-158653319 on Build GRCh38
rs770101036 Chr.3: 158653293-158653293 on Build GRCh38
rs1004082090 Chr.3: 158653674-158653674 on Build GRCh38
rs531887279 Chr.3: 158653456-158653456 on Build GRCh38
rs368676387 Chr.3: 158653482-158653482 on Build GRCh38
rs770039650 Chr.3: 158653433-158653433 on Build GRCh38
rs182872711 Chr.3: 158653630-158653630 on Build GRCh38
rs111345385 Chr.3: 158653440-158653440 on Build GRCh38
rs763546447 Chr.3: 158653366-158653366 on Build GRCh38
rs547093818 Chr.3: 158653620-158653620 on Build GRCh38
rs764670134 Chr.3: 158653503-158653503 on Build GRCh38
rs965034653 Chr.3: 158653320-158653320 on Build GRCh38
rs959808171 Chr.3: 158653430-158653430 on Build GRCh38
rs750029994 Chr.3: 158653510-158653510 on Build GRCh38
rs759440642 Chr.3: 158653312-158653312 on Build GRCh38
rs747535885 Chr.3: 158653275-158653275 on Build GRCh38
rs1009348771 Chr.3: 158653285-158653285 on Build GRCh38
rs140063324 Chr.3: 158653392-158653392 on Build GRCh38
rs752069831 Chr.3: 158653401-158653401 on Build GRCh38
rs761343297 Chr.3: 158653375-158653375 on Build GRCh38
rs145970222 Chr.3: 158653429-158653429 on Build GRCh38
rs773209906 Chr.3: 158653463-158653465 on Build GRCh38
rs775366703 Chr.3: 158653358-158653358 on Build GRCh38
rs766797056 Chr.3: 158653512-158653512 on Build GRCh38
rs934869857 Chr.3: 158653560-158653560 on Build GRCh38
rs547769190 Chr.3: 158653656-158653656 on Build GRCh38
rs755560938 Chr.3: 158653414-158653414 on Build GRCh38
rs1053209871 Chr.3: 158653569-158653569 on Build GRCh38
rs1043343558 Chr.3: 158653520-158653520 on Build GRCh38
rs775287626 Chr.3: 158653469-158653469 on Build GRCh38
rs945818683 Chr.3: 158653716-158653716 on Build GRCh38
rs535885259 Chr.3: 158653654-158653654 on Build GRCh38
rs944621086 Chr.3: 158653234-158653234 on Build GRCh38
rs1021136228 Chr.3: 158653400-158653400 on Build GRCh38
rs900511317 Chr.3: 158653633-158653633 on Build GRCh38
rs752014865 Chr.3: 158653513-158653513 on Build GRCh38
rs748329050 Chr.3: 158653288-158653288 on Build GRCh38
rs550421059 Chr.3: 158653457-158653457 on Build GRCh38
rs952603502 Chr.3: 158653667-158653667 on Build GRCh38
rs779114127 Chr.3: 158653396-158653396 on Build GRCh38
rs572518526 Chr.3: 158653295-158653295 on Build GRCh38
rs760637542 Chr.3: 158653330-158653330 on Build GRCh38
rs773985079 Chr.3: 158653306-158653306 on Build GRCh38
rs983205975 Chr.3: 158653616-158653616 on Build GRCh38
rs780573696 Chr.3: 158653399-158653399 on Build GRCh38
rs1054132634 Chr.3: 158653677-158653677 on Build GRCh38
rs112581636 Chr.3: 158653257-158653257 on Build GRCh38
rs768792924 Chr.3: 158653286-158653286 on Build GRCh38
rs776115676 Chr.3: 158653496-158653496 on Build GRCh38
rs950810293 Chr.3: 158653613-158653613 on Build GRCh38
rs768621926 Chr.3: 158653493-158653493 on Build GRCh38
rs748395191 Chr.3: 158653406-158653407 on Build GRCh38
rs187021781 Chr.3: 158653701-158653701 on Build GRCh38
rs921892115 Chr.3: 158653636-158653636 on Build GRCh38
rs1026873701 Chr.3: 158653363-158653363 on Build GRCh38
rs760266828 Chr.3: 158653379-158653379 on Build GRCh38
rs891566225 Chr.3: 158653381-158653381 on Build GRCh38
rs992952347 Chr.3: 158653437-158653437 on Build GRCh38
rs201689608 Chr.3: 158653282-158653282 on Build GRCh38
rs762547660 Chr.3: 158653511-158653511 on Build GRCh38
rs371938998 Chr.3: 158653325-158653325 on Build GRCh38

View Full Product Details