Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 167313787-167314333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBBX
Entrez Gene ID
79740
Gene Name
zinc finger B-box domain containing
Gene Aliases
-
UniGene
Hs.478143
Gene Chromosome Location
Chr.3: 167178399-167380405 on Build GRCh38
Transcripts
NM_001199201.1 NM_001199202.1 NM_024687.3 XM_011513144.1 XM_011513145.2 XM_011513148.2 XM_011513149.2 XM_011513151.2 XM_011513152.2 XM_011513153.2 XM_011513156.1 XM_017007186.1 XM_017007187.1 XM_017007188.1 XM_017007189.1 XM_017007190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7137216 c.1323C>T p.G441G Chr.3: 167314068-167314068 on Build GRCh38
5517086 c.1276T>A p.C426S Chr.3: 167314115-167314115 on Build GRCh38
1420426 c.1299G>T p.K433N Chr.3: 167314092-167314092 on Build GRCh38
5517087 c.1276T>A p.C426S Chr.3: 167314115-167314115 on Build GRCh38
1420425 c.1299G>T p.K433N Chr.3: 167314092-167314092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929106956 Chr.3: 167314124-167314124 on Build GRCh38
rs528697922 Chr.3: 167313846-167313846 on Build GRCh38
rs764512276 Chr.3: 167313949-167313949 on Build GRCh38
rs61737305 Chr.3: 167313973-167313973 on Build GRCh38
rs778275948 Chr.3: 167314045-167314045 on Build GRCh38
rs991213073 Chr.3: 167313922-167313922 on Build GRCh38
rs372865455 Chr.3: 167313967-167313967 on Build GRCh38
rs908045796 Chr.3: 167314081-167314081 on Build GRCh38
rs1051799279 Chr.3: 167314129-167314129 on Build GRCh38
rs758352137 Chr.3: 167313981-167313981 on Build GRCh38
rs894091749 Chr.3: 167313818-167313818 on Build GRCh38
rs147067456 Chr.3: 167313814-167313814 on Build GRCh38
rs749864015 Chr.3: 167314072-167314072 on Build GRCh38
rs1018189505 Chr.3: 167314268-167314268 on Build GRCh38
rs987114115 Chr.3: 167313914-167313914 on Build GRCh38
rs770283681 Chr.3: 167314033-167314033 on Build GRCh38
rs746322861 Chr.3: 167313973-167313973 on Build GRCh38
rs547273377 Chr.3: 167313847-167313847 on Build GRCh38
rs573456120 Chr.3: 167314154-167314154 on Build GRCh38
rs1043387000 Chr.3: 167314195-167314195 on Build GRCh38
rs777706259 Chr.3: 167314036-167314036 on Build GRCh38
rs952125010 Chr.3: 167314000-167314000 on Build GRCh38
rs1021982617 Chr.3: 167313826-167313826 on Build GRCh38
rs766312165 Chr.3: 167314150-167314150 on Build GRCh38
rs186557482 Chr.3: 167313961-167313961 on Build GRCh38
rs754209580 Chr.3: 167314280-167314280 on Build GRCh38
rs903642164 Chr.3: 167313845-167313845 on Build GRCh38
rs770292143 Chr.3: 167314152-167314153 on Build GRCh38
rs191822713 Chr.3: 167313823-167313823 on Build GRCh38
rs771995098 Chr.3: 167314106-167314106 on Build GRCh38
rs754630674 Chr.3: 167314012-167314012 on Build GRCh38
rs765086382 Chr.3: 167314139-167314139 on Build GRCh38
rs569668006 Chr.3: 167313907-167313907 on Build GRCh38
rs779488368 Chr.3: 167313990-167313990 on Build GRCh38
rs772978101 Chr.3: 167314147-167314147 on Build GRCh38
rs754541568 Chr.3: 167314153-167314153 on Build GRCh38
rs998620097 Chr.3: 167313898-167313898 on Build GRCh38
rs778327248 Chr.3: 167314166-167314166 on Build GRCh38
rs781025337 Chr.3: 167314013-167314013 on Build GRCh38
rs865911167 Chr.3: 167314176-167314176 on Build GRCh38
rs903590006 Chr.3: 167314224-167314224 on Build GRCh38
rs771443781 Chr.3: 167314082-167314082 on Build GRCh38
rs959979811 Chr.3: 167313909-167313909 on Build GRCh38
rs375036699 Chr.3: 167314025-167314025 on Build GRCh38
rs747757905 Chr.3: 167314030-167314030 on Build GRCh38
rs890951593 Chr.3: 167314254-167314254 on Build GRCh38
rs1008085002 Chr.3: 167314261-167314261 on Build GRCh38
rs534519913 Chr.3: 167314214-167314214 on Build GRCh38
rs571960998 Chr.3: 167313868-167313868 on Build GRCh38
rs999688947 Chr.3: 167314233-167314233 on Build GRCh38
rs759724853 Chr.3: 167313935-167313935 on Build GRCh38
rs775700752 Chr.3: 167314125-167314125 on Build GRCh38
rs184043170 Chr.3: 167313872-167313872 on Build GRCh38
rs764955003 Chr.3: 167313962-167313962 on Build GRCh38
rs369796514 Chr.3: 167313951-167313951 on Build GRCh38
rs1035407183 Chr.3: 167313908-167313908 on Build GRCh38
rs749041599 Chr.3: 167314048-167314048 on Build GRCh38
rs545318110 Chr.3: 167314277-167314277 on Build GRCh38
rs192717944 Chr.3: 167314248-167314248 on Build GRCh38
rs983600859 Chr.3: 167314069-167314069 on Build GRCh38
rs751061446 Chr.3: 167313993-167313993 on Build GRCh38
rs915547012 Chr.3: 167314019-167314019 on Build GRCh38
rs767748793 Chr.3: 167313942-167313942 on Build GRCh38
rs762848027 Chr.3: 167314148-167314148 on Build GRCh38
rs552791703 Chr.3: 167314221-167314221 on Build GRCh38
rs752462296 Chr.3: 167314157-167314157 on Build GRCh38
rs539387496 Chr.3: 167313869-167313869 on Build GRCh38
rs966845151 Chr.3: 167313871-167313871 on Build GRCh38
rs760821366 Chr.3: 167314133-167314133 on Build GRCh38
rs755811336 Chr.3: 167314163-167314163 on Build GRCh38
rs770979412 Chr.3: 167313933-167313933 on Build GRCh38
rs774334525 Chr.3: 167314098-167314098 on Build GRCh38
rs766831284 Chr.3: 167313817-167313817 on Build GRCh38
rs999287561 Chr.3: 167313855-167313855 on Build GRCh38
rs368899084 Chr.3: 167314046-167314046 on Build GRCh38
rs372606521 Chr.3: 167314099-167314099 on Build GRCh38
rs751417163 Chr.3: 167314151-167314151 on Build GRCh38
rs1031271562 Chr.3: 167313868-167313869 on Build GRCh38

View Full Product Details