Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170460099-170460610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A14
Entrez Gene ID
57709
Gene Name
solute carrier family 7 member 14
Gene Aliases
PPP1R142
UniGene
Hs.596660
Gene Chromosome Location
Chr.3: 170459554-170586074 on Build GRCh38
Transcripts
NM_020949.2

SNP Detail

SNP ID Chromosome Location
rs750746813 Chr.3: 170460163-170460163 on Build GRCh38
rs949528897 Chr.3: 170460334-170460334 on Build GRCh38
rs561360323 Chr.3: 170460442-170460442 on Build GRCh38
rs1014789849 Chr.3: 170460508-170460508 on Build GRCh38
rs533220115 Chr.3: 170460572-170460572 on Build GRCh38
rs386668163 Chr.3: 170460267-170460269 on Build GRCh38
rs1055443240 Chr.3: 170460417-170460417 on Build GRCh38
rs2287488 Chr.3: 170460267-170460267 on Build GRCh38
rs764817012 Chr.3: 170460513-170460513 on Build GRCh38
rs975926597 Chr.3: 170460215-170460215 on Build GRCh38
rs545290632 Chr.3: 170460347-170460347 on Build GRCh38
rs986601699 Chr.3: 170460564-170460564 on Build GRCh38
rs759275273 Chr.3: 170460448-170460448 on Build GRCh38
rs185290892 Chr.3: 170460390-170460390 on Build GRCh38
rs780125334 Chr.3: 170460193-170460193 on Build GRCh38
rs906559076 Chr.3: 170460480-170460480 on Build GRCh38
rs749722653 Chr.3: 170460218-170460218 on Build GRCh38
rs746385256 Chr.3: 170460327-170460327 on Build GRCh38
rs569665066 Chr.3: 170460141-170460141 on Build GRCh38
rs747572160 Chr.3: 170460518-170460518 on Build GRCh38
rs759249449 Chr.3: 170460461-170460461 on Build GRCh38
rs114978191 Chr.3: 170460399-170460399 on Build GRCh38
rs749571791 Chr.3: 170460241-170460241 on Build GRCh38
rs752194484 Chr.3: 170460514-170460514 on Build GRCh38
rs528541509 Chr.3: 170460503-170460503 on Build GRCh38
rs760463410 Chr.3: 170460340-170460340 on Build GRCh38
rs1005917275 Chr.3: 170460491-170460491 on Build GRCh38
rs768585892 Chr.3: 170460249-170460249 on Build GRCh38
rs181240787 Chr.3: 170460283-170460283 on Build GRCh38
rs947158184 Chr.3: 170460234-170460234 on Build GRCh38
rs555021447 Chr.3: 170460210-170460210 on Build GRCh38
rs1030333956 Chr.3: 170460216-170460216 on Build GRCh38
rs981396045 Chr.3: 170460149-170460149 on Build GRCh38
rs988380815 Chr.3: 170460295-170460295 on Build GRCh38
rs1037877544 Chr.3: 170460344-170460344 on Build GRCh38
rs2287489 Chr.3: 170460269-170460269 on Build GRCh38
rs76541465 Chr.3: 170460264-170460264 on Build GRCh38
rs991352360 Chr.3: 170460177-170460177 on Build GRCh38
rs563877961 Chr.3: 170460380-170460380 on Build GRCh38
rs1026398939 Chr.3: 170460461-170460461 on Build GRCh38
rs977177634 Chr.3: 170460545-170460545 on Build GRCh38
rs1014822524 Chr.3: 170460198-170460198 on Build GRCh38
rs1001170493 Chr.3: 170460156-170460156 on Build GRCh38
rs2287490 Chr.3: 170460517-170460517 on Build GRCh38
rs2287491 Chr.3: 170460534-170460534 on Build GRCh38
rs73040342 Chr.3: 170460164-170460164 on Build GRCh38

View Full Product Details