Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170996242-170996776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A2
Entrez Gene ID
6514
Gene Name
solute carrier family 2 member 2
Gene Aliases
GLUT2
UniGene
Hs.167584
Gene Chromosome Location
Chr.3: 170996341-171026979 on Build GRCh38
Transcripts
NM_000340.1 NM_001278658.1 NM_001278659.1 XM_011513087.2 XM_011513089.2

SNP Detail

SNP ID Chromosome Location
rs997578553 Chr.3: 170996663-170996663 on Build GRCh38
rs988203263 Chr.3: 170996390-170996390 on Build GRCh38
rs928555358 Chr.3: 170996466-170996466 on Build GRCh38
rs937911289 Chr.3: 170996522-170996527 on Build GRCh38
rs112957674 Chr.3: 170996554-170996554 on Build GRCh38
rs541058007 Chr.3: 170996542-170996542 on Build GRCh38
rs772261626 Chr.3: 170996526-170996526 on Build GRCh38
rs967238722 Chr.3: 170996413-170996413 on Build GRCh38
rs866750823 Chr.3: 170996600-170996600 on Build GRCh38
rs190326478 Chr.3: 170996325-170996325 on Build GRCh38
rs945207412 Chr.3: 170996611-170996611 on Build GRCh38
rs1021047546 Chr.3: 170996411-170996411 on Build GRCh38
rs1041773441 Chr.3: 170996634-170996634 on Build GRCh38
rs551252644 Chr.3: 170996486-170996486 on Build GRCh38
rs1009976007 Chr.3: 170996739-170996739 on Build GRCh38
rs963888147 Chr.3: 170996422-170996422 on Build GRCh38
rs991571904 Chr.3: 170996482-170996482 on Build GRCh38
rs767880748 Chr.3: 170996663-170996664 on Build GRCh38
rs997483592 Chr.3: 170996298-170996298 on Build GRCh38
rs138928037 Chr.3: 170996305-170996306 on Build GRCh38
rs535919268 Chr.3: 170996605-170996605 on Build GRCh38
rs1016930344 Chr.3: 170996691-170996691 on Build GRCh38
rs28579899 Chr.3: 170996696-170996696 on Build GRCh38
rs958370410 Chr.3: 170996372-170996372 on Build GRCh38
rs897563982 Chr.3: 170996522-170996522 on Build GRCh38
rs889380397 Chr.3: 170996664-170996664 on Build GRCh38
rs565839434 Chr.3: 170996431-170996431 on Build GRCh38
rs541564674 Chr.3: 170996481-170996481 on Build GRCh38
rs886058171 Chr.3: 170996739-170996739 on Build GRCh38
rs994085365 Chr.3: 170996710-170996710 on Build GRCh38
rs547190288 Chr.3: 170996417-170996417 on Build GRCh38
rs573454591 Chr.3: 170996643-170996643 on Build GRCh38
rs1007730372 Chr.3: 170996685-170996685 on Build GRCh38
rs564764920 Chr.3: 170996678-170996678 on Build GRCh38
rs1030316181 Chr.3: 170996321-170996321 on Build GRCh38
rs182503337 Chr.3: 170996483-170996483 on Build GRCh38
rs926301873 Chr.3: 170996518-170996518 on Build GRCh38
rs569662558 Chr.3: 170996535-170996535 on Build GRCh38
rs537828074 Chr.3: 170996537-170996537 on Build GRCh38
rs181123140 Chr.3: 170996675-170996675 on Build GRCh38
rs1053738564 Chr.3: 170996532-170996532 on Build GRCh38
rs753663594 Chr.3: 170996654-170996654 on Build GRCh38
rs903158742 Chr.3: 170996268-170996268 on Build GRCh38
rs941691459 Chr.3: 170996499-170996499 on Build GRCh38
rs981254810 Chr.3: 170996442-170996442 on Build GRCh38
rs571938295 Chr.3: 170996511-170996511 on Build GRCh38
rs55989805 Chr.3: 170996681-170996681 on Build GRCh38
rs55679742 Chr.3: 170996646-170996646 on Build GRCh38
rs1032314148 Chr.3: 170996365-170996365 on Build GRCh38
rs760619000 Chr.3: 170996592-170996592 on Build GRCh38
rs188353805 Chr.3: 170996558-170996558 on Build GRCh38
rs1026512319 Chr.3: 170996712-170996712 on Build GRCh38

View Full Product Details