Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170996566-170997091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A2
Entrez Gene ID
6514
Gene Name
solute carrier family 2 member 2
Gene Aliases
GLUT2
UniGene
Hs.167584
Gene Chromosome Location
Chr.3: 170996341-171026979 on Build GRCh38
Transcripts
NM_000340.1 NM_001278658.1 NM_001278659.1 XM_011513087.2 XM_011513089.2

SNP Detail

SNP ID Chromosome Location
rs561085655 Chr.3: 170997060-170997060 on Build GRCh38
rs997578553 Chr.3: 170996663-170996663 on Build GRCh38
rs943391907 Chr.3: 170996949-170996949 on Build GRCh38
rs866750823 Chr.3: 170996600-170996600 on Build GRCh38
rs1026896170 Chr.3: 170996995-170996995 on Build GRCh38
rs945207412 Chr.3: 170996611-170996611 on Build GRCh38
rs978309591 Chr.3: 170996909-170996909 on Build GRCh38
rs55669541 Chr.3: 170996771-170996771 on Build GRCh38
rs1041773441 Chr.3: 170996634-170996634 on Build GRCh38
rs185726034 Chr.3: 170996817-170996817 on Build GRCh38
rs1054076559 Chr.3: 170997030-170997030 on Build GRCh38
rs1009976007 Chr.3: 170996739-170996739 on Build GRCh38
rs767880748 Chr.3: 170996663-170996664 on Build GRCh38
rs139372968 Chr.3: 170996844-170996844 on Build GRCh38
rs751051821 Chr.3: 170996930-170996930 on Build GRCh38
rs543181298 Chr.3: 170996905-170996905 on Build GRCh38
rs535919268 Chr.3: 170996605-170996605 on Build GRCh38
rs775694029 Chr.3: 170996945-170996945 on Build GRCh38
rs575624852 Chr.3: 170996859-170996859 on Build GRCh38
rs915154282 Chr.3: 170996851-170996851 on Build GRCh38
rs547422449 Chr.3: 170996946-170996946 on Build GRCh38
rs1016930344 Chr.3: 170996691-170996691 on Build GRCh38
rs28579899 Chr.3: 170996696-170996696 on Build GRCh38
rs1035487106 Chr.3: 170996850-170996850 on Build GRCh38
rs889380397 Chr.3: 170996664-170996664 on Build GRCh38
rs886058171 Chr.3: 170996739-170996739 on Build GRCh38
rs971689457 Chr.3: 170996943-170996943 on Build GRCh38
rs994085365 Chr.3: 170996710-170996710 on Build GRCh38
rs780659539 Chr.3: 170996985-170996985 on Build GRCh38
rs959036976 Chr.3: 170996846-170996846 on Build GRCh38
rs991460014 Chr.3: 170996866-170996866 on Build GRCh38
rs573454591 Chr.3: 170996643-170996643 on Build GRCh38
rs56204521 Chr.3: 170996983-170996983 on Build GRCh38
rs1007730372 Chr.3: 170996685-170996685 on Build GRCh38
rs564764920 Chr.3: 170996678-170996678 on Build GRCh38
rs868164586 Chr.3: 170996887-170996887 on Build GRCh38
rs941732182 Chr.3: 170996939-170996939 on Build GRCh38
rs894036413 Chr.3: 170997036-170997036 on Build GRCh38
rs77733690 Chr.3: 170996994-170996994 on Build GRCh38
rs922477710 Chr.3: 170996931-170996931 on Build GRCh38
rs1037928394 Chr.3: 170996972-170996972 on Build GRCh38
rs561689694 Chr.3: 170996794-170996794 on Build GRCh38
rs970778526 Chr.3: 170996774-170996774 on Build GRCh38
rs181123140 Chr.3: 170996675-170996675 on Build GRCh38
rs753663594 Chr.3: 170996654-170996654 on Build GRCh38
rs943531821 Chr.3: 170997029-170997029 on Build GRCh38
rs909112257 Chr.3: 170996929-170996929 on Build GRCh38
rs140350942 Chr.3: 170996969-170996973 on Build GRCh38
rs763714148 Chr.3: 170996833-170996833 on Build GRCh38
rs757979511 Chr.3: 170996821-170996821 on Build GRCh38
rs55989805 Chr.3: 170996681-170996681 on Build GRCh38
rs55679742 Chr.3: 170996646-170996646 on Build GRCh38
rs12330351 Chr.3: 170997006-170997006 on Build GRCh38
rs760619000 Chr.3: 170996592-170996592 on Build GRCh38
rs981285768 Chr.3: 170996784-170996784 on Build GRCh38
rs1026512319 Chr.3: 170996712-170996712 on Build GRCh38
rs749737869 Chr.3: 170997037-170997037 on Build GRCh38

View Full Product Details