Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172632467-172633001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCEH1
Entrez Gene ID
57552
Gene Name
neutral cholesterol ester hydrolase 1
Gene Aliases
AADACL1, NCEH
UniGene
Hs.444099
Gene Chromosome Location
Chr.3: 172630645-172711218 on Build GRCh38
Transcripts
NM_001146276.1 NM_001146277.1 NM_001146278.1 NM_020792.4

SNP Detail

SNP ID Chromosome Location
rs1041454469 Chr.3: 172632901-172632901 on Build GRCh38
rs962852981 Chr.3: 172632657-172632657 on Build GRCh38
rs963239469 Chr.3: 172632888-172632888 on Build GRCh38
rs968996431 Chr.3: 172632955-172632955 on Build GRCh38
rs999734030 Chr.3: 172632970-172632970 on Build GRCh38
rs1011723359 Chr.3: 172632539-172632539 on Build GRCh38
rs1004895993 Chr.3: 172632868-172632868 on Build GRCh38
rs561706732 Chr.3: 172632678-172632678 on Build GRCh38
rs1055057 Chr.3: 172632572-172632572 on Build GRCh38
rs529274947 Chr.3: 172632682-172632682 on Build GRCh38
rs7432459 Chr.3: 172632550-172632550 on Build GRCh38
rs892769598 Chr.3: 172632816-172632816 on Build GRCh38
rs942599673 Chr.3: 172632796-172632796 on Build GRCh38
rs1040234035 Chr.3: 172632728-172632728 on Build GRCh38
rs531858422 Chr.3: 172632655-172632655 on Build GRCh38
rs981960064 Chr.3: 172632567-172632567 on Build GRCh38
rs1011338649 Chr.3: 172632870-172632870 on Build GRCh38
rs946811478 Chr.3: 172632706-172632706 on Build GRCh38
rs963143384 Chr.3: 172632517-172632517 on Build GRCh38
rs892915252 Chr.3: 172632508-172632508 on Build GRCh38
rs902687814 Chr.3: 172632940-172632940 on Build GRCh38
rs918425528 Chr.3: 172632686-172632686 on Build GRCh38
rs927655673 Chr.3: 172632971-172632971 on Build GRCh38
rs370635719 Chr.3: 172632560-172632560 on Build GRCh38
rs539597288 Chr.3: 172632976-172632976 on Build GRCh38
rs1035231186 Chr.3: 172632615-172632615 on Build GRCh38
rs1029893033 Chr.3: 172632825-172632825 on Build GRCh38
rs999788693 Chr.3: 172632592-172632592 on Build GRCh38
rs35145370 Chr.3: 172632975-172632976 on Build GRCh38
rs191771655 Chr.3: 172632663-172632663 on Build GRCh38
rs186938481 Chr.3: 172632647-172632647 on Build GRCh38
rs373723545 Chr.3: 172632742-172632742 on Build GRCh38
rs887376791 Chr.3: 172632863-172632863 on Build GRCh38
rs533406356 Chr.3: 172632573-172632574 on Build GRCh38
rs372681576 Chr.3: 172632523-172632523 on Build GRCh38
rs973136965 Chr.3: 172632947-172632947 on Build GRCh38
rs1015065590 Chr.3: 172632516-172632516 on Build GRCh38
rs182114699 Chr.3: 172632645-172632645 on Build GRCh38
rs981760355 Chr.3: 172632966-172632966 on Build GRCh38
rs998196362 Chr.3: 172632746-172632746 on Build GRCh38
rs547866245 Chr.3: 172632801-172632801 on Build GRCh38
rs929771022 Chr.3: 172632734-172632734 on Build GRCh38
rs745432826 Chr.3: 172632683-172632683 on Build GRCh38
rs1050879478 Chr.3: 172632777-172632777 on Build GRCh38
rs867627353 Chr.3: 172632660-172632660 on Build GRCh38
rs951494635 Chr.3: 172632493-172632495 on Build GRCh38
rs916557981 Chr.3: 172632656-172632656 on Build GRCh38
rs1022488689 Chr.3: 172632549-172632549 on Build GRCh38
rs973589501 Chr.3: 172632522-172632522 on Build GRCh38
rs771597410 Chr.3: 172632787-172632787 on Build GRCh38
rs772532717 Chr.3: 172632501-172632502 on Build GRCh38
rs1035180104 Chr.3: 172632975-172632975 on Build GRCh38
rs1023349371 Chr.3: 172632952-172632952 on Build GRCh38
rs566075505 Chr.3: 172632959-172632959 on Build GRCh38

View Full Product Details