Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172956537-172957055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA16
Entrez Gene ID
83893
Gene Name
spermatogenesis associated 16
Gene Aliases
NYD-SP12, SPGF6
UniGene
Hs.444236
Gene Chromosome Location
Chr.3: 172889357-173141268 on Build GRCh38
Transcripts
NM_031955.5 XM_006713778.2 XM_017007308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1041367 c.995C>T p.A332V Chr.3: 172956763-172956763 on Build GRCh38
3915388 c.1041C>T p.F347F Chr.3: 172956717-172956717 on Build GRCh38
1041368 c.974C>T p.S325L Chr.3: 172956784-172956784 on Build GRCh38
3205202 c.996G>A p.A332A Chr.3: 172956762-172956762 on Build GRCh38
1733609 c.955A>G p.T319A Chr.3: 172956803-172956803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778798906 Chr.3: 172956833-172956836 on Build GRCh38
rs779318248 Chr.3: 172956869-172956869 on Build GRCh38
rs745618585 Chr.3: 172956833-172956833 on Build GRCh38
rs776543945 Chr.3: 172956672-172956672 on Build GRCh38
rs907030864 Chr.3: 172956621-172956621 on Build GRCh38
rs564378486 Chr.3: 172956784-172956784 on Build GRCh38
rs774297969 Chr.3: 172956650-172956650 on Build GRCh38
rs13064708 Chr.3: 172956803-172956803 on Build GRCh38
rs769566618 Chr.3: 172956789-172956789 on Build GRCh38
rs954861029 Chr.3: 172956752-172956755 on Build GRCh38
rs776676290 Chr.3: 172956831-172956831 on Build GRCh38
rs765875315 Chr.3: 172956687-172956687 on Build GRCh38
rs529638825 Chr.3: 172956840-172956840 on Build GRCh38
rs562094988 Chr.3: 172956592-172956592 on Build GRCh38
rs753050752 Chr.3: 172956733-172956733 on Build GRCh38
rs955685835 Chr.3: 172956988-172956988 on Build GRCh38
rs146082029 Chr.3: 172956705-172956705 on Build GRCh38
rs377088060 Chr.3: 172956655-172956655 on Build GRCh38
rs761767334 Chr.3: 172956681-172956681 on Build GRCh38
rs544088248 Chr.3: 172956763-172956763 on Build GRCh38
rs772915401 Chr.3: 172956809-172956809 on Build GRCh38
rs562174338 Chr.3: 172956837-172956837 on Build GRCh38
rs779085714 Chr.3: 172956754-172956754 on Build GRCh38
rs954021436 Chr.3: 172956925-172956925 on Build GRCh38
rs973774768 Chr.3: 172956796-172956796 on Build GRCh38
rs754174712 Chr.3: 172956857-172956857 on Build GRCh38
rs200673599 Chr.3: 172956715-172956715 on Build GRCh38
rs199512283 Chr.3: 172956685-172956685 on Build GRCh38
rs949540930 Chr.3: 172956645-172956645 on Build GRCh38
rs532404090 Chr.3: 172956747-172956747 on Build GRCh38
rs375612371 Chr.3: 172956783-172956783 on Build GRCh38
rs978525753 Chr.3: 172956926-172956926 on Build GRCh38
rs888442850 Chr.3: 172956764-172956764 on Build GRCh38
rs547922321 Chr.3: 172956867-172956867 on Build GRCh38
rs149140561 Chr.3: 172956677-172956677 on Build GRCh38
rs138951243 Chr.3: 172956762-172956762 on Build GRCh38
rs764784167 Chr.3: 172956683-172956683 on Build GRCh38
rs755460688 Chr.3: 172956729-172956729 on Build GRCh38
rs368587027 Chr.3: 172956817-172956817 on Build GRCh38
rs949612902 Chr.3: 172957032-172957032 on Build GRCh38
rs776767984 Chr.3: 172956770-172956770 on Build GRCh38
rs988348639 Chr.3: 172956591-172956591 on Build GRCh38
rs988463860 Chr.3: 172957004-172957004 on Build GRCh38
rs1046527607 Chr.3: 172956682-172956682 on Build GRCh38
rs143211035 Chr.3: 172956872-172956872 on Build GRCh38
rs781691039 Chr.3: 172956732-172956732 on Build GRCh38
rs199868908 Chr.3: 172956746-172956746 on Build GRCh38
rs1028117645 Chr.3: 172956899-172956899 on Build GRCh38
rs189773685 Chr.3: 172957023-172957023 on Build GRCh38
rs778158832 Chr.3: 172956856-172956856 on Build GRCh38
rs11926761 Chr.3: 172956619-172956619 on Build GRCh38
rs928059420 Chr.3: 172956986-172956986 on Build GRCh38
rs751095220 Chr.3: 172956694-172956694 on Build GRCh38
rs370161882 Chr.3: 172956748-172956748 on Build GRCh38
rs916586571 Chr.3: 172956801-172956801 on Build GRCh38
rs756580561 Chr.3: 172956844-172956844 on Build GRCh38
rs770982158 Chr.3: 172956640-172956640 on Build GRCh38
rs1046466379 Chr.3: 172956946-172956946 on Build GRCh38
rs948168647 Chr.3: 172956609-172956609 on Build GRCh38
rs1033213904 Chr.3: 172956942-172956942 on Build GRCh38
rs764172678 Chr.3: 172956676-172956676 on Build GRCh38
rs995938659 Chr.3: 172956722-172956722 on Build GRCh38
rs908110846 Chr.3: 172956778-172956778 on Build GRCh38
rs762518840 Chr.3: 172956686-172956686 on Build GRCh38
rs916924843 Chr.3: 172957006-172957006 on Build GRCh38
rs766000019 Chr.3: 172956829-172956829 on Build GRCh38
rs964777837 Chr.3: 172956698-172956698 on Build GRCh38

View Full Product Details