Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 179413301-179413802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB4
Entrez Gene ID
59345
Gene Name
G protein subunit beta 4
Gene Aliases
CMTD1F
UniGene
Hs.173030
Gene Chromosome Location
Chr.3: 179396088-179451583 on Build GRCh38
Transcripts
NM_021629.3 XM_005247692.2 XM_006713721.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4149678 c.583G>C p.D195H Chr.3: 179413528-179413528 on Build GRCh38
1421058 c.669G>A p.T223T Chr.3: 179413442-179413442 on Build GRCh38
1421059 c.514G>A p.E172K Chr.3: 179413597-179413597 on Build GRCh38
729793 c.494C>T p.T165I Chr.3: 179413718-179413718 on Build GRCh38
3590424 c.455T>C p.L152S Chr.3: 179413757-179413757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199615251 Chr.3: 179413452-179413452 on Build GRCh38
rs375356541 Chr.3: 179413692-179413692 on Build GRCh38
rs755864696 Chr.3: 179413597-179413597 on Build GRCh38
rs776836938 Chr.3: 179413519-179413519 on Build GRCh38
rs766413591 Chr.3: 179413420-179413420 on Build GRCh38
rs760894139 Chr.3: 179413515-179413515 on Build GRCh38
rs751438259 Chr.3: 179413442-179413442 on Build GRCh38
rs957501114 Chr.3: 179413335-179413335 on Build GRCh38
rs367709368 Chr.3: 179413598-179413598 on Build GRCh38
rs771469568 Chr.3: 179413480-179413480 on Build GRCh38
rs569442187 Chr.3: 179413657-179413657 on Build GRCh38
rs755596977 Chr.3: 179413466-179413466 on Build GRCh38
rs762690340 Chr.3: 179413589-179413589 on Build GRCh38
rs775578986 Chr.3: 179413513-179413513 on Build GRCh38
rs748257526 Chr.3: 179413719-179413719 on Build GRCh38
rs954963340 Chr.3: 179413541-179413541 on Build GRCh38
rs753438422 Chr.3: 179413644-179413644 on Build GRCh38
rs768745441 Chr.3: 179413517-179413517 on Build GRCh38
rs748747265 Chr.3: 179413471-179413471 on Build GRCh38
rs144385061 Chr.3: 179413443-179413443 on Build GRCh38
rs756637757 Chr.3: 179413664-179413674 on Build GRCh38
rs369750242 Chr.3: 179413697-179413697 on Build GRCh38
rs375876321 Chr.3: 179413373-179413373 on Build GRCh38
rs752798775 Chr.3: 179413646-179413648 on Build GRCh38
rs556759178 Chr.3: 179413537-179413537 on Build GRCh38
rs747328798 Chr.3: 179413687-179413687 on Build GRCh38
rs569616038 Chr.3: 179413381-179413381 on Build GRCh38
rs111501349 Chr.3: 179413773-179413773 on Build GRCh38
rs755210355 Chr.3: 179413540-179413541 on Build GRCh38
rs375864239 Chr.3: 179413387-179413387 on Build GRCh38
rs149704487 Chr.3: 179413362-179413362 on Build GRCh38
rs746253436 Chr.3: 179413490-179413490 on Build GRCh38
rs372819629 Chr.3: 179413396-179413396 on Build GRCh38
rs774377330 Chr.3: 179413552-179413552 on Build GRCh38
rs200527279 Chr.3: 179413686-179413686 on Build GRCh38
rs772386850 Chr.3: 179413499-179413499 on Build GRCh38
rs774430510 Chr.3: 179413752-179413752 on Build GRCh38
rs756429902 Chr.3: 179413536-179413539 on Build GRCh38
rs1016827159 Chr.3: 179413400-179413400 on Build GRCh38
rs538233963 Chr.3: 179413414-179413414 on Build GRCh38
rs767366352 Chr.3: 179413577-179413577 on Build GRCh38
rs769949519 Chr.3: 179413721-179413721 on Build GRCh38
rs766466679 Chr.3: 179413545-179413545 on Build GRCh38
rs759431955 Chr.3: 179413571-179413571 on Build GRCh38
rs779372670 Chr.3: 179413481-179413481 on Build GRCh38
rs372351074 Chr.3: 179413648-179413648 on Build GRCh38
rs1005993737 Chr.3: 179413353-179413353 on Build GRCh38
rs764168618 Chr.3: 179413392-179413392 on Build GRCh38
rs775641492 Chr.3: 179413382-179413382 on Build GRCh38
rs767744394 Chr.3: 179413628-179413629 on Build GRCh38
rs371996539 Chr.3: 179413649-179413649 on Build GRCh38
rs772933399 Chr.3: 179413461-179413461 on Build GRCh38
rs758775096 Chr.3: 179413685-179413685 on Build GRCh38
rs1031628985 Chr.3: 179413333-179413333 on Build GRCh38

View Full Product Details