Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183816752-183817244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP6D1
Entrez Gene ID
79929
Gene Name
MAP6 domain containing 1
Gene Aliases
MAPO6D1, SL21
UniGene
Hs.478465
Gene Chromosome Location
Chr.3: 183815876-183825605 on Build GRCh38
Transcripts
NM_024871.2

SNP Detail

SNP ID Chromosome Location
rs376986843 Chr.3: 183817174-183817174 on Build GRCh38
rs185652406 Chr.3: 183816962-183816962 on Build GRCh38
rs971941434 Chr.3: 183817018-183817018 on Build GRCh38
rs181105166 Chr.3: 183816958-183816958 on Build GRCh38
rs759748735 Chr.3: 183816943-183816943 on Build GRCh38
rs564962900 Chr.3: 183816957-183816957 on Build GRCh38
rs547012198 Chr.3: 183817144-183817144 on Build GRCh38
rs1013468636 Chr.3: 183816839-183816839 on Build GRCh38
rs576336578 Chr.3: 183816865-183816865 on Build GRCh38
rs543266029 Chr.3: 183816947-183816947 on Build GRCh38
rs751342936 Chr.3: 183817070-183817070 on Build GRCh38
rs938673196 Chr.3: 183816917-183816918 on Build GRCh38
rs1010814195 Chr.3: 183817043-183817043 on Build GRCh38
rs574112481 Chr.3: 183816941-183816941 on Build GRCh38
rs531107254 Chr.3: 183817123-183817123 on Build GRCh38
rs963205595 Chr.3: 183817009-183817009 on Build GRCh38
rs181632135 Chr.3: 183817140-183817140 on Build GRCh38
rs964676710 Chr.3: 183816780-183816780 on Build GRCh38
rs558973166 Chr.3: 183817004-183817004 on Build GRCh38
rs376788207 Chr.3: 183816815-183816815 on Build GRCh38
rs769035965 Chr.3: 183817186-183817186 on Build GRCh38
rs2255019 Chr.3: 183817125-183817125 on Build GRCh38
rs954245531 Chr.3: 183817124-183817124 on Build GRCh38
rs1042853527 Chr.3: 183817075-183817075 on Build GRCh38
rs554868297 Chr.3: 183817213-183817213 on Build GRCh38
rs1056807490 Chr.3: 183817066-183817066 on Build GRCh38
rs761901824 Chr.3: 183816811-183816811 on Build GRCh38
rs776051691 Chr.3: 183817166-183817166 on Build GRCh38
rs772450671 Chr.3: 183817165-183817165 on Build GRCh38
rs750426448 Chr.3: 183816802-183816802 on Build GRCh38
rs867166498 Chr.3: 183817142-183817142 on Build GRCh38
rs752778407 Chr.3: 183817122-183817122 on Build GRCh38
rs999092855 Chr.3: 183816866-183816866 on Build GRCh38
rs189944617 Chr.3: 183817047-183817047 on Build GRCh38
rs977924964 Chr.3: 183817082-183817082 on Build GRCh38
rs948420774 Chr.3: 183817193-183817193 on Build GRCh38
rs761059739 Chr.3: 183817181-183817181 on Build GRCh38
rs540286868 Chr.3: 183817100-183817100 on Build GRCh38
rs1047499067 Chr.3: 183816977-183816977 on Build GRCh38
rs867958101 Chr.3: 183817183-183817183 on Build GRCh38
rs900381269 Chr.3: 183817013-183817013 on Build GRCh38
rs919209628 Chr.3: 183817046-183817046 on Build GRCh38
rs758239217 Chr.3: 183816890-183816890 on Build GRCh38
rs369660922 Chr.3: 183817093-183817093 on Build GRCh38
rs965364550 Chr.3: 183816933-183816933 on Build GRCh38
rs999061752 Chr.3: 183817083-183817083 on Build GRCh38
rs1011010175 Chr.3: 183817218-183817218 on Build GRCh38
rs563656252 Chr.3: 183817101-183817101 on Build GRCh38
rs1021808028 Chr.3: 183816848-183816848 on Build GRCh38
rs916126302 Chr.3: 183816930-183816930 on Build GRCh38
rs553908883 Chr.3: 183816945-183816946 on Build GRCh38
rs777123778 Chr.3: 183817150-183817150 on Build GRCh38
rs746424540 Chr.3: 183817163-183817163 on Build GRCh38
rs2255015 Chr.3: 183817188-183817188 on Build GRCh38
rs530309974 Chr.3: 183817005-183817005 on Build GRCh38
rs948517581 Chr.3: 183817072-183817072 on Build GRCh38
rs780756797 Chr.3: 183817109-183817109 on Build GRCh38
rs938388386 Chr.3: 183816794-183816794 on Build GRCh38
rs989678976 Chr.3: 183816783-183816783 on Build GRCh38
rs992858326 Chr.3: 183816928-183816928 on Build GRCh38
rs904680272 Chr.3: 183817078-183817078 on Build GRCh38
rs148164193 Chr.3: 183817138-183817138 on Build GRCh38
rs890222829 Chr.3: 183817001-183817001 on Build GRCh38
rs750252262 Chr.3: 183816876-183816876 on Build GRCh38
rs867710528 Chr.3: 183817107-183817107 on Build GRCh38
rs1057275924 Chr.3: 183816798-183816798 on Build GRCh38
rs920871649 Chr.3: 183817147-183817148 on Build GRCh38
rs909067077 Chr.3: 183817008-183817008 on Build GRCh38

View Full Product Details