Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185443260-185443798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K13
Entrez Gene ID
9175
Gene Name
mitogen-activated protein kinase kinase kinase 13
Gene Aliases
LZK, MEKK13, MLK
UniGene
Hs.591306
Gene Chromosome Location
Chr.3: 185282938-185489094 on Build GRCh38
Transcripts
NM_001242314.1 NM_001242317.1 NM_004721.4 XM_011513310.2 XM_017007456.1 XM_017007457.1 XM_017007458.1 XM_017007459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1633034 c.827T>C p.I276T Chr.3: 185443612-185443612 on Build GRCh38
6971636 c.350C>G p.T117R Chr.3: 185443567-185443567 on Build GRCh38
3781118 c.400C>T p.R134C Chr.3: 185443617-185443617 on Build GRCh38
6917168 c.737G>A p.R246Q Chr.3: 185443522-185443522 on Build GRCh38
6973698 c.777G>C p.W259C Chr.3: 185443562-185443562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764242255 Chr.3: 185443522-185443522 on Build GRCh38
rs375457810 Chr.3: 185443506-185443506 on Build GRCh38
rs900413015 Chr.3: 185443591-185443591 on Build GRCh38
rs766638708 Chr.3: 185443548-185443548 on Build GRCh38
rs769390178 Chr.3: 185443472-185443472 on Build GRCh38
rs556754135 Chr.3: 185443713-185443713 on Build GRCh38
rs577644672 Chr.3: 185443714-185443714 on Build GRCh38
rs188088193 Chr.3: 185443731-185443731 on Build GRCh38
rs945444889 Chr.3: 185443363-185443363 on Build GRCh38
rs550046997 Chr.3: 185443350-185443350 on Build GRCh38
rs374237213 Chr.3: 185443380-185443380 on Build GRCh38
rs780508755 Chr.3: 185443456-185443456 on Build GRCh38
rs776809053 Chr.3: 185443528-185443528 on Build GRCh38
rs115725370 Chr.3: 185443698-185443698 on Build GRCh38
rs1003712733 Chr.3: 185443763-185443763 on Build GRCh38
rs755371719 Chr.3: 185443556-185443556 on Build GRCh38
rs752222546 Chr.3: 185443401-185443401 on Build GRCh38
rs765601571 Chr.3: 185443545-185443545 on Build GRCh38
rs200723196 Chr.3: 185443586-185443586 on Build GRCh38
rs533941966 Chr.3: 185443529-185443529 on Build GRCh38
rs765725489 Chr.3: 185443407-185443407 on Build GRCh38
rs770777601 Chr.3: 185443672-185443672 on Build GRCh38
rs1034299180 Chr.3: 185443774-185443774 on Build GRCh38
rs540031137 Chr.3: 185443708-185443708 on Build GRCh38
rs574041234 Chr.3: 185443689-185443689 on Build GRCh38
rs752118068 Chr.3: 185443555-185443555 on Build GRCh38
rs1010543891 Chr.3: 185443426-185443426 on Build GRCh38
rs770631849 Chr.3: 185443514-185443514 on Build GRCh38
rs970858925 Chr.3: 185443285-185443285 on Build GRCh38
rs368504985 Chr.3: 185443568-185443568 on Build GRCh38
rs371199844 Chr.3: 185443468-185443468 on Build GRCh38
rs143565195 Chr.3: 185443340-185443340 on Build GRCh38
rs758898925 Chr.3: 185443547-185443547 on Build GRCh38
rs749122155 Chr.3: 185443494-185443494 on Build GRCh38
rs747466011 Chr.3: 185443441-185443441 on Build GRCh38
rs889537345 Chr.3: 185443353-185443353 on Build GRCh38
rs569242652 Chr.3: 185443354-185443354 on Build GRCh38
rs758810422 Chr.3: 185443415-185443415 on Build GRCh38
rs557844192 Chr.3: 185443397-185443397 on Build GRCh38
rs141609876 Chr.3: 185443546-185443546 on Build GRCh38
rs750763481 Chr.3: 185443414-185443414 on Build GRCh38
rs757971984 Chr.3: 185443445-185443445 on Build GRCh38
rs566824809 Chr.3: 185443518-185443518 on Build GRCh38
rs755591040 Chr.3: 185443575-185443575 on Build GRCh38
rs772829736 Chr.3: 185443481-185443481 on Build GRCh38
rs56315431 Chr.3: 185443580-185443580 on Build GRCh38
rs770044702 Chr.3: 185443312-185443312 on Build GRCh38
rs757485772 Chr.3: 185443404-185443404 on Build GRCh38
rs780650204 Chr.3: 185443422-185443422 on Build GRCh38
rs376304258 Chr.3: 185443683-185443683 on Build GRCh38
rs759336796 Chr.3: 185443521-185443521 on Build GRCh38
rs989489189 Chr.3: 185443351-185443351 on Build GRCh38
rs915271009 Chr.3: 185443358-185443358 on Build GRCh38
rs906955656 Chr.3: 185443723-185443723 on Build GRCh38
rs35492967 Chr.3: 185443721-185443722 on Build GRCh38
rs545060902 Chr.3: 185443719-185443719 on Build GRCh38
rs982697481 Chr.3: 185443298-185443298 on Build GRCh38
rs574698720 Chr.3: 185443668-185443668 on Build GRCh38
rs780778687 Chr.3: 185443566-185443566 on Build GRCh38

View Full Product Details