Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2249996-2250499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4800
Entrez Gene ID
100616358
Gene Name
microRNA 4800
Gene Aliases
mir-4800
UniGene
Gene Chromosome Location
Chr.4: 2250077-2250156 on Build GRCh38
Transcripts
NR_039964.1 NM_006454.2 XM_011513388.2

Gene Symbol
MXD4
Entrez Gene ID
10608
Gene Name
MAX dimerization protein 4
Gene Aliases
MAD4, MST149, MSTP149, bHLHc12
UniGene
Hs.655020
Gene Chromosome Location
Chr.4: 2247432-2262143 on Build GRCh38
Transcripts
NR_039964.1 NM_006454.2 XM_011513388.2

SNP Detail

SNP ID Chromosome Location
rs769948735 Chr.4: 2250066-2250067 on Build GRCh38
rs374688743 Chr.4: 2250127-2250127 on Build GRCh38
rs556600842 Chr.4: 2250061-2250061 on Build GRCh38
rs562559045 Chr.4: 2250302-2250302 on Build GRCh38
rs764708793 Chr.4: 2250106-2250106 on Build GRCh38
rs962277119 Chr.4: 2250295-2250295 on Build GRCh38
rs992024573 Chr.4: 2250193-2250193 on Build GRCh38
rs924505988 Chr.4: 2250221-2250221 on Build GRCh38
rs893004234 Chr.4: 2250239-2250239 on Build GRCh38
rs1001217012 Chr.4: 2250388-2250388 on Build GRCh38
rs1053193426 Chr.4: 2250040-2250040 on Build GRCh38
rs767539762 Chr.4: 2250432-2250441 on Build GRCh38
rs1052682 Chr.4: 2250474-2250474 on Build GRCh38
rs539805722 Chr.4: 2250393-2250393 on Build GRCh38
rs535656716 Chr.4: 2250460-2250460 on Build GRCh38
rs1015291995 Chr.4: 2250294-2250294 on Build GRCh38
rs776542469 Chr.4: 2250038-2250038 on Build GRCh38
rs531580878 Chr.4: 2250307-2250307 on Build GRCh38
rs570786698 Chr.4: 2250392-2250392 on Build GRCh38
rs527559612 Chr.4: 2250361-2250361 on Build GRCh38
rs909231000 Chr.4: 2250292-2250292 on Build GRCh38
rs568375492 Chr.4: 2250360-2250360 on Build GRCh38
rs1049904403 Chr.4: 2250257-2250257 on Build GRCh38
rs1052685 Chr.4: 2250447-2250447 on Build GRCh38
rs751884475 Chr.4: 2250314-2250314 on Build GRCh38
rs559864870 Chr.4: 2250211-2250211 on Build GRCh38
rs993911107 Chr.4: 2250449-2250449 on Build GRCh38
rs576264332 Chr.4: 2250115-2250115 on Build GRCh38
rs555920639 Chr.4: 2250468-2250468 on Build GRCh38
rs761356585 Chr.4: 2250058-2250058 on Build GRCh38
rs532111217 Chr.4: 2250219-2250219 on Build GRCh38
rs943361027 Chr.4: 2250300-2250300 on Build GRCh38
rs61789367 Chr.4: 2250166-2250166 on Build GRCh38
rs1019173696 Chr.4: 2250081-2250081 on Build GRCh38
rs996605525 Chr.4: 2250240-2250240 on Build GRCh38
rs540155874 Chr.4: 2250290-2250290 on Build GRCh38
rs543840480 Chr.4: 2250458-2250458 on Build GRCh38
rs201952115 Chr.4: 2250199-2250199 on Build GRCh38
rs548354801 Chr.4: 2250308-2250308 on Build GRCh38
rs149969742 Chr.4: 2250469-2250469 on Build GRCh38
rs1053729915 Chr.4: 2250346-2250346 on Build GRCh38
rs934600586 Chr.4: 2250222-2250222 on Build GRCh38
rs746016577 Chr.4: 2250100-2250100 on Build GRCh38
rs868531986 Chr.4: 2250139-2250139 on Build GRCh38
rs969731675 Chr.4: 2250105-2250105 on Build GRCh38
rs755358306 Chr.4: 2250362-2250362 on Build GRCh38
rs997027245 Chr.4: 2250039-2250039 on Build GRCh38
rs920793163 Chr.4: 2250157-2250157 on Build GRCh38
rs891959799 Chr.4: 2250352-2250352 on Build GRCh38
rs763507461 Chr.4: 2250158-2250158 on Build GRCh38
rs1054233454 Chr.4: 2250023-2250023 on Build GRCh38
rs1011822866 Chr.4: 2250077-2250077 on Build GRCh38
rs562090462 Chr.4: 2250155-2250155 on Build GRCh38
rs2233043 Chr.4: 2250287-2250287 on Build GRCh38
rs916002830 Chr.4: 2250198-2250198 on Build GRCh38
rs952273810 Chr.4: 2250334-2250334 on Build GRCh38
rs115519692 Chr.4: 2250387-2250387 on Build GRCh38
rs1054203627 Chr.4: 2250235-2250235 on Build GRCh38
rs781587060 Chr.4: 2250373-2250373 on Build GRCh38
rs556281141 Chr.4: 2250434-2250434 on Build GRCh38

View Full Product Details