Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115840817-115841318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHLH2
Entrez Gene ID
4808
Gene Name
nescient helix-loop-helix 2
Gene Aliases
HEN2, NSCL2, bHLHa34
UniGene
Hs.46296
Gene Chromosome Location
Chr.1: 115829707-115841127 on Build GRCh38
Transcripts
NM_005599.3 XM_017001370.1

SNP Detail

SNP ID Chromosome Location
rs538535054 Chr.1: 115840847-115840847 on Build GRCh38
rs1018218749 Chr.1: 115841013-115841013 on Build GRCh38
rs775043149 Chr.1: 115840854-115840854 on Build GRCh38
rs964179545 Chr.1: 115841218-115841218 on Build GRCh38
rs905423635 Chr.1: 115840909-115840909 on Build GRCh38
rs908798710 Chr.1: 115841269-115841269 on Build GRCh38
rs1047709614 Chr.1: 115840859-115840859 on Build GRCh38
rs1029663378 Chr.1: 115841041-115841041 on Build GRCh38
rs775109877 Chr.1: 115840937-115840937 on Build GRCh38
rs910036116 Chr.1: 115840861-115840861 on Build GRCh38
rs968563958 Chr.1: 115841004-115841004 on Build GRCh38
rs951573003 Chr.1: 115841068-115841068 on Build GRCh38
rs963925007 Chr.1: 115841014-115841014 on Build GRCh38
rs895470413 Chr.1: 115841273-115841273 on Build GRCh38
rs377289069 Chr.1: 115840917-115840917 on Build GRCh38
rs1047332326 Chr.1: 115840921-115840921 on Build GRCh38
rs941629234 Chr.1: 115840878-115840878 on Build GRCh38
rs1009980622 Chr.1: 115840965-115840965 on Build GRCh38
rs184201611 Chr.1: 115840996-115840996 on Build GRCh38
rs533517044 Chr.1: 115841055-115841055 on Build GRCh38
rs909931828 Chr.1: 115841094-115841094 on Build GRCh38
rs1045306721 Chr.1: 115840896-115840896 on Build GRCh38
rs975620449 Chr.1: 115840881-115840881 on Build GRCh38
rs1056248247 Chr.1: 115841032-115841032 on Build GRCh38
rs908243362 Chr.1: 115840975-115840975 on Build GRCh38
rs976754501 Chr.1: 115841028-115841028 on Build GRCh38
rs193100666 Chr.1: 115840845-115840845 on Build GRCh38
rs187434172 Chr.1: 115841243-115841243 on Build GRCh38
rs922794595 Chr.1: 115841223-115841223 on Build GRCh38
rs945810850 Chr.1: 115841275-115841275 on Build GRCh38
rs1033396404 Chr.1: 115841183-115841183 on Build GRCh38
rs56788298 Chr.1: 115841268-115841268 on Build GRCh38
rs573647098 Chr.1: 115841216-115841216 on Build GRCh38
rs572888241 Chr.1: 115841005-115841005 on Build GRCh38
rs939602436 Chr.1: 115841246-115841246 on Build GRCh38
rs555334568 Chr.1: 115841125-115841125 on Build GRCh38
rs765219133 Chr.1: 115841149-115841149 on Build GRCh38
rs941576943 Chr.1: 115841283-115841283 on Build GRCh38
rs1002335114 Chr.1: 115841031-115841031 on Build GRCh38
rs1038054466 Chr.1: 115841294-115841294 on Build GRCh38
rs761856850 Chr.1: 115841093-115841093 on Build GRCh38
rs59577930 Chr.1: 115841221-115841221 on Build GRCh38
rs539532339 Chr.1: 115840919-115840919 on Build GRCh38
rs560531477 Chr.1: 115840911-115840911 on Build GRCh38
rs934255061 Chr.1: 115840920-115840920 on Build GRCh38
rs980871150 Chr.1: 115841172-115841172 on Build GRCh38
rs1000208582 Chr.1: 115841179-115841179 on Build GRCh38

View Full Product Details