Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 9908000-9908510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A9
Entrez Gene ID
56606
Gene Name
solute carrier family 2 member 9
Gene Aliases
GLUT9, GLUTX, UAQTL2, URATv1
UniGene
Hs.444612
Gene Chromosome Location
Chr.4: 9771125-10040248 on Build GRCh38
Transcripts
NM_001001290.1 NM_020041.2 XM_006713968.3 XM_006713969.2 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513859.2 XM_011513860.2 XM_011513861.2 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513865.2 XM_011513866.2 XM_011513867.2 XM_011513868.2 XM_017008457.1 XM_017008458.1 XM_017008459.1 XM_017008460.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5569771 c.1080A>C p.T360T Chr.4: 9908268-9908268 on Build GRCh38
6781412 c.998_999insG p.I334fs*14 Chr.4: 9908262-9908263 on Build GRCh38
4727092 c.978C>T p.Y326Y Chr.4: 9908283-9908283 on Build GRCh38
1226258 c.1003-1G>T p.? Chr.4: 9908346-9908346 on Build GRCh38
1226259 c.916-1G>T p.? Chr.4: 9908346-9908346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199577702 Chr.4: 9908374-9908374 on Build GRCh38
rs947261309 Chr.4: 9908436-9908436 on Build GRCh38
rs911278538 Chr.4: 9908104-9908104 on Build GRCh38
rs778725193 Chr.4: 9908334-9908334 on Build GRCh38
rs555736223 Chr.4: 9908407-9908407 on Build GRCh38
rs540525608 Chr.4: 9908338-9908338 on Build GRCh38
rs184307796 Chr.4: 9908069-9908069 on Build GRCh38
rs766506983 Chr.4: 9908188-9908188 on Build GRCh38
rs372542335 Chr.4: 9908315-9908315 on Build GRCh38
rs576420234 Chr.4: 9908298-9908298 on Build GRCh38
rs956680307 Chr.4: 9908412-9908412 on Build GRCh38
rs533262270 Chr.4: 9908312-9908312 on Build GRCh38
rs368289310 Chr.4: 9908303-9908303 on Build GRCh38
rs1007488758 Chr.4: 9908328-9908328 on Build GRCh38
rs756166192 Chr.4: 9908184-9908184 on Build GRCh38
rs1017206161 Chr.4: 9908114-9908114 on Build GRCh38
rs972959337 Chr.4: 9908468-9908468 on Build GRCh38
rs2280205 Chr.4: 9908299-9908299 on Build GRCh38
rs749169614 Chr.4: 9908244-9908244 on Build GRCh38
rs901171332 Chr.4: 9908339-9908339 on Build GRCh38
rs755483541 Chr.4: 9908204-9908204 on Build GRCh38
rs780600463 Chr.4: 9908198-9908199 on Build GRCh38
rs981869059 Chr.4: 9908179-9908179 on Build GRCh38
rs773908978 Chr.4: 9908357-9908357 on Build GRCh38
rs769414057 Chr.4: 9908235-9908235 on Build GRCh38
rs748093061 Chr.4: 9908386-9908386 on Build GRCh38
rs573658321 Chr.4: 9908410-9908410 on Build GRCh38
rs374834951 Chr.4: 9908282-9908282 on Build GRCh38
rs748838061 Chr.4: 9908344-9908344 on Build GRCh38
rs542920882 Chr.4: 9908267-9908267 on Build GRCh38
rs147545059 Chr.4: 9908168-9908168 on Build GRCh38
rs979538012 Chr.4: 9908403-9908403 on Build GRCh38
rs149428247 Chr.4: 9908107-9908107 on Build GRCh38
rs768651091 Chr.4: 9908259-9908259 on Build GRCh38
rs368382152 Chr.4: 9908390-9908390 on Build GRCh38
rs527710024 Chr.4: 9908452-9908452 on Build GRCh38
rs755833997 Chr.4: 9908322-9908322 on Build GRCh38
rs548989476 Chr.4: 9908457-9908457 on Build GRCh38
rs1028420506 Chr.4: 9908361-9908361 on Build GRCh38
rs1029822909 Chr.4: 9908041-9908041 on Build GRCh38
rs370426243 Chr.4: 9908125-9908125 on Build GRCh38
rs182011901 Chr.4: 9908203-9908203 on Build GRCh38
rs367692082 Chr.4: 9908263-9908263 on Build GRCh38
rs971714092 Chr.4: 9908120-9908120 on Build GRCh38
rs1028948134 Chr.4: 9908054-9908054 on Build GRCh38
rs998700573 Chr.4: 9908340-9908340 on Build GRCh38
rs560972980 Chr.4: 9908243-9908243 on Build GRCh38
rs557674068 Chr.4: 9908229-9908229 on Build GRCh38
rs961569993 Chr.4: 9908116-9908116 on Build GRCh38
rs189116938 Chr.4: 9908112-9908112 on Build GRCh38
rs371591260 Chr.4: 9908272-9908272 on Build GRCh38
rs779092846 Chr.4: 9908217-9908217 on Build GRCh38
rs116742917 Chr.4: 9908302-9908302 on Build GRCh38
rs998272902 Chr.4: 9908024-9908024 on Build GRCh38
rs2280204 Chr.4: 9908226-9908226 on Build GRCh38
rs779653371 Chr.4: 9908342-9908342 on Build GRCh38
rs935275677 Chr.4: 9908165-9908165 on Build GRCh38
rs756816874 Chr.4: 9908220-9908220 on Build GRCh38
rs772992516 Chr.4: 9908392-9908392 on Build GRCh38
rs778886684 Chr.4: 9908382-9908382 on Build GRCh38
rs754341360 Chr.4: 9908195-9908195 on Build GRCh38
rs889845657 Chr.4: 9908398-9908398 on Build GRCh38
rs968541386 Chr.4: 9908366-9908366 on Build GRCh38
rs992101223 Chr.4: 9908435-9908435 on Build GRCh38
rs917786422 Chr.4: 9908169-9908169 on Build GRCh38
rs754464408 Chr.4: 9908194-9908194 on Build GRCh38
rs267600301 Chr.4: 9908289-9908289 on Build GRCh38
rs985590238 Chr.4: 9908095-9908095 on Build GRCh38
rs753123621 Chr.4: 9908219-9908219 on Build GRCh38
rs766017898 Chr.4: 9908121-9908121 on Build GRCh38
rs760956353 Chr.4: 9908283-9908283 on Build GRCh38
rs750097140 Chr.4: 9908307-9908307 on Build GRCh38
rs947970473 Chr.4: 9908475-9908475 on Build GRCh38
rs955530365 Chr.4: 9908042-9908042 on Build GRCh38

View Full Product Details