Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17815005-17815527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAPG
Entrez Gene ID
64151
Gene Name
non-SMC condensin I complex subunit G
Gene Aliases
CAPG, CHCG, NY-MEL-3, YCG1
UniGene
Hs.567567
Gene Chromosome Location
Chr.4: 17810813-17844865 on Build GRCh38
Transcripts
NM_022346.4 NR_073124.1 XM_011513876.2 XM_017008543.1 XM_017008544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4506034 c.709C>T p.H237Y Chr.4: 17815292-17815292 on Build GRCh38
3337418 c.749T>G p.L250R Chr.4: 17815332-17815332 on Build GRCh38
3825562 c.747C>T p.L249L Chr.4: 17815330-17815330 on Build GRCh38
381000 c.718G>A p.A240T Chr.4: 17815301-17815301 on Build GRCh38
6198482 c.692T>C p.V231A Chr.4: 17815275-17815275 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs954724067 Chr.4: 17815085-17815085 on Build GRCh38
rs566543649 Chr.4: 17815088-17815088 on Build GRCh38
rs1037510683 Chr.4: 17815217-17815217 on Build GRCh38
rs968856880 Chr.4: 17815330-17815330 on Build GRCh38
rs959711359 Chr.4: 17815480-17815480 on Build GRCh38
rs141319271 Chr.4: 17815327-17815327 on Build GRCh38
rs552168639 Chr.4: 17815421-17815421 on Build GRCh38
rs761710022 Chr.4: 17815263-17815263 on Build GRCh38
rs996333450 Chr.4: 17815336-17815336 on Build GRCh38
rs774448552 Chr.4: 17815384-17815384 on Build GRCh38
rs768221062 Chr.4: 17815380-17815380 on Build GRCh38
rs145611655 Chr.4: 17815438-17815438 on Build GRCh38
rs759066389 Chr.4: 17815048-17815048 on Build GRCh38
rs35746704 Chr.4: 17815335-17815336 on Build GRCh38
rs776360576 Chr.4: 17815042-17815042 on Build GRCh38
rs779399137 Chr.4: 17815315-17815315 on Build GRCh38
rs769380659 Chr.4: 17815224-17815224 on Build GRCh38
rs1021710763 Chr.4: 17815328-17815328 on Build GRCh38
rs1029063321 Chr.4: 17815338-17815338 on Build GRCh38
rs751472911 Chr.4: 17815259-17815259 on Build GRCh38
rs767164631 Chr.4: 17815267-17815267 on Build GRCh38
rs528154518 Chr.4: 17815472-17815472 on Build GRCh38
rs769385019 Chr.4: 17815359-17815359 on Build GRCh38
rs922058537 Chr.4: 17815081-17815081 on Build GRCh38
rs762531812 Chr.4: 17815241-17815241 on Build GRCh38
rs775197767 Chr.4: 17815367-17815367 on Build GRCh38
rs576975932 Chr.4: 17815302-17815302 on Build GRCh38
rs759736095 Chr.4: 17815300-17815300 on Build GRCh38
rs1044830263 Chr.4: 17815251-17815251 on Build GRCh38
rs143738542 Chr.4: 17815307-17815307 on Build GRCh38
rs373948375 Chr.4: 17815242-17815242 on Build GRCh38
rs940622269 Chr.4: 17815206-17815206 on Build GRCh38
rs948207872 Chr.4: 17815194-17815194 on Build GRCh38
rs116257556 Chr.4: 17815227-17815227 on Build GRCh38
rs987903923 Chr.4: 17815101-17815101 on Build GRCh38
rs532155146 Chr.4: 17815105-17815105 on Build GRCh38
rs767456889 Chr.4: 17815386-17815386 on Build GRCh38
rs376754267 Chr.4: 17815306-17815306 on Build GRCh38
rs1052549274 Chr.4: 17815313-17815313 on Build GRCh38
rs907764443 Chr.4: 17815193-17815193 on Build GRCh38
rs772929707 Chr.4: 17815410-17815410 on Build GRCh38
rs762554057 Chr.4: 17815375-17815375 on Build GRCh38
rs1004115667 Chr.4: 17815433-17815433 on Build GRCh38
rs750688828 Chr.4: 17815281-17815281 on Build GRCh38
rs140365009 Chr.4: 17815458-17815458 on Build GRCh38
rs538080054 Chr.4: 17815430-17815430 on Build GRCh38
rs565803509 Chr.4: 17815431-17815431 on Build GRCh38
rs151066063 Chr.4: 17815325-17815325 on Build GRCh38
rs564722105 Chr.4: 17815076-17815076 on Build GRCh38
rs41268409 Chr.4: 17815385-17815385 on Build GRCh38
rs756329340 Chr.4: 17815295-17815295 on Build GRCh38
rs373215411 Chr.4: 17815416-17815416 on Build GRCh38

View Full Product Details