Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 24807245-24807751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC149
Entrez Gene ID
91050
Gene Name
coiled-coil domain containing 149
Gene Aliases
-
UniGene
Hs.106432
Gene Chromosome Location
Chr.4: 24806117-24980204 on Build GRCh38
Transcripts
NM_001130726.2 NM_173463.4 XM_005248210.3 XM_011513906.2 XM_011513907.2 XM_011513908.1 XM_017008827.1 XM_017008828.1

SNP Detail

SNP ID Chromosome Location
rs1032274355 Chr.4: 24807720-24807720 on Build GRCh38
rs890284242 Chr.4: 24807296-24807308 on Build GRCh38
rs187041258 Chr.4: 24807539-24807539 on Build GRCh38
rs192241451 Chr.4: 24807558-24807558 on Build GRCh38
rs528637377 Chr.4: 24807686-24807686 on Build GRCh38
rs959244854 Chr.4: 24807281-24807281 on Build GRCh38
rs1033977701 Chr.4: 24807670-24807670 on Build GRCh38
rs979357166 Chr.4: 24807274-24807274 on Build GRCh38
rs369681293 Chr.4: 24807642-24807642 on Build GRCh38
rs537706131 Chr.4: 24807311-24807311 on Build GRCh38
rs796392977 Chr.4: 24807651-24807651 on Build GRCh38
rs1048646767 Chr.4: 24807578-24807578 on Build GRCh38
rs1027894529 Chr.4: 24807615-24807615 on Build GRCh38
rs995122217 Chr.4: 24807592-24807592 on Build GRCh38
rs138285987 Chr.4: 24807455-24807455 on Build GRCh38
rs868191596 Chr.4: 24807714-24807714 on Build GRCh38
rs780229088 Chr.4: 24807549-24807549 on Build GRCh38
rs931511606 Chr.4: 24807554-24807554 on Build GRCh38
rs889921148 Chr.4: 24807597-24807597 on Build GRCh38
rs925981011 Chr.4: 24807418-24807418 on Build GRCh38
rs966352252 Chr.4: 24807664-24807664 on Build GRCh38
rs11936137 Chr.4: 24807433-24807433 on Build GRCh38
rs1041634358 Chr.4: 24807622-24807622 on Build GRCh38
rs369113763 Chr.4: 24807619-24807619 on Build GRCh38
rs917988887 Chr.4: 24807393-24807393 on Build GRCh38
rs952339238 Chr.4: 24807280-24807280 on Build GRCh38
rs905414768 Chr.4: 24807625-24807625 on Build GRCh38
rs558051335 Chr.4: 24807350-24807350 on Build GRCh38
rs114358954 Chr.4: 24807647-24807647 on Build GRCh38
rs959574942 Chr.4: 24807723-24807723 on Build GRCh38
rs182709518 Chr.4: 24807411-24807411 on Build GRCh38
rs901589937 Chr.4: 24807630-24807630 on Build GRCh38
rs925069915 Chr.4: 24807277-24807277 on Build GRCh38
rs890560670 Chr.4: 24807471-24807471 on Build GRCh38
rs752709535 Chr.4: 24807609-24807609 on Build GRCh38
rs897983260 Chr.4: 24807589-24807589 on Build GRCh38
rs1001158256 Chr.4: 24807676-24807676 on Build GRCh38
rs150813469 Chr.4: 24807454-24807454 on Build GRCh38
rs992678686 Chr.4: 24807324-24807324 on Build GRCh38

View Full Product Details