Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37588762-37589270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf19
Entrez Gene ID
55286
Gene Name
chromosome 4 open reading frame 19
Gene Aliases
-
UniGene
Hs.107527
Gene Chromosome Location
Chr.4: 37453454-37596146 on Build GRCh38
Transcripts
NM_001104629.1 NM_018302.2 XM_011513712.2 XM_011513713.2 XM_017008361.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3603454 c.4G>A p.G2R Chr.4: 37588871-37588871 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs996303878 Chr.4: 37589086-37589086 on Build GRCh38
rs748279506 Chr.4: 37589171-37589171 on Build GRCh38
rs1019117077 Chr.4: 37589096-37589096 on Build GRCh38
rs772345417 Chr.4: 37589192-37589192 on Build GRCh38
rs755097236 Chr.4: 37588985-37588985 on Build GRCh38
rs577504158 Chr.4: 37588923-37588923 on Build GRCh38
rs768923646 Chr.4: 37588927-37588927 on Build GRCh38
rs777473074 Chr.4: 37588837-37588837 on Build GRCh38
rs1009572288 Chr.4: 37589074-37589074 on Build GRCh38
rs969297552 Chr.4: 37589204-37589204 on Build GRCh38
rs112023101 Chr.4: 37588820-37588820 on Build GRCh38
rs987539089 Chr.4: 37589157-37589157 on Build GRCh38
rs770357113 Chr.4: 37588877-37588877 on Build GRCh38
rs367671308 Chr.4: 37588972-37588972 on Build GRCh38
rs1005133165 Chr.4: 37588998-37588999 on Build GRCh38
rs774602845 Chr.4: 37588937-37588937 on Build GRCh38
rs1029005527 Chr.4: 37589124-37589124 on Build GRCh38
rs10013825 Chr.4: 37589055-37589055 on Build GRCh38
rs2279994 Chr.4: 37588948-37588948 on Build GRCh38
rs371075759 Chr.4: 37588801-37588801 on Build GRCh38
rs375728507 Chr.4: 37588860-37588860 on Build GRCh38
rs760924069 Chr.4: 37589075-37589076 on Build GRCh38
rs34248425 Chr.4: 37589055-37589056 on Build GRCh38
rs761991267 Chr.4: 37588938-37588938 on Build GRCh38
rs776151990 Chr.4: 37588905-37588905 on Build GRCh38
rs946377344 Chr.4: 37589018-37589018 on Build GRCh38
rs993561785 Chr.4: 37589148-37589148 on Build GRCh38
rs934576778 Chr.4: 37588873-37588873 on Build GRCh38
rs191336554 Chr.4: 37588831-37588831 on Build GRCh38
rs574910225 Chr.4: 37588817-37588817 on Build GRCh38
rs372698549 Chr.4: 37588858-37588858 on Build GRCh38
rs767804267 Chr.4: 37588940-37588940 on Build GRCh38
rs376842002 Chr.4: 37588788-37588788 on Build GRCh38
rs954778819 Chr.4: 37589135-37589135 on Build GRCh38
rs564971504 Chr.4: 37588973-37588973 on Build GRCh38
rs893462076 Chr.4: 37588999-37588999 on Build GRCh38
rs989172373 Chr.4: 37588894-37588894 on Build GRCh38
rs1044126427 Chr.4: 37588944-37588944 on Build GRCh38
rs991482381 Chr.4: 37589178-37589178 on Build GRCh38
rs952121645 Chr.4: 37589172-37589172 on Build GRCh38
rs1023387587 Chr.4: 37589203-37589203 on Build GRCh38
rs1016963015 Chr.4: 37589016-37589016 on Build GRCh38
rs981926392 Chr.4: 37588921-37588921 on Build GRCh38
rs778985689 Chr.4: 37589103-37589103 on Build GRCh38
rs201083900 Chr.4: 37588849-37588849 on Build GRCh38
rs982469962 Chr.4: 37589227-37589227 on Build GRCh38
rs1050535615 Chr.4: 37589023-37589023 on Build GRCh38
rs867958625 Chr.4: 37589221-37589221 on Build GRCh38
rs899298933 Chr.4: 37589085-37589085 on Build GRCh38
rs889334675 Chr.4: 37589028-37589028 on Build GRCh38
rs749334732 Chr.4: 37588967-37588967 on Build GRCh38
rs746676926 Chr.4: 37588845-37588845 on Build GRCh38
rs752138870 Chr.4: 37588818-37588818 on Build GRCh38

View Full Product Details