Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 38796881-38797386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR1
Entrez Gene ID
7096
Gene Name
toll like receptor 1
Gene Aliases
CD281, TIL, TIL. LPRS5, rsc786
UniGene
Hs.621817
Gene Chromosome Location
Chr.4: 38787555-38805800 on Build GRCh38
Transcripts
NM_003263.3 XM_005262662.4 XM_011513742.2 XM_011513743.2 XM_011513744.2 XM_011513745.2 XM_017008571.1 XM_017008572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5995636 c.1472G>A p.S491N Chr.4: 38797360-38797360 on Build GRCh38
4124533 c.1803C>T p.C601C Chr.4: 38797029-38797029 on Build GRCh38
3409271 c.1852C>T p.Q618* Chr.4: 38796980-38796980 on Build GRCh38
4934008 c.1794C>T p.T598T Chr.4: 38797038-38797038 on Build GRCh38
5975127 c.1859G>A p.R620Q Chr.4: 38796973-38796973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202172776 Chr.4: 38796959-38796959 on Build GRCh38
rs190895342 Chr.4: 38796930-38796930 on Build GRCh38
rs111807776 Chr.4: 38797042-38797042 on Build GRCh38
rs760904516 Chr.4: 38796956-38796956 on Build GRCh38
rs773181138 Chr.4: 38796950-38796950 on Build GRCh38
rs753983618 Chr.4: 38797321-38797321 on Build GRCh38
rs112542063 Chr.4: 38797120-38797120 on Build GRCh38
rs767391851 Chr.4: 38797041-38797041 on Build GRCh38
rs1043697206 Chr.4: 38797261-38797261 on Build GRCh38
rs5743614 Chr.4: 38797314-38797314 on Build GRCh38
rs5743618 Chr.4: 38797027-38797027 on Build GRCh38
rs1041165741 Chr.4: 38797233-38797233 on Build GRCh38
rs140382462 Chr.4: 38797074-38797074 on Build GRCh38
rs755637224 Chr.4: 38797055-38797055 on Build GRCh38
rs763844691 Chr.4: 38797308-38797308 on Build GRCh38
rs767387683 Chr.4: 38797079-38797079 on Build GRCh38
rs747969548 Chr.4: 38797197-38797197 on Build GRCh38
rs370009768 Chr.4: 38797073-38797073 on Build GRCh38
rs776090836 Chr.4: 38797223-38797223 on Build GRCh38
rs762675323 Chr.4: 38797243-38797243 on Build GRCh38
rs772124432 Chr.4: 38796947-38796947 on Build GRCh38
rs749925721 Chr.4: 38797043-38797043 on Build GRCh38
rs377068130 Chr.4: 38797246-38797246 on Build GRCh38
rs752511598 Chr.4: 38796960-38796960 on Build GRCh38
rs368881067 Chr.4: 38797267-38797267 on Build GRCh38
rs5743619 Chr.4: 38796940-38796940 on Build GRCh38
rs200607815 Chr.4: 38797250-38797250 on Build GRCh38
rs200912870 Chr.4: 38796918-38796918 on Build GRCh38
rs568135901 Chr.4: 38797156-38797156 on Build GRCh38
rs778343132 Chr.4: 38797157-38797157 on Build GRCh38
rs781549406 Chr.4: 38796990-38796991 on Build GRCh38
rs761878041 Chr.4: 38796943-38796943 on Build GRCh38
rs764829877 Chr.4: 38797167-38797167 on Build GRCh38
rs756702006 Chr.4: 38797317-38797317 on Build GRCh38
rs774554376 Chr.4: 38797312-38797312 on Build GRCh38
rs769128800 Chr.4: 38796934-38796934 on Build GRCh38
rs772480838 Chr.4: 38797007-38797007 on Build GRCh38
rs372081210 Chr.4: 38796906-38796906 on Build GRCh38
rs774254323 Chr.4: 38797097-38797097 on Build GRCh38
rs755401705 Chr.4: 38797001-38797001 on Build GRCh38
rs200594956 Chr.4: 38797292-38797292 on Build GRCh38
rs760664665 Chr.4: 38797139-38797139 on Build GRCh38
rs200088165 Chr.4: 38796970-38796970 on Build GRCh38
rs786201036 Chr.4: 38797285-38797285 on Build GRCh38
rs143576765 Chr.4: 38797036-38797036 on Build GRCh38
rs564514530 Chr.4: 38797059-38797059 on Build GRCh38
rs770835697 Chr.4: 38797276-38797276 on Build GRCh38
rs779254336 Chr.4: 38797003-38797003 on Build GRCh38
rs953159848 Chr.4: 38797147-38797147 on Build GRCh38
rs150358789 Chr.4: 38797081-38797081 on Build GRCh38
rs201366752 Chr.4: 38796973-38796973 on Build GRCh38
rs757805205 Chr.4: 38797247-38797247 on Build GRCh38
rs200506989 Chr.4: 38797235-38797235 on Build GRCh38
rs148537628 Chr.4: 38796975-38796975 on Build GRCh38
rs143576719 Chr.4: 38797350-38797350 on Build GRCh38
rs764312573 Chr.4: 38797245-38797245 on Build GRCh38
rs187823248 Chr.4: 38797268-38797268 on Build GRCh38
rs755126139 Chr.4: 38797339-38797339 on Build GRCh38
rs961961000 Chr.4: 38797135-38797135 on Build GRCh38
rs369737596 Chr.4: 38796971-38796971 on Build GRCh38
rs746796724 Chr.4: 38797355-38797355 on Build GRCh38
rs771205452 Chr.4: 38797082-38797082 on Build GRCh38
rs759151280 Chr.4: 38797153-38797153 on Build GRCh38
rs750892111 Chr.4: 38797315-38797315 on Build GRCh38
rs5743617 Chr.4: 38797072-38797072 on Build GRCh38
rs755759428 Chr.4: 38796977-38796977 on Build GRCh38
rs748597526 Chr.4: 38797005-38797005 on Build GRCh38
rs775083312 Chr.4: 38797286-38797286 on Build GRCh38
rs5743615 Chr.4: 38797248-38797248 on Build GRCh38
rs769352235 Chr.4: 38796919-38796919 on Build GRCh38
rs372658963 Chr.4: 38797142-38797142 on Build GRCh38
rs530839614 Chr.4: 38797161-38797161 on Build GRCh38
rs776860609 Chr.4: 38797083-38797083 on Build GRCh38
rs907757231 Chr.4: 38796927-38796927 on Build GRCh38
rs151036585 Chr.4: 38797019-38797019 on Build GRCh38
rs374121599 Chr.4: 38797030-38797030 on Build GRCh38
rs770888784 Chr.4: 38797212-38797212 on Build GRCh38
rs786201035 Chr.4: 38797328-38797328 on Build GRCh38
rs764860555 Chr.4: 38797236-38797236 on Build GRCh38
rs759045186 Chr.4: 38797279-38797279 on Build GRCh38
rs778197468 Chr.4: 38797038-38797038 on Build GRCh38
rs137853173 Chr.4: 38797171-38797171 on Build GRCh38
rs753369710 Chr.4: 38797146-38797146 on Build GRCh38
rs771697062 Chr.4: 38797201-38797201 on Build GRCh38
rs892023944 Chr.4: 38796998-38796998 on Build GRCh38
rs770115894 Chr.4: 38797178-38797178 on Build GRCh38
rs753422993 Chr.4: 38797064-38797064 on Build GRCh38
rs773303713 Chr.4: 38797008-38797008 on Build GRCh38
rs1015047971 Chr.4: 38797345-38797345 on Build GRCh38
rs780186155 Chr.4: 38797257-38797257 on Build GRCh38
rs780197874 Chr.4: 38797194-38797194 on Build GRCh38
rs773313089 Chr.4: 38797087-38797087 on Build GRCh38
rs142855687 Chr.4: 38796986-38796986 on Build GRCh38
rs375868027 Chr.4: 38797068-38797068 on Build GRCh38
rs1014362334 Chr.4: 38797320-38797320 on Build GRCh38
rs760557774 Chr.4: 38797208-38797208 on Build GRCh38
rs762840851 Chr.4: 38797029-38797029 on Build GRCh38
rs761768106 Chr.4: 38797039-38797039 on Build GRCh38
rs5743616 Chr.4: 38797155-38797155 on Build GRCh38
rs201271982 Chr.4: 38797255-38797255 on Build GRCh38
rs149065118 Chr.4: 38797138-38797138 on Build GRCh38
rs770960887 Chr.4: 38797047-38797052 on Build GRCh38
rs370784389 Chr.4: 38797360-38797360 on Build GRCh38
rs749638831 Chr.4: 38796993-38796993 on Build GRCh38
rs749783321 Chr.4: 38797039-38797044 on Build GRCh38
rs375952631 Chr.4: 38797358-38797358 on Build GRCh38
rs55812771 Chr.4: 38797285-38797285 on Build GRCh38
rs192439045 Chr.4: 38797269-38797269 on Build GRCh38
rs199539854 Chr.4: 38797281-38797281 on Build GRCh38
rs575708724 Chr.4: 38797359-38797359 on Build GRCh38
rs776609872 Chr.4: 38797277-38797277 on Build GRCh38
rs137853172 Chr.4: 38797207-38797207 on Build GRCh38
rs770186713 Chr.4: 38797008-38797008 on Build GRCh38
rs144775976 Chr.4: 38796974-38796974 on Build GRCh38
rs757995061 Chr.4: 38796967-38796968 on Build GRCh38
rs138016555 Chr.4: 38797099-38797099 on Build GRCh38

View Full Product Details