Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 44622254-44622748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YIPF7
Entrez Gene ID
285525
Gene Name
Yip1 domain family member 7
Gene Aliases
FinGER9
UniGene
Hs.596000
Gene Chromosome Location
Chr.4: 44619578-44656868 on Build GRCh38
Transcripts
NM_182592.2 XM_011513679.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6824445 c.710T>C p.V237A Chr.4: 44622547-44622547 on Build GRCh38
6588888 c.744G>A p.K248K Chr.4: 44622513-44622513 on Build GRCh38
6512153 c.815G>T p.G272V Chr.4: 44622442-44622442 on Build GRCh38
2848260 c.696C>T p.I232I Chr.4: 44622561-44622561 on Build GRCh38
5541874 c.768G>A p.M256I Chr.4: 44622489-44622489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751920683 Chr.4: 44622448-44622448 on Build GRCh38
rs375303056 Chr.4: 44622484-44622484 on Build GRCh38
rs781210910 Chr.4: 44622613-44622613 on Build GRCh38
rs376355006 Chr.4: 44622574-44622574 on Build GRCh38
rs968198370 Chr.4: 44622434-44622434 on Build GRCh38
rs762406788 Chr.4: 44622580-44622580 on Build GRCh38
rs752318273 Chr.4: 44622432-44622432 on Build GRCh38
rs780373980 Chr.4: 44622536-44622536 on Build GRCh38
rs771882911 Chr.4: 44622483-44622483 on Build GRCh38
rs190628991 Chr.4: 44622401-44622401 on Build GRCh38
rs775238612 Chr.4: 44622572-44622572 on Build GRCh38
rs750736016 Chr.4: 44622585-44622585 on Build GRCh38
rs374504611 Chr.4: 44622378-44622378 on Build GRCh38
rs774290730 Chr.4: 44622386-44622386 on Build GRCh38
rs556877019 Chr.4: 44622348-44622348 on Build GRCh38
rs764742866 Chr.4: 44622436-44622436 on Build GRCh38
rs1021460617 Chr.4: 44622706-44622706 on Build GRCh38
rs372470281 Chr.4: 44622542-44622542 on Build GRCh38
rs1050319375 Chr.4: 44622671-44622671 on Build GRCh38
rs767984005 Chr.4: 44622531-44622531 on Build GRCh38
rs1048785036 Chr.4: 44622320-44622320 on Build GRCh38
rs367834984 Chr.4: 44622398-44622398 on Build GRCh38
rs1004954403 Chr.4: 44622333-44622333 on Build GRCh38
rs887570944 Chr.4: 44622325-44622325 on Build GRCh38
rs769281013 Chr.4: 44622570-44622570 on Build GRCh38
rs764639314 Chr.4: 44622426-44622426 on Build GRCh38
rs772114477 Chr.4: 44622400-44622400 on Build GRCh38
rs764981243 Chr.4: 44622511-44622511 on Build GRCh38
rs543227451 Chr.4: 44622682-44622682 on Build GRCh38
rs776467075 Chr.4: 44622502-44622502 on Build GRCh38
rs777627619 Chr.4: 44622543-44622543 on Build GRCh38
rs3213899 Chr.4: 44622335-44622335 on Build GRCh38
rs776258208 Chr.4: 44622554-44622554 on Build GRCh38
rs572638472 Chr.4: 44622375-44622375 on Build GRCh38
rs749279067 Chr.4: 44622537-44622537 on Build GRCh38
rs769770629 Chr.4: 44622383-44622383 on Build GRCh38
rs1022793585 Chr.4: 44622423-44622423 on Build GRCh38
rs901725366 Chr.4: 44622339-44622339 on Build GRCh38
rs201334946 Chr.4: 44622533-44622533 on Build GRCh38
rs1036033859 Chr.4: 44622338-44622338 on Build GRCh38
rs757159193 Chr.4: 44622602-44622602 on Build GRCh38
rs182873860 Chr.4: 44622624-44622624 on Build GRCh38
rs752118988 Chr.4: 44622530-44622530 on Build GRCh38
rs781501297 Chr.4: 44622454-44622454 on Build GRCh38
rs750979990 Chr.4: 44622466-44622466 on Build GRCh38
rs774063953 Chr.4: 44622576-44622576 on Build GRCh38
rs754098355 Chr.4: 44622590-44622590 on Build GRCh38
rs367859833 Chr.4: 44622504-44622504 on Build GRCh38
rs770921580 Chr.4: 44622551-44622551 on Build GRCh38
rs890765879 Chr.4: 44622722-44622722 on Build GRCh38
rs763712181 Chr.4: 44622575-44622575 on Build GRCh38
rs781585187 Chr.4: 44622380-44622380 on Build GRCh38
rs759027158 Chr.4: 44622424-44622424 on Build GRCh38
rs745725182 Chr.4: 44622561-44622561 on Build GRCh38
rs948247929 Chr.4: 44622295-44622295 on Build GRCh38
rs945765712 Chr.4: 44622709-44622709 on Build GRCh38
rs778278053 Chr.4: 44622374-44622374 on Build GRCh38
rs1050880606 Chr.4: 44622711-44622711 on Build GRCh38
rs756775517 Chr.4: 44622449-44622449 on Build GRCh38
rs760514470 Chr.4: 44622409-44622409 on Build GRCh38
rs746094408 Chr.4: 44622461-44622461 on Build GRCh38
rs753490719 Chr.4: 44622418-44622418 on Build GRCh38
rs151304151 Chr.4: 44622683-44622683 on Build GRCh38
rs575135983 Chr.4: 44622353-44622353 on Build GRCh38
rs762544227 Chr.4: 44622525-44622525 on Build GRCh38
rs868272187 Chr.4: 44622343-44622343 on Build GRCh38
rs532623434 Chr.4: 44622470-44622470 on Build GRCh38
rs778957035 Chr.4: 44622600-44622600 on Build GRCh38
rs761126595 Chr.4: 44622586-44622586 on Build GRCh38
rs186628194 Chr.4: 44622373-44622373 on Build GRCh38
rs765968750 Chr.4: 44622415-44622415 on Build GRCh38
rs750043407 Chr.4: 44622485-44622485 on Build GRCh38
rs371612304 Chr.4: 44622481-44622481 on Build GRCh38
rs923972388 Chr.4: 44622452-44622452 on Build GRCh38
rs369548094 Chr.4: 44622593-44622593 on Build GRCh38
rs890818367 Chr.4: 44622370-44622370 on Build GRCh38
rs745608509 Chr.4: 44622618-44622618 on Build GRCh38
rs746933998 Chr.4: 44622545-44622545 on Build GRCh38
rs76572894 Chr.4: 44622601-44622601 on Build GRCh38
rs774836488 Chr.4: 44622522-44622522 on Build GRCh38
rs373578037 Chr.4: 44622637-44622637 on Build GRCh38
rs768178405 Chr.4: 44622584-44622584 on Build GRCh38
rs565150538 Chr.4: 44622450-44622450 on Build GRCh38
rs764652928 Chr.4: 44622529-44622529 on Build GRCh38
rs542656446 Chr.4: 44622362-44622362 on Build GRCh38

View Full Product Details