Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 48421869-48422402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLAIN2
Entrez Gene ID
57606
Gene Name
SLAIN motif family member 2
Gene Aliases
KIAA1458
UniGene
Hs.479677
Gene Chromosome Location
Chr.4: 48341480-48426198 on Build GRCh38
Transcripts
NM_020846.1 XM_005248121.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7425599 c.1705G>T p.G569C Chr.4: 48422036-48422036 on Build GRCh38
4530655 c.1717G>A p.D573N Chr.4: 48422048-48422048 on Build GRCh38
3825931 c.1722C>T p.D574D Chr.4: 48422053-48422053 on Build GRCh38
5668232 c.1727G>T p.W576L Chr.4: 48422058-48422058 on Build GRCh38
6251196 c.1686G>T p.L562F Chr.4: 48422017-48422017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528532972 Chr.4: 48422169-48422169 on Build GRCh38
rs142036147 Chr.4: 48422140-48422140 on Build GRCh38
rs921550224 Chr.4: 48422116-48422116 on Build GRCh38
rs751133012 Chr.4: 48422095-48422095 on Build GRCh38
rs375209023 Chr.4: 48422020-48422020 on Build GRCh38
rs979066224 Chr.4: 48422043-48422043 on Build GRCh38
rs17609614 Chr.4: 48422041-48422041 on Build GRCh38
rs756687611 Chr.4: 48422102-48422102 on Build GRCh38
rs1055048825 Chr.4: 48422243-48422243 on Build GRCh38
rs769874345 Chr.4: 48422263-48422263 on Build GRCh38
rs368866915 Chr.4: 48421910-48421910 on Build GRCh38
rs775250210 Chr.4: 48422034-48422034 on Build GRCh38
rs749829346 Chr.4: 48422127-48422127 on Build GRCh38
rs545630093 Chr.4: 48421936-48421936 on Build GRCh38
rs975652143 Chr.4: 48422115-48422115 on Build GRCh38
rs898393203 Chr.4: 48421907-48421907 on Build GRCh38
rs147593344 Chr.4: 48422098-48422098 on Build GRCh38
rs765223535 Chr.4: 48422017-48422017 on Build GRCh38
rs1032126273 Chr.4: 48422359-48422359 on Build GRCh38
rs748411181 Chr.4: 48421964-48421964 on Build GRCh38
rs964369596 Chr.4: 48422366-48422366 on Build GRCh38
rs185822384 Chr.4: 48422155-48422155 on Build GRCh38
rs1038293733 Chr.4: 48422221-48422221 on Build GRCh38
rs866580874 Chr.4: 48421918-48421918 on Build GRCh38
rs780767585 Chr.4: 48422107-48422107 on Build GRCh38
rs896418437 Chr.4: 48422253-48422253 on Build GRCh38
rs759433637 Chr.4: 48421983-48421983 on Build GRCh38
rs375571223 Chr.4: 48422100-48422100 on Build GRCh38
rs756433215 Chr.4: 48422304-48422304 on Build GRCh38
rs902240055 Chr.4: 48422331-48422331 on Build GRCh38
rs766915580 Chr.4: 48422099-48422099 on Build GRCh38
rs564997493 Chr.4: 48421894-48421894 on Build GRCh38
rs940126466 Chr.4: 48422192-48422192 on Build GRCh38
rs202221339 Chr.4: 48422046-48422046 on Build GRCh38
rs974427912 Chr.4: 48422378-48422378 on Build GRCh38
rs571424541 Chr.4: 48422289-48422289 on Build GRCh38
rs756877539 Chr.4: 48422312-48422315 on Build GRCh38
rs761764785 Chr.4: 48422176-48422176 on Build GRCh38
rs1000622291 Chr.4: 48422333-48422333 on Build GRCh38
rs898510760 Chr.4: 48422231-48422231 on Build GRCh38
rs370753867 Chr.4: 48421970-48421970 on Build GRCh38
rs879045086 Chr.4: 48422156-48422156 on Build GRCh38
rs546630410 Chr.4: 48422285-48422285 on Build GRCh38
rs773325999 Chr.4: 48421981-48421981 on Build GRCh38
rs1020685191 Chr.4: 48422022-48422022 on Build GRCh38
rs34144414 Chr.4: 48422304-48422305 on Build GRCh38
rs372469639 Chr.4: 48422235-48422235 on Build GRCh38
rs373849837 Chr.4: 48421975-48421975 on Build GRCh38

View Full Product Details