Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 51863490-51864018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D4
Entrez Gene ID
23142
Gene Name
defective in cullin neddylation 1 domain containing 4
Gene Aliases
-
UniGene
Hs.605388
Gene Chromosome Location
Chr.4: 51843000-51916837 on Build GRCh38
Transcripts
NM_001040402.2 NM_001287755.1 NM_001287757.1 NM_015115.3 XM_005265731.3 XM_011534379.2 XM_011534380.1 XM_017007912.1 XM_017007913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5014423 c.109G>C p.D37H Chr.4: 51863682-51863682 on Build GRCh38
5014424 c.241G>C p.D81H Chr.4: 51863682-51863682 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371381672 Chr.4: 51863645-51863645 on Build GRCh38
rs190090640 Chr.4: 51863550-51863550 on Build GRCh38
rs182152245 Chr.4: 51863874-51863874 on Build GRCh38
rs931661315 Chr.4: 51863932-51863932 on Build GRCh38
rs187376469 Chr.4: 51863638-51863638 on Build GRCh38
rs577411590 Chr.4: 51863656-51863656 on Build GRCh38
rs758360065 Chr.4: 51863525-51863525 on Build GRCh38
rs200103544 Chr.4: 51863707-51863707 on Build GRCh38
rs957122769 Chr.4: 51863791-51863791 on Build GRCh38
rs1040283571 Chr.4: 51863930-51863930 on Build GRCh38
rs747729869 Chr.4: 51863632-51863632 on Build GRCh38
rs201327644 Chr.4: 51863747-51863747 on Build GRCh38
rs144145405 Chr.4: 51863845-51863845 on Build GRCh38
rs761835130 Chr.4: 51863750-51863750 on Build GRCh38
rs754834486 Chr.4: 51863541-51863544 on Build GRCh38
rs930732466 Chr.4: 51863905-51863905 on Build GRCh38
rs971157702 Chr.4: 51863815-51863815 on Build GRCh38
rs756033386 Chr.4: 51863535-51863535 on Build GRCh38
rs901754354 Chr.4: 51863931-51863931 on Build GRCh38
rs74856733 Chr.4: 51863711-51863711 on Build GRCh38
rs140179536 Chr.4: 51863689-51863689 on Build GRCh38
rs995847135 Chr.4: 51863787-51863787 on Build GRCh38
rs559714735 Chr.4: 51863754-51863754 on Build GRCh38
rs1034991569 Chr.4: 51863853-51863853 on Build GRCh38
rs1050079752 Chr.4: 51863938-51863938 on Build GRCh38
rs778385219 Chr.4: 51863629-51863629 on Build GRCh38
rs892888367 Chr.4: 51863942-51863942 on Build GRCh38
rs1036999038 Chr.4: 51863966-51863966 on Build GRCh38
rs1004776826 Chr.4: 51863770-51863770 on Build GRCh38
rs779971053 Chr.4: 51863618-51863618 on Build GRCh38
rs557459347 Chr.4: 51863646-51863646 on Build GRCh38
rs995685430 Chr.4: 51863548-51863548 on Build GRCh38
rs576150785 Chr.4: 51863786-51863786 on Build GRCh38
rs112280906 Chr.4: 51863572-51863572 on Build GRCh38
rs752749575 Chr.4: 51863675-51863675 on Build GRCh38
rs1018611608 Chr.4: 51863545-51863545 on Build GRCh38
rs758053478 Chr.4: 51863697-51863697 on Build GRCh38
rs771613997 Chr.4: 51863710-51863710 on Build GRCh38
rs932460169 Chr.4: 51863888-51863888 on Build GRCh38
rs551295687 Chr.4: 51863530-51863530 on Build GRCh38
rs984637381 Chr.4: 51863910-51863910 on Build GRCh38
rs921205038 Chr.4: 51863882-51863882 on Build GRCh38
rs753659465 Chr.4: 51863670-51863670 on Build GRCh38
rs766283956 Chr.4: 51863668-51863668 on Build GRCh38
rs565648088 Chr.4: 51863991-51863991 on Build GRCh38
rs777545860 Chr.4: 51863735-51863735 on Build GRCh38
rs907688429 Chr.4: 51863914-51863914 on Build GRCh38
rs749055274 Chr.4: 51863624-51863624 on Build GRCh38
rs1022389797 Chr.4: 51863832-51863832 on Build GRCh38
rs940028894 Chr.4: 51863698-51863698 on Build GRCh38
rs538406455 Chr.4: 51863878-51863878 on Build GRCh38
rs752870149 Chr.4: 51863522-51863522 on Build GRCh38
rs746266323 Chr.4: 51863727-51863727 on Build GRCh38
rs886385681 Chr.4: 51863768-51863768 on Build GRCh38
rs765517082 Chr.4: 51863647-51863647 on Build GRCh38
rs919933401 Chr.4: 51863972-51863972 on Build GRCh38
rs765524028 Chr.4: 51863517-51863517 on Build GRCh38
rs756087324 Chr.4: 51863664-51863664 on Build GRCh38
rs150604690 Chr.4: 51863852-51863852 on Build GRCh38
rs931559059 Chr.4: 51863993-51863993 on Build GRCh38
rs528533188 Chr.4: 51863781-51863781 on Build GRCh38
rs940592522 Chr.4: 51863929-51863929 on Build GRCh38
rs1040082550 Chr.4: 51863771-51863771 on Build GRCh38
rs996167160 Chr.4: 51863760-51863760 on Build GRCh38
rs763108332 Chr.4: 51863653-51863653 on Build GRCh38
rs561376361 Chr.4: 51863955-51863955 on Build GRCh38
rs750271606 Chr.4: 51863533-51863533 on Build GRCh38
rs373794586 Chr.4: 51863701-51863701 on Build GRCh38
rs573464883 Chr.4: 51863601-51863602 on Build GRCh38
rs377727373 Chr.4: 51863628-51863628 on Build GRCh38
rs192099923 Chr.4: 51863644-51863644 on Build GRCh38
rs1029197723 Chr.4: 51863790-51863790 on Build GRCh38
rs73246084 Chr.4: 51863601-51863601 on Build GRCh38
rs779893698 Chr.4: 51863542-51863542 on Build GRCh38
rs958413411 Chr.4: 51863592-51863592 on Build GRCh38
rs185189288 Chr.4: 51863974-51863974 on Build GRCh38
rs139701173 Chr.4: 51863990-51863990 on Build GRCh38
rs763152952 Chr.4: 51863740-51863740 on Build GRCh38
rs775534147 Chr.4: 51863652-51863652 on Build GRCh38
rs764348543 Chr.4: 51863742-51863742 on Build GRCh38
rs1011256614 Chr.4: 51863975-51863975 on Build GRCh38
rs1011517733 Chr.4: 51863809-51863809 on Build GRCh38
rs751656404 Chr.4: 51863659-51863659 on Build GRCh38
rs777228405 Chr.4: 51863722-51863722 on Build GRCh38
rs773779189 Chr.4: 51863641-51863641 on Build GRCh38
rs202136157 Chr.4: 51863728-51863728 on Build GRCh38
rs756035331 Chr.4: 51863731-51863731 on Build GRCh38
rs961026187 Chr.4: 51863871-51863871 on Build GRCh38
rs778637100 Chr.4: 51863685-51863685 on Build GRCh38

View Full Product Details